Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151121369-151121914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3D19
Entrez Gene ID
152503
Gene Name
SH3 domain containing 19
Gene Aliases
EBP, EVE1, Kryn, SH3P19
UniGene
Hs.744983
Gene Chromosome Location
Chr.4: 151120281-151228030 on Build GRCh38
Transcripts
NM_001009555.3 NM_001128923.1 NM_001128924.1 NM_001243349.1 XM_005262767.2 XM_011531646.1 XM_011531647.1 XM_011531648.1 XM_011531649.1 XM_017007778.1 XM_017007779.1 XM_017007780.1 XM_017007781.1 XM_017007782.1 XM_017007783.1

SNP Detail

SNP ID Chromosome Location
rs35415171 Chr.4: 151121436-151121439 on Build GRCh38
rs376379590 Chr.4: 151121441-151121441 on Build GRCh38
rs574920942 Chr.4: 151121559-151121559 on Build GRCh38
rs138385834 Chr.4: 151121783-151121783 on Build GRCh38
rs1037453390 Chr.4: 151121717-151121717 on Build GRCh38
rs78369655 Chr.4: 151121734-151121734 on Build GRCh38
rs978588384 Chr.4: 151121496-151121496 on Build GRCh38
rs1032811250 Chr.4: 151121803-151121803 on Build GRCh38
rs925786070 Chr.4: 151121504-151121504 on Build GRCh38
rs1046133 Chr.4: 151121511-151121511 on Build GRCh38
rs74392174 Chr.4: 151121440-151121440 on Build GRCh38
rs1014020947 Chr.4: 151121448-151121448 on Build GRCh38
rs999869522 Chr.4: 151121800-151121800 on Build GRCh38
rs1025365930 Chr.4: 151121461-151121461 on Build GRCh38
rs925154473 Chr.4: 151121477-151121477 on Build GRCh38
rs1051454627 Chr.4: 151121671-151121671 on Build GRCh38
rs536074075 Chr.4: 151121603-151121603 on Build GRCh38
rs564862749 Chr.4: 151121744-151121744 on Build GRCh38
rs557026009 Chr.4: 151121440-151121443 on Build GRCh38
rs147048191 Chr.4: 151121695-151121695 on Build GRCh38
rs967140419 Chr.4: 151121483-151121483 on Build GRCh38
rs755543058 Chr.4: 151121467-151121467 on Build GRCh38
rs1038581393 Chr.4: 151121715-151121715 on Build GRCh38
rs933208829 Chr.4: 151121653-151121653 on Build GRCh38
rs922647202 Chr.4: 151121590-151121590 on Build GRCh38
rs749533920 Chr.4: 151121826-151121826 on Build GRCh38
rs995699824 Chr.4: 151121772-151121772 on Build GRCh38
rs992175725 Chr.4: 151121433-151121433 on Build GRCh38
rs908645842 Chr.4: 151121673-151121673 on Build GRCh38
rs975314657 Chr.4: 151121887-151121887 on Build GRCh38
rs1019929509 Chr.4: 151121780-151121780 on Build GRCh38
rs1014366237 Chr.4: 151121766-151121766 on Build GRCh38
rs538489336 Chr.4: 151121615-151121615 on Build GRCh38
rs186691181 Chr.4: 151121543-151121543 on Build GRCh38
rs781596777 Chr.4: 151121475-151121475 on Build GRCh38
rs886094405 Chr.4: 151121677-151121677 on Build GRCh38
rs543951950 Chr.4: 151121871-151121871 on Build GRCh38
rs778069790 Chr.4: 151121792-151121792 on Build GRCh38
rs371351376 Chr.4: 151121575-151121575 on Build GRCh38
rs898702150 Chr.4: 151121747-151121747 on Build GRCh38
rs562639019 Chr.4: 151121879-151121879 on Build GRCh38
rs1046126 Chr.4: 151121531-151121531 on Build GRCh38
rs905326290 Chr.4: 151121723-151121723 on Build GRCh38
rs1056705222 Chr.4: 151121751-151121751 on Build GRCh38
rs1002456769 Chr.4: 151121727-151121727 on Build GRCh38
rs895496387 Chr.4: 151121756-151121756 on Build GRCh38
rs554479615 Chr.4: 151121613-151121613 on Build GRCh38
rs199706309 Chr.4: 151121439-151121439 on Build GRCh38

View Full Product Details