Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151715767-151716280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATB
Entrez Gene ID
5188
Gene Name
glutamyl-tRNA amidotransferase subunit B
Gene Aliases
HSPC199, PET112, PET112L
UniGene
Hs.119316
Gene Chromosome Location
Chr.4: 151669773-151761023 on Build GRCh38
Transcripts
NM_004564.2 XM_005263063.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744049 c.701C>A p.T234K Chr.4: 151716071-151716071 on Build GRCh38
1220206 c.695C>T p.A232V Chr.4: 151716077-151716077 on Build GRCh38
1052080 c.673G>A p.D225N Chr.4: 151716099-151716099 on Build GRCh38
5672860 c.721C>T p.L241L Chr.4: 151716051-151716051 on Build GRCh38
1220204 c.752C>T p.A251V Chr.4: 151716020-151716020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548831054 Chr.4: 151716219-151716219 on Build GRCh38
rs776318374 Chr.4: 151716167-151716168 on Build GRCh38
rs922252866 Chr.4: 151716055-151716055 on Build GRCh38
rs563623888 Chr.4: 151715921-151715921 on Build GRCh38
rs567338181 Chr.4: 151716236-151716236 on Build GRCh38
rs999947490 Chr.4: 151716059-151716059 on Build GRCh38
rs137948992 Chr.4: 151716111-151716111 on Build GRCh38
rs963964742 Chr.4: 151715998-151715998 on Build GRCh38
rs565097850 Chr.4: 151715852-151715853 on Build GRCh38
rs759714882 Chr.4: 151715855-151715856 on Build GRCh38
rs201506834 Chr.4: 151716101-151716101 on Build GRCh38
rs934227493 Chr.4: 151715908-151715908 on Build GRCh38
rs1038575117 Chr.4: 151715801-151715801 on Build GRCh38
rs775131201 Chr.4: 151716124-151716124 on Build GRCh38
rs890097258 Chr.4: 151715932-151715932 on Build GRCh38
rs757681630 Chr.4: 151716019-151716019 on Build GRCh38
rs766714240 Chr.4: 151715839-151715840 on Build GRCh38
rs371744323 Chr.4: 151715992-151715992 on Build GRCh38
rs145476419 Chr.4: 151716020-151716020 on Build GRCh38
rs760313567 Chr.4: 151716126-151716126 on Build GRCh38
rs764723952 Chr.4: 151716016-151716016 on Build GRCh38
rs569270398 Chr.4: 151716179-151716179 on Build GRCh38
rs763697147 Chr.4: 151715975-151715976 on Build GRCh38
rs759075070 Chr.4: 151715985-151715985 on Build GRCh38
rs761262245 Chr.4: 151716007-151716007 on Build GRCh38
rs543107277 Chr.4: 151716066-151716066 on Build GRCh38
rs769705880 Chr.4: 151716084-151716084 on Build GRCh38
rs553025822 Chr.4: 151715864-151715864 on Build GRCh38
rs138063181 Chr.4: 151716065-151716065 on Build GRCh38
rs550965719 Chr.4: 151716137-151716137 on Build GRCh38
rs192455594 Chr.4: 151715867-151715867 on Build GRCh38
rs377634370 Chr.4: 151715851-151715851 on Build GRCh38
rs531883894 Chr.4: 151715836-151715843 on Build GRCh38
rs201975276 Chr.4: 151716114-151716114 on Build GRCh38
rs151106436 Chr.4: 151715789-151715789 on Build GRCh38
rs547158537 Chr.4: 151716100-151716100 on Build GRCh38
rs941448056 Chr.4: 151716109-151716109 on Build GRCh38
rs905424869 Chr.4: 151715823-151715823 on Build GRCh38
rs750682382 Chr.4: 151716058-151716058 on Build GRCh38
rs35487764 Chr.4: 151716076-151716076 on Build GRCh38
rs1020986868 Chr.4: 151715844-151715844 on Build GRCh38
rs765921802 Chr.4: 151715924-151715927 on Build GRCh38
rs1038511892 Chr.4: 151716168-151716168 on Build GRCh38
rs760124470 Chr.4: 151715853-151715855 on Build GRCh38
rs536765841 Chr.4: 151716209-151716209 on Build GRCh38
rs766981648 Chr.4: 151715836-151715839 on Build GRCh38
rs761423190 Chr.4: 151716169-151716170 on Build GRCh38
rs993137030 Chr.4: 151716069-151716069 on Build GRCh38
rs941459403 Chr.4: 151715850-151715850 on Build GRCh38
rs934197533 Chr.4: 151716206-151716206 on Build GRCh38
rs770464398 Chr.4: 151715967-151715967 on Build GRCh38
rs777812349 Chr.4: 151716086-151716086 on Build GRCh38
rs749000969 Chr.4: 151716099-151716099 on Build GRCh38
rs909941935 Chr.4: 151715825-151715825 on Build GRCh38
rs774088913 Chr.4: 151715976-151715976 on Build GRCh38
rs34326468 Chr.4: 151716190-151716191 on Build GRCh38
rs767155504 Chr.4: 151715994-151715994 on Build GRCh38
rs1051378562 Chr.4: 151715923-151715923 on Build GRCh38
rs764794594 Chr.4: 151716180-151716181 on Build GRCh38
rs879368498 Chr.4: 151715852-151715853 on Build GRCh38
rs1056436187 Chr.4: 151715835-151715836 on Build GRCh38
rs1033278810 Chr.4: 151715912-151715912 on Build GRCh38
rs909890997 Chr.4: 151716096-151716096 on Build GRCh38
rs545462506 Chr.4: 151715903-151715903 on Build GRCh38
rs753191998 Chr.4: 151716005-151716005 on Build GRCh38
rs904239290 Chr.4: 151716051-151716051 on Build GRCh38
rs374430639 Chr.4: 151716239-151716239 on Build GRCh38
rs150296974 Chr.4: 151716077-151716077 on Build GRCh38

View Full Product Details