Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152880823-152881340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625237 c.442G>T p.G148C Chr.4: 152880993-152880993 on Build GRCh38
3600957 c.524T>C p.I175T Chr.4: 152881075-152881075 on Build GRCh38
1194676 c.631C>T p.H211Y Chr.4: 152881182-152881182 on Build GRCh38
447505 c.507G>C p.K169N Chr.4: 152881058-152881058 on Build GRCh38
6535466 c.513A>G p.K171K Chr.4: 152881064-152881064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750222943 Chr.4: 152881235-152881235 on Build GRCh38
rs762435930 Chr.4: 152881138-152881138 on Build GRCh38
rs140439590 Chr.4: 152880983-152880983 on Build GRCh38
rs756154277 Chr.4: 152880941-152880941 on Build GRCh38
rs972277548 Chr.4: 152880861-152880861 on Build GRCh38
rs749527148 Chr.4: 152880997-152880997 on Build GRCh38
rs533435202 Chr.4: 152881094-152881094 on Build GRCh38
rs764586733 Chr.4: 152881013-152881013 on Build GRCh38
rs1057078404 Chr.4: 152881210-152881210 on Build GRCh38
rs201384562 Chr.4: 152881035-152881035 on Build GRCh38
rs746533086 Chr.4: 152881122-152881122 on Build GRCh38
rs749315002 Chr.4: 152881088-152881088 on Build GRCh38
rs767677232 Chr.4: 152881206-152881206 on Build GRCh38
rs1013347638 Chr.4: 152880887-152880887 on Build GRCh38
rs949290195 Chr.4: 152881253-152881253 on Build GRCh38
rs770818342 Chr.4: 152880957-152880957 on Build GRCh38
rs73865265 Chr.4: 152881091-152881091 on Build GRCh38
rs73865262 Chr.4: 152880853-152880853 on Build GRCh38
rs896409428 Chr.4: 152880860-152880860 on Build GRCh38
rs780296505 Chr.4: 152880924-152880925 on Build GRCh38
rs749052109 Chr.4: 152880955-152880955 on Build GRCh38
rs777560559 Chr.4: 152881086-152881086 on Build GRCh38
rs757241558 Chr.4: 152881212-152881212 on Build GRCh38
rs80127385 Chr.4: 152880908-152880908 on Build GRCh38
rs753303689 Chr.4: 152881304-152881304 on Build GRCh38
rs778530610 Chr.4: 152881217-152881217 on Build GRCh38
rs753696765 Chr.4: 152881207-152881207 on Build GRCh38
rs758720563 Chr.4: 152881044-152881044 on Build GRCh38
rs924695460 Chr.4: 152881119-152881119 on Build GRCh38
rs919210257 Chr.4: 152880889-152880889 on Build GRCh38
rs745573991 Chr.4: 152880969-152880969 on Build GRCh38
rs1016466593 Chr.4: 152880961-152880961 on Build GRCh38
rs73865263 Chr.4: 152881072-152881072 on Build GRCh38
rs762737649 Chr.4: 152880948-152880948 on Build GRCh38
rs191714736 Chr.4: 152881155-152881155 on Build GRCh38
rs111887580 Chr.4: 152881301-152881301 on Build GRCh38
rs776995083 Chr.4: 152881134-152881134 on Build GRCh38
rs776993971 Chr.4: 152881017-152881017 on Build GRCh38
rs909877715 Chr.4: 152880967-152880967 on Build GRCh38
rs768210406 Chr.4: 152881133-152881133 on Build GRCh38
rs371990041 Chr.4: 152881028-152881028 on Build GRCh38
rs765474391 Chr.4: 152881144-152881144 on Build GRCh38
rs777645718 Chr.4: 152880945-152880945 on Build GRCh38
rs948023789 Chr.4: 152880970-152880970 on Build GRCh38
rs377259978 Chr.4: 152881100-152881100 on Build GRCh38
rs34460189 Chr.4: 152880974-152880975 on Build GRCh38
rs551587540 Chr.4: 152881099-152881099 on Build GRCh38
rs892744749 Chr.4: 152881079-152881079 on Build GRCh38
rs766723304 Chr.4: 152881158-152881158 on Build GRCh38
rs1044986726 Chr.4: 152881275-152881275 on Build GRCh38
rs767845994 Chr.4: 152881303-152881303 on Build GRCh38
rs773872530 Chr.4: 152880968-152880968 on Build GRCh38
rs369258055 Chr.4: 152881176-152881176 on Build GRCh38
rs755104357 Chr.4: 152881190-152881190 on Build GRCh38
rs545602334 Chr.4: 152880851-152880851 on Build GRCh38
rs766529190 Chr.4: 152881046-152881046 on Build GRCh38
rs866277840 Chr.4: 152881005-152881005 on Build GRCh38
rs144212409 Chr.4: 152881042-152881042 on Build GRCh38
rs775001196 Chr.4: 152880996-152880996 on Build GRCh38
rs748052903 Chr.4: 152880937-152880937 on Build GRCh38
rs930537286 Chr.4: 152880898-152880898 on Build GRCh38
rs146568129 Chr.4: 152881051-152881051 on Build GRCh38
rs576572614 Chr.4: 152880880-152880880 on Build GRCh38
rs934640176 Chr.4: 152881121-152881121 on Build GRCh38
rs774912995 Chr.4: 152881106-152881106 on Build GRCh38
rs773658881 Chr.4: 152881146-152881146 on Build GRCh38
rs780334178 Chr.4: 152880929-152880929 on Build GRCh38

View Full Product Details