Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10286260-10286758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMBL
Entrez Gene ID
134147
Gene Name
carboxymethylenebutenolidase homolog
Gene Aliases
JS-1
UniGene
Hs.192586
Gene Chromosome Location
Chr.5: 10277595-10308056 on Build GRCh38
Transcripts
NM_138809.3 XM_017009034.1 XM_017009035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5466242 c.339C>A p.I113I Chr.5: 10286481-10286481 on Build GRCh38
735160 c.333T>C p.S111S Chr.5: 10286487-10286487 on Build GRCh38
7235000 c.460G>T p.V154F Chr.5: 10286360-10286360 on Build GRCh38
2990062 c.329T>C p.I110T Chr.5: 10286491-10286491 on Build GRCh38
4849404 c.369C>T p.A123A Chr.5: 10286451-10286451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942514871 Chr.5: 10286515-10286515 on Build GRCh38
rs773199941 Chr.5: 10286416-10286416 on Build GRCh38
rs769211982 Chr.5: 10286393-10286393 on Build GRCh38
rs762378958 Chr.5: 10286327-10286327 on Build GRCh38
rs895996466 Chr.5: 10286660-10286660 on Build GRCh38
rs13180748 Chr.5: 10286630-10286630 on Build GRCh38
rs777716530 Chr.5: 10286443-10286443 on Build GRCh38
rs1038513366 Chr.5: 10286606-10286606 on Build GRCh38
rs1032626650 Chr.5: 10286686-10286686 on Build GRCh38
rs979254805 Chr.5: 10286497-10286497 on Build GRCh38
rs146696783 Chr.5: 10286437-10286437 on Build GRCh38
rs996192791 Chr.5: 10286718-10286718 on Build GRCh38
rs114991197 Chr.5: 10286296-10286296 on Build GRCh38
rs386685404 Chr.5: 10286562-10286564 on Build GRCh38
rs745576546 Chr.5: 10286465-10286465 on Build GRCh38
rs1002379153 Chr.5: 10286624-10286624 on Build GRCh38
rs763442330 Chr.5: 10286529-10286529 on Build GRCh38
rs750463180 Chr.5: 10286338-10286338 on Build GRCh38
rs528008731 Chr.5: 10286286-10286286 on Build GRCh38
rs1034529005 Chr.5: 10286404-10286404 on Build GRCh38
rs756034402 Chr.5: 10286440-10286440 on Build GRCh38
rs375310155 Chr.5: 10286392-10286392 on Build GRCh38
rs372248737 Chr.5: 10286483-10286483 on Build GRCh38
rs78337728 Chr.5: 10286564-10286564 on Build GRCh38
rs761072004 Chr.5: 10286317-10286317 on Build GRCh38
rs35489000 Chr.5: 10286356-10286356 on Build GRCh38
rs868855509 Chr.5: 10286402-10286402 on Build GRCh38
rs777318390 Chr.5: 10286324-10286324 on Build GRCh38
rs922458330 Chr.5: 10286544-10286544 on Build GRCh38
rs370274529 Chr.5: 10286706-10286706 on Build GRCh38
rs557128485 Chr.5: 10286596-10286596 on Build GRCh38
rs770272540 Chr.5: 10286415-10286415 on Build GRCh38
rs999727892 Chr.5: 10286670-10286670 on Build GRCh38
rs115891820 Chr.5: 10286609-10286609 on Build GRCh38
rs139472834 Chr.5: 10286433-10286433 on Build GRCh38
rs886772486 Chr.5: 10286567-10286567 on Build GRCh38
rs201454015 Chr.5: 10286476-10286476 on Build GRCh38
rs144167278 Chr.5: 10286435-10286435 on Build GRCh38
rs777042773 Chr.5: 10286403-10286403 on Build GRCh38
rs376709839 Chr.5: 10286458-10286458 on Build GRCh38
rs763043668 Chr.5: 10286419-10286419 on Build GRCh38
rs1001737071 Chr.5: 10286397-10286397 on Build GRCh38
rs766412476 Chr.5: 10286420-10286420 on Build GRCh38
rs534928794 Chr.5: 10286438-10286438 on Build GRCh38
rs926083642 Chr.5: 10286514-10286514 on Build GRCh38
rs34487157 Chr.5: 10286372-10286372 on Build GRCh38
rs754803877 Chr.5: 10286359-10286359 on Build GRCh38
rs1014507874 Chr.5: 10286676-10286676 on Build GRCh38
rs756019209 Chr.5: 10286367-10286367 on Build GRCh38
rs764532159 Chr.5: 10286321-10286321 on Build GRCh38
rs753695106 Chr.5: 10286445-10286445 on Build GRCh38
rs765293407 Chr.5: 10286336-10286336 on Build GRCh38
rs902720074 Chr.5: 10286643-10286643 on Build GRCh38
rs186812717 Chr.5: 10286315-10286315 on Build GRCh38
rs766552139 Chr.5: 10286354-10286354 on Build GRCh38
rs759738230 Chr.5: 10286436-10286436 on Build GRCh38
rs745485656 Chr.5: 10286386-10286386 on Build GRCh38
rs773822133 Chr.5: 10286494-10286494 on Build GRCh38
rs778758411 Chr.5: 10286382-10286382 on Build GRCh38
rs756867942 Chr.5: 10286451-10286451 on Build GRCh38
rs758556381 Chr.5: 10286342-10286342 on Build GRCh38
rs958243044 Chr.5: 10286703-10286703 on Build GRCh38
rs762771059 Chr.5: 10286575-10286575 on Build GRCh38
rs146320422 Chr.5: 10286376-10286376 on Build GRCh38
rs772915744 Chr.5: 10286315-10286315 on Build GRCh38
rs538689219 Chr.5: 10286411-10286411 on Build GRCh38
rs931028118 Chr.5: 10286588-10286588 on Build GRCh38
rs770229561 Chr.5: 10286490-10286490 on Build GRCh38
rs1005706352 Chr.5: 10286364-10286364 on Build GRCh38
rs780957157 Chr.5: 10286360-10286360 on Build GRCh38
rs142499932 Chr.5: 10286361-10286361 on Build GRCh38
rs771794082 Chr.5: 10286466-10286466 on Build GRCh38
rs865859684 Chr.5: 10286410-10286410 on Build GRCh38
rs539773959 Chr.5: 10286698-10286698 on Build GRCh38
rs777150555 Chr.5: 10286369-10286369 on Build GRCh38
rs753333458 Chr.5: 10286634-10286635 on Build GRCh38
rs371292499 Chr.5: 10286481-10286481 on Build GRCh38
rs772080555 Chr.5: 10286308-10286308 on Build GRCh38
rs75142029 Chr.5: 10286562-10286562 on Build GRCh38
rs762763030 Chr.5: 10286375-10286377 on Build GRCh38

View Full Product Details