Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35057942-35058454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRLR
Entrez Gene ID
5618
Gene Name
prolactin receptor
Gene Aliases
HPRL, MFAB, hPRLrI
UniGene
Hs.368587
Gene Chromosome Location
Chr.5: 35048295-35230589 on Build GRCh38
Transcripts
NM_000949.6 NM_001204314.2 NM_001204316.1 NM_001204317.1 NM_001204318.1 NR_037910.1 XM_006714484.2 XM_011514068.2 XM_011514069.2 XM_017009645.1 XM_017009646.1

SNP Detail

SNP ID Chromosome Location
rs1029088603 Chr.5: 35058093-35058093 on Build GRCh38
rs62355477 Chr.5: 35058379-35058379 on Build GRCh38
rs757180317 Chr.5: 35058051-35058051 on Build GRCh38
rs954852770 Chr.5: 35058423-35058423 on Build GRCh38
rs1030868575 Chr.5: 35058042-35058042 on Build GRCh38
rs1009224275 Chr.5: 35058396-35058396 on Build GRCh38
rs968721362 Chr.5: 35058055-35058055 on Build GRCh38
rs899939354 Chr.5: 35058310-35058310 on Build GRCh38
rs899714736 Chr.5: 35058391-35058391 on Build GRCh38
rs531705350 Chr.5: 35058190-35058190 on Build GRCh38
rs1015181623 Chr.5: 35058148-35058148 on Build GRCh38
rs538455406 Chr.5: 35058249-35058252 on Build GRCh38
rs1043774789 Chr.5: 35058308-35058308 on Build GRCh38
rs973148080 Chr.5: 35058194-35058194 on Build GRCh38
rs1967132 Chr.5: 35058105-35058105 on Build GRCh38
rs567450085 Chr.5: 35058217-35058217 on Build GRCh38
rs906184034 Chr.5: 35058283-35058283 on Build GRCh38
rs569337817 Chr.5: 35058056-35058056 on Build GRCh38
rs996756514 Chr.5: 35058418-35058418 on Build GRCh38
rs553990549 Chr.5: 35058195-35058195 on Build GRCh38
rs946907467 Chr.5: 35058306-35058306 on Build GRCh38
rs914051425 Chr.5: 35058305-35058305 on Build GRCh38
rs773739251 Chr.5: 35058111-35058111 on Build GRCh38
rs928197803 Chr.5: 35058270-35058270 on Build GRCh38
rs996781320 Chr.5: 35058334-35058334 on Build GRCh38
rs939606705 Chr.5: 35058271-35058271 on Build GRCh38
rs987954858 Chr.5: 35058110-35058110 on Build GRCh38
rs1044811548 Chr.5: 35057992-35057992 on Build GRCh38
rs879572344 Chr.5: 35058050-35058050 on Build GRCh38
rs575662535 Chr.5: 35058263-35058263 on Build GRCh38
rs149523165 Chr.5: 35058419-35058419 on Build GRCh38
rs939154527 Chr.5: 35058286-35058286 on Build GRCh38
rs973637616 Chr.5: 35058218-35058218 on Build GRCh38
rs1052597896 Chr.5: 35058303-35058303 on Build GRCh38
rs571413123 Chr.5: 35058170-35058170 on Build GRCh38
rs531822938 Chr.5: 35058082-35058082 on Build GRCh38
rs947694361 Chr.5: 35057981-35057981 on Build GRCh38
rs535813489 Chr.5: 35058228-35058228 on Build GRCh38
rs559732516 Chr.5: 35058139-35058139 on Build GRCh38
rs1053034121 Chr.5: 35058000-35058000 on Build GRCh38
rs375941619 Chr.5: 35058048-35058048 on Build GRCh38
rs555725931 Chr.5: 35058252-35058252 on Build GRCh38
rs892330802 Chr.5: 35058353-35058353 on Build GRCh38
rs920821762 Chr.5: 35058224-35058224 on Build GRCh38
rs369106628 Chr.5: 35058078-35058078 on Build GRCh38
rs578093555 Chr.5: 35058398-35058398 on Build GRCh38
rs1003795237 Chr.5: 35058019-35058019 on Build GRCh38
rs908058210 Chr.5: 35058136-35058136 on Build GRCh38
rs920367279 Chr.5: 35058232-35058232 on Build GRCh38
rs558410023 Chr.5: 35058390-35058390 on Build GRCh38
rs906253281 Chr.5: 35058011-35058011 on Build GRCh38
rs940841277 Chr.5: 35058172-35058172 on Build GRCh38

View Full Product Details