Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 56922566-56923103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIER3
Entrez Gene ID
166968
Gene Name
MIER family member 3
Gene Aliases
-
UniGene
Hs.657594
Gene Chromosome Location
Chr.5: 56919602-56952130 on Build GRCh38
Transcripts
NM_001297598.1 NM_001297599.1 NM_152622.4 XM_011543216.1 XM_011543217.1 XM_017009149.1 NM_001171990.2 NR_136555.1

Gene Symbol
SETD9
Entrez Gene ID
133383
Gene Name
SET domain containing 9
Gene Aliases
C5orf35
UniGene
Hs.85950
Gene Chromosome Location
Chr.5: 56909260-56928900 on Build GRCh38
Transcripts
NM_001297598.1 NM_001297599.1 NM_152622.4 XM_011543216.1 XM_011543217.1 XM_017009149.1 NM_001171990.2 NR_136555.1

SNP Detail

SNP ID Chromosome Location
rs970690407 Chr.5: 56922835-56922837 on Build GRCh38
rs1026762855 Chr.5: 56922656-56922656 on Build GRCh38
rs982213804 Chr.5: 56922851-56922851 on Build GRCh38
rs533487516 Chr.5: 56922948-56922948 on Build GRCh38
rs943619660 Chr.5: 56922957-56922957 on Build GRCh38
rs994656439 Chr.5: 56922783-56922783 on Build GRCh38
rs1014196823 Chr.5: 56922720-56922720 on Build GRCh38
rs863838 Chr.5: 56922724-56922724 on Build GRCh38
rs930746475 Chr.5: 56923050-56923050 on Build GRCh38
rs983732715 Chr.5: 56922798-56922798 on Build GRCh38
rs979894086 Chr.5: 56922772-56922772 on Build GRCh38
rs969885244 Chr.5: 56922732-56922732 on Build GRCh38
rs912112646 Chr.5: 56922956-56922956 on Build GRCh38
rs1008613716 Chr.5: 56922663-56922663 on Build GRCh38
rs1039211597 Chr.5: 56922999-56922999 on Build GRCh38
rs550603957 Chr.5: 56922826-56922826 on Build GRCh38
rs988568076 Chr.5: 56922860-56922860 on Build GRCh38
rs994923555 Chr.5: 56922655-56922655 on Build GRCh38
rs899318506 Chr.5: 56923022-56923022 on Build GRCh38
rs989682915 Chr.5: 56922867-56922867 on Build GRCh38
rs749622190 Chr.5: 56922607-56922607 on Build GRCh38
rs933635599 Chr.5: 56922910-56922910 on Build GRCh38
rs555903580 Chr.5: 56922612-56922612 on Build GRCh38
rs899284666 Chr.5: 56922632-56922632 on Build GRCh38
rs950700626 Chr.5: 56922797-56922797 on Build GRCh38
rs1032844838 Chr.5: 56922787-56922787 on Build GRCh38
rs561850246 Chr.5: 56922765-56922765 on Build GRCh38
rs527497108 Chr.5: 56922700-56922700 on Build GRCh38
rs957384542 Chr.5: 56922796-56922796 on Build GRCh38
rs938065091 Chr.5: 56922858-56922858 on Build GRCh38
rs774889097 Chr.5: 56923078-56923078 on Build GRCh38
rs779726499 Chr.5: 56923079-56923081 on Build GRCh38
rs574212172 Chr.5: 56922718-56922718 on Build GRCh38
rs748179415 Chr.5: 56922978-56922978 on Build GRCh38
rs551932633 Chr.5: 56923057-56923057 on Build GRCh38
rs148014477 Chr.5: 56922819-56922819 on Build GRCh38
rs926721089 Chr.5: 56922857-56922857 on Build GRCh38
rs774353674 Chr.5: 56922829-56922829 on Build GRCh38
rs912912707 Chr.5: 56922871-56922871 on Build GRCh38
rs1015605110 Chr.5: 56922801-56922801 on Build GRCh38
rs886421212 Chr.5: 56922662-56922662 on Build GRCh38
rs569533394 Chr.5: 56922933-56922933 on Build GRCh38

View Full Product Details