Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64804135-64804634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWC27
Entrez Gene ID
10283
Gene Name
CWC27 spliceosome associated protein homolog
Gene Aliases
NY-CO-10, SDCCAG-10, SDCCAG10
UniGene
Hs.371372
Gene Chromosome Location
Chr.5: 64768918-65018763 on Build GRCh38
Transcripts
NM_001297644.1 NM_001297645.1 NM_001318000.1 NM_005869.3 XM_011543095.2 XM_011543096.1 XM_017008943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7183120 c.811G>A p.E271K Chr.5: 64804259-64804259 on Build GRCh38
3345960 c.851G>T p.R284I Chr.5: 64804299-64804299 on Build GRCh38
5758626 c.935G>A p.R312H Chr.5: 64804383-64804383 on Build GRCh38
266329 c.828T>C p.D276D Chr.5: 64804276-64804276 on Build GRCh38
194049 c.884C>T p.A295V Chr.5: 64804332-64804332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000790319 Chr.5: 64804223-64804223 on Build GRCh38
rs146471959 Chr.5: 64804358-64804358 on Build GRCh38
rs551945418 Chr.5: 64804326-64804326 on Build GRCh38
rs971348778 Chr.5: 64804167-64804167 on Build GRCh38
rs149929520 Chr.5: 64804332-64804332 on Build GRCh38
rs752999264 Chr.5: 64804285-64804285 on Build GRCh38
rs141017155 Chr.5: 64804368-64804368 on Build GRCh38
rs747116169 Chr.5: 64804360-64804360 on Build GRCh38
rs755632501 Chr.5: 64804324-64804324 on Build GRCh38
rs894760771 Chr.5: 64804465-64804465 on Build GRCh38
rs181875545 Chr.5: 64804585-64804585 on Build GRCh38
rs747909538 Chr.5: 64804340-64804340 on Build GRCh38
rs369432186 Chr.5: 64804266-64804266 on Build GRCh38
rs191313541 Chr.5: 64804490-64804490 on Build GRCh38
rs778866747 Chr.5: 64804333-64804333 on Build GRCh38
rs539579911 Chr.5: 64804541-64804541 on Build GRCh38
rs771390228 Chr.5: 64804264-64804264 on Build GRCh38
rs568361888 Chr.5: 64804482-64804482 on Build GRCh38
rs776503579 Chr.5: 64804372-64804372 on Build GRCh38
rs780013474 Chr.5: 64804201-64804201 on Build GRCh38
rs778118563 Chr.5: 64804349-64804349 on Build GRCh38
rs765409962 Chr.5: 64804536-64804536 on Build GRCh38
rs200712636 Chr.5: 64804293-64804293 on Build GRCh38
rs760170656 Chr.5: 64804409-64804409 on Build GRCh38
rs971401408 Chr.5: 64804533-64804533 on Build GRCh38
rs765918730 Chr.5: 64804429-64804429 on Build GRCh38
rs1024961855 Chr.5: 64804310-64804310 on Build GRCh38
rs1013589386 Chr.5: 64804495-64804495 on Build GRCh38
rs973595470 Chr.5: 64804434-64804434 on Build GRCh38
rs936526728 Chr.5: 64804550-64804550 on Build GRCh38
rs919537940 Chr.5: 64804441-64804441 on Build GRCh38
rs759880526 Chr.5: 64804273-64804273 on Build GRCh38
rs201403830 Chr.5: 64804291-64804291 on Build GRCh38
rs563411865 Chr.5: 64804237-64804237 on Build GRCh38
rs140234898 Chr.5: 64804315-64804315 on Build GRCh38
rs770234813 Chr.5: 64804382-64804382 on Build GRCh38
rs1049682409 Chr.5: 64804431-64804431 on Build GRCh38
rs760410222 Chr.5: 64804212-64804213 on Build GRCh38
rs760775772 Chr.5: 64804253-64804253 on Build GRCh38
rs138755572 Chr.5: 64804484-64804484 on Build GRCh38
rs749089890 Chr.5: 64804203-64804203 on Build GRCh38
rs1053617331 Chr.5: 64804559-64804559 on Build GRCh38
rs892378517 Chr.5: 64804282-64804282 on Build GRCh38
rs912211059 Chr.5: 64804505-64804505 on Build GRCh38
rs750081188 Chr.5: 64804401-64804401 on Build GRCh38
rs956545920 Chr.5: 64804216-64804216 on Build GRCh38
rs915193174 Chr.5: 64804289-64804289 on Build GRCh38
rs773455166 Chr.5: 64804238-64804238 on Build GRCh38
rs775866124 Chr.5: 64804383-64804383 on Build GRCh38
rs892405725 Chr.5: 64804582-64804582 on Build GRCh38
rs768485141 Chr.5: 64804217-64804217 on Build GRCh38
rs772461502 Chr.5: 64804344-64804344 on Build GRCh38
rs760098385 Chr.5: 64804375-64804375 on Build GRCh38
rs745337361 Chr.5: 64804195-64804195 on Build GRCh38
rs753811241 Chr.5: 64804330-64804330 on Build GRCh38
rs748387936 Chr.5: 64804232-64804232 on Build GRCh38
rs376548134 Chr.5: 64804190-64804190 on Build GRCh38
rs764283583 Chr.5: 64804354-64804356 on Build GRCh38
rs753231367 Chr.5: 64804430-64804430 on Build GRCh38
rs924661992 Chr.5: 64804207-64804207 on Build GRCh38
rs34570664 Chr.5: 64804384-64804384 on Build GRCh38
rs76515392 Chr.5: 64804468-64804469 on Build GRCh38
rs749944995 Chr.5: 64804320-64804320 on Build GRCh38
rs767210188 Chr.5: 64804388-64804388 on Build GRCh38
rs540603042 Chr.5: 64804182-64804182 on Build GRCh38
rs755005084 Chr.5: 64804346-64804346 on Build GRCh38
rs756441840 Chr.5: 64804467-64804467 on Build GRCh38
rs141258299 Chr.5: 64804506-64804506 on Build GRCh38
rs982881729 Chr.5: 64804478-64804478 on Build GRCh38
rs774781868 Chr.5: 64804221-64804221 on Build GRCh38
rs548145437 Chr.5: 64804442-64804442 on Build GRCh38
rs756833834 Chr.5: 64804181-64804181 on Build GRCh38
rs756040365 Chr.5: 64804198-64804198 on Build GRCh38

View Full Product Details