Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 72223596-72224086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS27
Entrez Gene ID
23107
Gene Name
mitochondrial ribosomal protein S27
Gene Aliases
MRP-S27, S27mt
UniGene
Hs.482491
Gene Chromosome Location
Chr.5: 72219409-72320257 on Build GRCh38
Transcripts
NM_001286748.1 NM_001286751.1 NM_015084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5868771 c.974C>T p.S325F Chr.5: 72223714-72223714 on Build GRCh38
3761309 c.851G>A p.G284D Chr.5: 72223837-72223837 on Build GRCh38
5884261 c.998G>A p.R333Q Chr.5: 72223690-72223690 on Build GRCh38
7000015 c.1005+8A>T p.? Chr.5: 72223675-72223675 on Build GRCh38
1743814 c.910G>A p.D304N Chr.5: 72223778-72223778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755815900 Chr.5: 72223656-72223659 on Build GRCh38
rs372653734 Chr.5: 72223629-72223629 on Build GRCh38
rs1040819269 Chr.5: 72223924-72223924 on Build GRCh38
rs932492420 Chr.5: 72223947-72223947 on Build GRCh38
rs3209157 Chr.5: 72223837-72223837 on Build GRCh38
rs146439858 Chr.5: 72223817-72223817 on Build GRCh38
rs180902760 Chr.5: 72223868-72223868 on Build GRCh38
rs201174216 Chr.5: 72223775-72223775 on Build GRCh38
rs543465707 Chr.5: 72223903-72223903 on Build GRCh38
rs762871981 Chr.5: 72223830-72223830 on Build GRCh38
rs912246252 Chr.5: 72223723-72223723 on Build GRCh38
rs748303069 Chr.5: 72223711-72223711 on Build GRCh38
rs1033741923 Chr.5: 72224038-72224038 on Build GRCh38
rs908315878 Chr.5: 72223653-72223653 on Build GRCh38
rs376015004 Chr.5: 72223789-72223789 on Build GRCh38
rs748380106 Chr.5: 72223672-72223675 on Build GRCh38
rs771325947 Chr.5: 72223826-72223826 on Build GRCh38
rs765735710 Chr.5: 72223644-72223644 on Build GRCh38
rs748590134 Chr.5: 72223795-72223795 on Build GRCh38
rs925051285 Chr.5: 72224050-72224050 on Build GRCh38
rs376664405 Chr.5: 72223666-72223666 on Build GRCh38
rs767833114 Chr.5: 72223649-72223649 on Build GRCh38
rs776962586 Chr.5: 72223827-72223827 on Build GRCh38
rs779088732 Chr.5: 72223879-72223879 on Build GRCh38
rs898544029 Chr.5: 72223928-72223928 on Build GRCh38
rs1020975144 Chr.5: 72223759-72223759 on Build GRCh38
rs995442344 Chr.5: 72224057-72224057 on Build GRCh38
rs772042178 Chr.5: 72223719-72223719 on Build GRCh38
rs538301314 Chr.5: 72223761-72223761 on Build GRCh38
rs753449750 Chr.5: 72223887-72223888 on Build GRCh38
rs780961059 Chr.5: 72223687-72223687 on Build GRCh38
rs893735915 Chr.5: 72223971-72223971 on Build GRCh38
rs74783714 Chr.5: 72223648-72223648 on Build GRCh38
rs11557156 Chr.5: 72223848-72223848 on Build GRCh38
rs753431770 Chr.5: 72223855-72223855 on Build GRCh38
rs547666613 Chr.5: 72223631-72223631 on Build GRCh38
rs751446042 Chr.5: 72223762-72223762 on Build GRCh38
rs753254378 Chr.5: 72223880-72223880 on Build GRCh38
rs763181395 Chr.5: 72223684-72223684 on Build GRCh38
rs754280558 Chr.5: 72223777-72223777 on Build GRCh38
rs564313652 Chr.5: 72223862-72223862 on Build GRCh38
rs200556076 Chr.5: 72223835-72223835 on Build GRCh38
rs755720097 Chr.5: 72223772-72223772 on Build GRCh38
rs1025456607 Chr.5: 72223959-72223959 on Build GRCh38
rs144450614 Chr.5: 72223667-72223667 on Build GRCh38
rs750078052 Chr.5: 72223770-72223770 on Build GRCh38
rs772206505 Chr.5: 72223802-72223802 on Build GRCh38
rs773741498 Chr.5: 72223656-72223656 on Build GRCh38
rs766950966 Chr.5: 72223767-72223767 on Build GRCh38
rs561632774 Chr.5: 72224039-72224039 on Build GRCh38
rs767485854 Chr.5: 72223927-72223927 on Build GRCh38
rs975017034 Chr.5: 72224049-72224049 on Build GRCh38
rs749311608 Chr.5: 72223879-72223879 on Build GRCh38
rs1042402731 Chr.5: 72224012-72224012 on Build GRCh38
rs768620494 Chr.5: 72223638-72223638 on Build GRCh38
rs760924486 Chr.5: 72223735-72223735 on Build GRCh38
rs373837365 Chr.5: 72223676-72223676 on Build GRCh38
rs1010013754 Chr.5: 72223726-72223726 on Build GRCh38
rs982485002 Chr.5: 72223889-72223889 on Build GRCh38
rs995359319 Chr.5: 72223940-72223940 on Build GRCh38
rs773627016 Chr.5: 72223730-72223730 on Build GRCh38
rs779270735 Chr.5: 72223708-72223708 on Build GRCh38
rs779544739 Chr.5: 72223786-72223786 on Build GRCh38
rs951236137 Chr.5: 72223966-72223966 on Build GRCh38
rs371221332 Chr.5: 72223675-72223675 on Build GRCh38
rs756690405 Chr.5: 72223683-72223683 on Build GRCh38
rs919561830 Chr.5: 72223624-72223624 on Build GRCh38
rs529032593 Chr.5: 72224061-72224061 on Build GRCh38
rs745503396 Chr.5: 72223690-72223690 on Build GRCh38
rs749889500 Chr.5: 72223847-72223847 on Build GRCh38
rs1009518100 Chr.5: 72223977-72223977 on Build GRCh38
rs761586667 Chr.5: 72223845-72223845 on Build GRCh38
rs188517497 Chr.5: 72223705-72223705 on Build GRCh38
rs755271237 Chr.5: 72223778-72223778 on Build GRCh38
rs747255768 Chr.5: 72223823-72223823 on Build GRCh38
rs368017037 Chr.5: 72223697-72223697 on Build GRCh38
rs1053698539 Chr.5: 72223951-72223951 on Build GRCh38
rs199822876 Chr.5: 72223665-72223665 on Build GRCh38
rs769716487 Chr.5: 72223739-72223739 on Build GRCh38
rs767042602 Chr.5: 72223846-72223846 on Build GRCh38
rs11557156 Chr.5: 72223848-72223848 on Build GRCh38
rs752461652 Chr.5: 72223649-72223651 on Build GRCh38
rs556389272 Chr.5: 72223763-72223763 on Build GRCh38
rs1003542962 Chr.5: 72224037-72224037 on Build GRCh38
rs943736766 Chr.5: 72223923-72223923 on Build GRCh38
rs906549785 Chr.5: 72224026-72224026 on Build GRCh38
rs775375023 Chr.5: 72223752-72223752 on Build GRCh38
rs758934418 Chr.5: 72223883-72223883 on Build GRCh38
rs759470947 Chr.5: 72223658-72223658 on Build GRCh38

View Full Product Details