Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73015676-73016128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO2
Entrez Gene ID
115548
Gene Name
FCH domain only 2
Gene Aliases
-
UniGene
Hs.165762
Gene Chromosome Location
Chr.5: 72955981-73090522 on Build GRCh38
Transcripts
NM_001146032.1 NM_138782.2 XM_017009016.1 XM_017009017.1 XM_017009018.1 XM_017009019.1 XM_017009020.1 XM_017009021.1 XM_017009022.1 XM_017009023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5724759 c.657C>T p.S219S Chr.5: 73015682-73015682 on Build GRCh38
5724761 c.657C>T p.S219S Chr.5: 73015682-73015682 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs895871391 Chr.5: 73015872-73015872 on Build GRCh38
rs537763156 Chr.5: 73015802-73015802 on Build GRCh38
rs550031554 Chr.5: 73015869-73015869 on Build GRCh38
rs951268217 Chr.5: 73015818-73015818 on Build GRCh38
rs189388969 Chr.5: 73015953-73015953 on Build GRCh38
rs951216136 Chr.5: 73016018-73016018 on Build GRCh38
rs752282144 Chr.5: 73015764-73015767 on Build GRCh38
rs947294801 Chr.5: 73016095-73016095 on Build GRCh38
rs745742229 Chr.5: 73015730-73015730 on Build GRCh38
rs913167076 Chr.5: 73015815-73015815 on Build GRCh38
rs763846094 Chr.5: 73015709-73015709 on Build GRCh38
rs371198234 Chr.5: 73015764-73015764 on Build GRCh38
rs924981661 Chr.5: 73016091-73016091 on Build GRCh38
rs988681030 Chr.5: 73016090-73016090 on Build GRCh38
rs994022818 Chr.5: 73016014-73016014 on Build GRCh38
rs942317580 Chr.5: 73015854-73015854 on Build GRCh38
rs1052616565 Chr.5: 73015883-73015883 on Build GRCh38
rs936423103 Chr.5: 73015875-73015875 on Build GRCh38
rs986648210 Chr.5: 73016022-73016022 on Build GRCh38
rs973893363 Chr.5: 73015782-73015782 on Build GRCh38
rs2607123 Chr.5: 73016038-73016038 on Build GRCh38
rs963966626 Chr.5: 73016061-73016061 on Build GRCh38
rs1030104545 Chr.5: 73016073-73016073 on Build GRCh38
rs947265294 Chr.5: 73015821-73015821 on Build GRCh38
rs374985447 Chr.5: 73015767-73015767 on Build GRCh38
rs963802121 Chr.5: 73015781-73015781 on Build GRCh38
rs751339379 Chr.5: 73015716-73015716 on Build GRCh38
rs1025936851 Chr.5: 73015788-73015788 on Build GRCh38
rs892663254 Chr.5: 73015891-73015891 on Build GRCh38
rs912212621 Chr.5: 73015844-73015844 on Build GRCh38
rs975495631 Chr.5: 73015862-73015862 on Build GRCh38
rs368908338 Chr.5: 73015729-73015729 on Build GRCh38
rs949626588 Chr.5: 73015898-73015898 on Build GRCh38
rs748574463 Chr.5: 73015765-73015765 on Build GRCh38
rs954585085 Chr.5: 73016075-73016075 on Build GRCh38
rs1016680478 Chr.5: 73015990-73015990 on Build GRCh38
rs367687360 Chr.5: 73015743-73015743 on Build GRCh38
rs986742752 Chr.5: 73015828-73015828 on Build GRCh38
rs994376860 Chr.5: 73015921-73015921 on Build GRCh38
rs780851001 Chr.5: 73015729-73015730 on Build GRCh38
rs1046653666 Chr.5: 73015948-73015948 on Build GRCh38
rs975070180 Chr.5: 73016076-73016076 on Build GRCh38
rs1025460572 Chr.5: 73016016-73016016 on Build GRCh38
rs1015887842 Chr.5: 73015998-73015998 on Build GRCh38
rs937319318 Chr.5: 73015867-73015867 on Build GRCh38
rs145815990 Chr.5: 73015771-73015771 on Build GRCh38
rs1005716933 Chr.5: 73015960-73015960 on Build GRCh38
rs1015858396 Chr.5: 73015735-73015735 on Build GRCh38
rs1002125457 Chr.5: 73015989-73015989 on Build GRCh38
rs756964118 Chr.5: 73015723-73015723 on Build GRCh38
rs1019512190 Chr.5: 73016024-73016024 on Build GRCh38
rs772273118 Chr.5: 73015766-73015766 on Build GRCh38
rs372243004 Chr.5: 73015738-73015738 on Build GRCh38
rs755689017 Chr.5: 73015732-73015732 on Build GRCh38
rs936427411 Chr.5: 73016102-73016102 on Build GRCh38
rs768330058 Chr.5: 73015756-73015756 on Build GRCh38
rs899243580 Chr.5: 73015997-73015997 on Build GRCh38
rs1026202460 Chr.5: 73015937-73015938 on Build GRCh38

View Full Product Details