Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73554277-73554807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRA2
Entrez Gene ID
57763
Gene Name
ankyrin repeat family A member 2
Gene Aliases
ANKRA
UniGene
Hs.239154
Gene Chromosome Location
Chr.5: 73552200-73565705 on Build GRCh38
Transcripts
NM_023039.4 XM_005248560.2 XM_017009678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224282 c.790G>A p.V264I Chr.5: 73554337-73554337 on Build GRCh38
739265 c.739-1G>A p.? Chr.5: 73554389-73554389 on Build GRCh38
6898086 c.742G>A p.G248R Chr.5: 73554385-73554385 on Build GRCh38
7336822 c.778C>T p.H260Y Chr.5: 73554349-73554349 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750457418 Chr.5: 73554386-73554386 on Build GRCh38
rs937032834 Chr.5: 73554554-73554554 on Build GRCh38
rs549997929 Chr.5: 73554667-73554667 on Build GRCh38
rs762338173 Chr.5: 73554357-73554357 on Build GRCh38
rs964069938 Chr.5: 73554358-73554358 on Build GRCh38
rs2544638 Chr.5: 73554619-73554619 on Build GRCh38
rs539982924 Chr.5: 73554483-73554483 on Build GRCh38
rs769949628 Chr.5: 73554726-73554726 on Build GRCh38
rs1002033483 Chr.5: 73554588-73554588 on Build GRCh38
rs774685297 Chr.5: 73554355-73554355 on Build GRCh38
rs766704724 Chr.5: 73554397-73554397 on Build GRCh38
rs759343082 Chr.5: 73554671-73554671 on Build GRCh38
rs376097818 Chr.5: 73554313-73554313 on Build GRCh38
rs756407727 Chr.5: 73554392-73554392 on Build GRCh38
rs1008331184 Chr.5: 73554303-73554303 on Build GRCh38
rs775841513 Chr.5: 73554312-73554312 on Build GRCh38
rs975402998 Chr.5: 73554379-73554379 on Build GRCh38
rs761906464 Chr.5: 73554743-73554743 on Build GRCh38
rs537222529 Chr.5: 73554543-73554543 on Build GRCh38
rs343105 Chr.5: 73554570-73554570 on Build GRCh38
rs2562283 Chr.5: 73554317-73554317 on Build GRCh38
rs928385170 Chr.5: 73554498-73554498 on Build GRCh38
rs1055474461 Chr.5: 73554564-73554564 on Build GRCh38
rs751936968 Chr.5: 73554524-73554528 on Build GRCh38
rs745319626 Chr.5: 73554437-73554437 on Build GRCh38
rs1013675585 Chr.5: 73554768-73554768 on Build GRCh38
rs780584037 Chr.5: 73554431-73554431 on Build GRCh38
rs960790787 Chr.5: 73554742-73554742 on Build GRCh38
rs557300161 Chr.5: 73554594-73554594 on Build GRCh38
rs343104 Chr.5: 73554457-73554457 on Build GRCh38
rs752597173 Chr.5: 73554407-73554407 on Build GRCh38
rs764631608 Chr.5: 73554333-73554333 on Build GRCh38
rs1047784885 Chr.5: 73554696-73554696 on Build GRCh38
rs1051251252 Chr.5: 73554443-73554443 on Build GRCh38
rs113077883 Chr.5: 73554653-73554653 on Build GRCh38
rs767512583 Chr.5: 73554365-73554365 on Build GRCh38
rs1008365320 Chr.5: 73554754-73554754 on Build GRCh38
rs774345624 Chr.5: 73554376-73554376 on Build GRCh38
rs139166858 Chr.5: 73554422-73554422 on Build GRCh38
rs558171197 Chr.5: 73554484-73554484 on Build GRCh38
rs757172160 Chr.5: 73554423-73554423 on Build GRCh38
rs776684601 Chr.5: 73554617-73554617 on Build GRCh38
rs930991967 Chr.5: 73554585-73554585 on Build GRCh38
rs758246764 Chr.5: 73554410-73554410 on Build GRCh38
rs555834571 Chr.5: 73554571-73554571 on Build GRCh38
rs777806507 Chr.5: 73554414-73554414 on Build GRCh38
rs1033467028 Chr.5: 73554712-73554712 on Build GRCh38
rs769506714 Chr.5: 73554439-73554439 on Build GRCh38

View Full Product Details