Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80672610-80673124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH3
Entrez Gene ID
4437
Gene Name
mutS homolog 3
Gene Aliases
DUP, MRP1
UniGene
Hs.280987
Gene Chromosome Location
Chr.5: 80654648-80876815 on Build GRCh38
Transcripts
NM_002439.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947753 c.924G>A p.K308K Chr.5: 80672755-80672755 on Build GRCh38
6358217 c.892G>C p.V298L Chr.5: 80672750-80672750 on Build GRCh38
6475098 c.979A>G p.K327E Chr.5: 80672837-80672837 on Build GRCh38
370162 c.883-1G>A p.? Chr.5: 80672740-80672740 on Build GRCh38
6715749 c.1006A>C p.K336Q Chr.5: 80672837-80672837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796776649 Chr.5: 80672917-80672920 on Build GRCh38
rs929659061 Chr.5: 80672794-80672794 on Build GRCh38
rs770200319 Chr.5: 80672770-80672770 on Build GRCh38
rs757003720 Chr.5: 80672897-80672897 on Build GRCh38
rs763366083 Chr.5: 80672826-80672826 on Build GRCh38
rs967606333 Chr.5: 80673071-80673071 on Build GRCh38
rs114833767 Chr.5: 80672681-80672681 on Build GRCh38
rs750622145 Chr.5: 80672864-80672864 on Build GRCh38
rs762169960 Chr.5: 80672843-80672843 on Build GRCh38
rs774838967 Chr.5: 80672832-80672832 on Build GRCh38
rs1029100804 Chr.5: 80672973-80672973 on Build GRCh38
rs573316612 Chr.5: 80672908-80672908 on Build GRCh38
rs987503360 Chr.5: 80673049-80673049 on Build GRCh38
rs776158265 Chr.5: 80672810-80672810 on Build GRCh38
rs764577777 Chr.5: 80672828-80672828 on Build GRCh38
rs967477502 Chr.5: 80672991-80672991 on Build GRCh38
rs368709702 Chr.5: 80672874-80672874 on Build GRCh38
rs6151644 Chr.5: 80672635-80672635 on Build GRCh38
rs745412081 Chr.5: 80672813-80672813 on Build GRCh38
rs954847355 Chr.5: 80672994-80672994 on Build GRCh38
rs536975089 Chr.5: 80672855-80672855 on Build GRCh38
rs911030215 Chr.5: 80672660-80672660 on Build GRCh38
rs767758514 Chr.5: 80672713-80672713 on Build GRCh38
rs201336852 Chr.5: 80672814-80672814 on Build GRCh38
rs979021564 Chr.5: 80673083-80673083 on Build GRCh38
rs770547789 Chr.5: 80672790-80672790 on Build GRCh38
rs1055578357 Chr.5: 80672680-80672680 on Build GRCh38
rs751322507 Chr.5: 80672893-80672893 on Build GRCh38
rs746243211 Chr.5: 80672754-80672754 on Build GRCh38
rs758255356 Chr.5: 80672886-80672886 on Build GRCh38
rs747974815 Chr.5: 80672767-80672767 on Build GRCh38
rs545083362 Chr.5: 80672910-80672910 on Build GRCh38
rs937085030 Chr.5: 80672678-80672678 on Build GRCh38
rs913440539 Chr.5: 80673067-80673067 on Build GRCh38
rs757543733 Chr.5: 80672705-80672705 on Build GRCh38
rs185557248 Chr.5: 80672911-80672911 on Build GRCh38
rs1043213408 Chr.5: 80672665-80672665 on Build GRCh38
rs777592841 Chr.5: 80672891-80672891 on Build GRCh38
rs1022159921 Chr.5: 80672969-80672969 on Build GRCh38
rs149662421 Chr.5: 80672784-80672784 on Build GRCh38
rs575674036 Chr.5: 80672919-80672919 on Build GRCh38
rs756110352 Chr.5: 80672746-80672746 on Build GRCh38
rs1046694553 Chr.5: 80672799-80672799 on Build GRCh38
rs367818642 Chr.5: 80672916-80672917 on Build GRCh38
rs1030316269 Chr.5: 80673061-80673061 on Build GRCh38
rs1034232918 Chr.5: 80672821-80672821 on Build GRCh38
rs963905950 Chr.5: 80672929-80672929 on Build GRCh38
rs544674944 Chr.5: 80673069-80673069 on Build GRCh38
rs986682500 Chr.5: 80673085-80673085 on Build GRCh38
rs887040509 Chr.5: 80672802-80672802 on Build GRCh38
rs367621789 Chr.5: 80673005-80673005 on Build GRCh38
rs776070993 Chr.5: 80672819-80672819 on Build GRCh38
rs759719304 Chr.5: 80672900-80672901 on Build GRCh38
rs996247738 Chr.5: 80672960-80672960 on Build GRCh38
rs181036898 Chr.5: 80672873-80672873 on Build GRCh38
rs750501889 Chr.5: 80672714-80672714 on Build GRCh38
rs760818925 Chr.5: 80672869-80672869 on Build GRCh38
rs778050509 Chr.5: 80672671-80672671 on Build GRCh38
rs1677655 Chr.5: 80672677-80672677 on Build GRCh38
rs774758214 Chr.5: 80672698-80672699 on Build GRCh38
rs115177266 Chr.5: 80672782-80672782 on Build GRCh38

View Full Product Details