Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80674875-80675315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH3
Entrez Gene ID
4437
Gene Name
mutS homolog 3
Gene Aliases
DUP, MRP1
UniGene
Hs.280987
Gene Chromosome Location
Chr.5: 80654648-80876815 on Build GRCh38
Transcripts
NM_002439.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618322 c.1017C>T p.I339I Chr.5: 80674999-80674999 on Build GRCh38
6436771 c.1140C>A p.D380E Chr.5: 80675095-80675095 on Build GRCh38
3994486 c.1127A>T p.N376I Chr.5: 80675109-80675109 on Build GRCh38
3776893 c.1095C>T p.T365T Chr.5: 80675050-80675050 on Build GRCh38
6475134 c.1010C>A p.P337H Chr.5: 80674992-80674992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139170496 Chr.5: 80675099-80675099 on Build GRCh38
rs146703236 Chr.5: 80675077-80675077 on Build GRCh38
rs1050041501 Chr.5: 80675221-80675221 on Build GRCh38
rs999157792 Chr.5: 80675062-80675062 on Build GRCh38
rs779579188 Chr.5: 80675057-80675057 on Build GRCh38
rs978126073 Chr.5: 80674952-80674952 on Build GRCh38
rs760855166 Chr.5: 80675137-80675137 on Build GRCh38
rs555101803 Chr.5: 80675007-80675007 on Build GRCh38
rs139015837 Chr.5: 80674926-80674926 on Build GRCh38
rs531466601 Chr.5: 80675056-80675056 on Build GRCh38
rs71539691 Chr.5: 80675128-80675129 on Build GRCh38
rs776871293 Chr.5: 80675104-80675104 on Build GRCh38
rs879133824 Chr.5: 80675254-80675254 on Build GRCh38
rs772505587 Chr.5: 80674987-80674987 on Build GRCh38
rs931546318 Chr.5: 80675206-80675206 on Build GRCh38
rs545826957 Chr.5: 80675001-80675001 on Build GRCh38
rs774961818 Chr.5: 80674978-80674978 on Build GRCh38
rs761726585 Chr.5: 80675133-80675133 on Build GRCh38
rs928612707 Chr.5: 80674910-80674910 on Build GRCh38
rs748719147 Chr.5: 80675176-80675176 on Build GRCh38
rs1023881607 Chr.5: 80675106-80675106 on Build GRCh38
rs71539688 Chr.5: 80675109-80675109 on Build GRCh38
rs552453605 Chr.5: 80675284-80675284 on Build GRCh38
rs754537091 Chr.5: 80675169-80675169 on Build GRCh38
rs948622384 Chr.5: 80675273-80675273 on Build GRCh38
rs944240457 Chr.5: 80675288-80675288 on Build GRCh38
rs71539683 Chr.5: 80675019-80675019 on Build GRCh38
rs765235885 Chr.5: 80675111-80675111 on Build GRCh38
rs992109956 Chr.5: 80675231-80675231 on Build GRCh38
rs146475000 Chr.5: 80675118-80675118 on Build GRCh38
rs377063765 Chr.5: 80674922-80674922 on Build GRCh38
rs751866806 Chr.5: 80675063-80675063 on Build GRCh38
rs773431505 Chr.5: 80675079-80675079 on Build GRCh38
rs889997385 Chr.5: 80674969-80674969 on Build GRCh38
rs764225623 Chr.5: 80675172-80675173 on Build GRCh38
rs750148437 Chr.5: 80675035-80675035 on Build GRCh38
rs759555645 Chr.5: 80675110-80675110 on Build GRCh38
rs748596644 Chr.5: 80675059-80675059 on Build GRCh38
rs760799464 Chr.5: 80674996-80674996 on Build GRCh38
rs374581205 Chr.5: 80675139-80675139 on Build GRCh38
rs749860215 Chr.5: 80674956-80674956 on Build GRCh38
rs778559988 Chr.5: 80675171-80675171 on Build GRCh38
rs35425529 Chr.5: 80675200-80675201 on Build GRCh38
rs55976728 Chr.5: 80675195-80675195 on Build GRCh38
rs939391017 Chr.5: 80675211-80675211 on Build GRCh38
rs71539689 Chr.5: 80675116-80675116 on Build GRCh38
rs373336770 Chr.5: 80675040-80675040 on Build GRCh38
rs758948335 Chr.5: 80675177-80675177 on Build GRCh38
rs587776701 Chr.5: 80675103-80675103 on Build GRCh38
rs562793293 Chr.5: 80675043-80675043 on Build GRCh38
rs748784115 Chr.5: 80675044-80675044 on Build GRCh38
rs761005861 Chr.5: 80675096-80675096 on Build GRCh38
rs764234846 Chr.5: 80674931-80674931 on Build GRCh38
rs769258876 Chr.5: 80674977-80674977 on Build GRCh38
rs527400902 Chr.5: 80675192-80675192 on Build GRCh38
rs373961048 Chr.5: 80675006-80675006 on Build GRCh38
rs561642413 Chr.5: 80675152-80675152 on Build GRCh38
rs540294263 Chr.5: 80674920-80674920 on Build GRCh38
rs773569755 Chr.5: 80674992-80674992 on Build GRCh38
rs550630377 Chr.5: 80675199-80675199 on Build GRCh38
rs140543135 Chr.5: 80675115-80675115 on Build GRCh38
rs71539687 Chr.5: 80675103-80675103 on Build GRCh38
rs767851159 Chr.5: 80675093-80675094 on Build GRCh38
rs751448831 Chr.5: 80675018-80675018 on Build GRCh38
rs911363170 Chr.5: 80675256-80675256 on Build GRCh38
rs188507741 Chr.5: 80675204-80675204 on Build GRCh38
rs753475310 Chr.5: 80675158-80675158 on Build GRCh38
rs769413360 Chr.5: 80674945-80674945 on Build GRCh38
rs56116918 Chr.5: 80674953-80674953 on Build GRCh38
rs71539684 Chr.5: 80675079-80675079 on Build GRCh38
rs71539685 Chr.5: 80675082-80675082 on Build GRCh38
rs763165611 Chr.5: 80674929-80674929 on Build GRCh38
rs766534747 Chr.5: 80674999-80674999 on Build GRCh38
rs917189663 Chr.5: 80675245-80675245 on Build GRCh38
rs1008382885 Chr.5: 80674982-80674982 on Build GRCh38
rs765972998 Chr.5: 80675155-80675155 on Build GRCh38
rs757051997 Chr.5: 80675020-80675020 on Build GRCh38
rs928721752 Chr.5: 80675129-80675129 on Build GRCh38
rs904291316 Chr.5: 80674908-80674908 on Build GRCh38
rs940026025 Chr.5: 80674919-80674919 on Build GRCh38
rs71539686 Chr.5: 80675088-80675088 on Build GRCh38
rs780963280 Chr.5: 80675026-80675026 on Build GRCh38
rs368084849 Chr.5: 80675140-80675140 on Build GRCh38

View Full Product Details