Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134857801-134858298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf24
Entrez Gene ID
134553
Gene Name
chromosome 5 open reading frame 24
Gene Aliases
-
UniGene
Hs.708536
Gene Chromosome Location
Chr.5: 134833603-134859735 on Build GRCh38
Transcripts
NM_001135586.1 NM_001300894.1 NM_152409.3 XM_005271889.3 XM_017009049.1 XM_017009050.1

SNP Detail

SNP ID Chromosome Location
rs180750598 Chr.5: 134858020-134858020 on Build GRCh38
rs982199487 Chr.5: 134858245-134858245 on Build GRCh38
rs531216416 Chr.5: 134858069-134858069 on Build GRCh38
rs969894610 Chr.5: 134858230-134858230 on Build GRCh38
rs80346634 Chr.5: 134858122-134858122 on Build GRCh38
rs189595352 Chr.5: 134857858-134857858 on Build GRCh38
rs1033131877 Chr.5: 134857841-134857841 on Build GRCh38
rs1036670109 Chr.5: 134858046-134858046 on Build GRCh38
rs767756050 Chr.5: 134858039-134858039 on Build GRCh38
rs928493225 Chr.5: 134858259-134858259 on Build GRCh38
rs756716331 Chr.5: 134857938-134857938 on Build GRCh38
rs575372488 Chr.5: 134858033-134858033 on Build GRCh38
rs762668716 Chr.5: 134857987-134857987 on Build GRCh38
rs879188084 Chr.5: 134858122-134858122 on Build GRCh38
rs529574675 Chr.5: 134858224-134858224 on Build GRCh38
rs1048612747 Chr.5: 134858009-134858009 on Build GRCh38
rs988604918 Chr.5: 134858276-134858276 on Build GRCh38
rs988788973 Chr.5: 134857886-134857886 on Build GRCh38
rs891507184 Chr.5: 134858051-134858051 on Build GRCh38
rs540680017 Chr.5: 134858183-134858183 on Build GRCh38
rs574420490 Chr.5: 134857961-134857961 on Build GRCh38
rs947038110 Chr.5: 134858059-134858059 on Build GRCh38
rs963338151 Chr.5: 134857916-134857916 on Build GRCh38
rs367598207 Chr.5: 134858048-134858048 on Build GRCh38
rs999998657 Chr.5: 134858163-134858163 on Build GRCh38
rs185600585 Chr.5: 134858272-134858272 on Build GRCh38
rs118050206 Chr.5: 134858131-134858131 on Build GRCh38
rs374934904 Chr.5: 134858218-134858218 on Build GRCh38
rs922077914 Chr.5: 134857946-134857946 on Build GRCh38
rs564435043 Chr.5: 134858130-134858130 on Build GRCh38
rs753210581 Chr.5: 134857914-134857914 on Build GRCh38
rs546234260 Chr.5: 134858040-134858040 on Build GRCh38
rs895512682 Chr.5: 134858060-134858060 on Build GRCh38
rs1030941708 Chr.5: 134858121-134858122 on Build GRCh38
rs948128312 Chr.5: 134857962-134857962 on Build GRCh38
rs980355292 Chr.5: 134858015-134858015 on Build GRCh38
rs552844778 Chr.5: 134857957-134857957 on Build GRCh38
rs758053651 Chr.5: 134858070-134858070 on Build GRCh38
rs574552737 Chr.5: 134858244-134858244 on Build GRCh38
rs944912476 Chr.5: 134857911-134857911 on Build GRCh38
rs551312196 Chr.5: 134858168-134858168 on Build GRCh38
rs541608932 Chr.5: 134857964-134857964 on Build GRCh38
rs963556411 Chr.5: 134858190-134858190 on Build GRCh38
rs911899845 Chr.5: 134857895-134857895 on Build GRCh38

View Full Product Details