Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139817316-139817810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSD2
Entrez Gene ID
84249
Gene Name
pleckstrin and Sec7 domain containing 2
Gene Aliases
EFA6C
UniGene
Hs.21963
Gene Chromosome Location
Chr.5: 139683980-139844466 on Build GRCh38
Transcripts
NM_032289.2 XM_011537696.2 XM_017009975.1 XM_017009976.1 XM_017009977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3918751 c.1021G>A p.E341K Chr.5: 139817485-139817485 on Build GRCh38
5823551 c.1027A>G p.S343G Chr.5: 139817491-139817491 on Build GRCh38
3994000 c.1020C>T p.N340N Chr.5: 139817484-139817484 on Build GRCh38
3850548 c.1070C>T p.S357L Chr.5: 139817534-139817534 on Build GRCh38
3393170 c.1087G>A p.G363R Chr.5: 139817551-139817551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756626496 Chr.5: 139817537-139817537 on Build GRCh38
rs747518596 Chr.5: 139817550-139817550 on Build GRCh38
rs927141558 Chr.5: 139817710-139817710 on Build GRCh38
rs1006713542 Chr.5: 139817627-139817627 on Build GRCh38
rs369670477 Chr.5: 139817603-139817603 on Build GRCh38
rs766537676 Chr.5: 139817523-139817523 on Build GRCh38
rs745379075 Chr.5: 139817440-139817440 on Build GRCh38
rs1048829997 Chr.5: 139817698-139817698 on Build GRCh38
rs748064197 Chr.5: 139817492-139817492 on Build GRCh38
rs377120564 Chr.5: 139817582-139817582 on Build GRCh38
rs186814250 Chr.5: 139817473-139817473 on Build GRCh38
rs936312669 Chr.5: 139817679-139817679 on Build GRCh38
rs763089302 Chr.5: 139817650-139817650 on Build GRCh38
rs878905135 Chr.5: 139817445-139817445 on Build GRCh38
rs35714177 Chr.5: 139817551-139817551 on Build GRCh38
rs577662249 Chr.5: 139817713-139817713 on Build GRCh38
rs139242669 Chr.5: 139817535-139817535 on Build GRCh38
rs773140424 Chr.5: 139817644-139817644 on Build GRCh38
rs541733560 Chr.5: 139817729-139817729 on Build GRCh38
rs367820622 Chr.5: 139817446-139817446 on Build GRCh38
rs371792247 Chr.5: 139817461-139817461 on Build GRCh38
rs1055116654 Chr.5: 139817725-139817725 on Build GRCh38
rs773205125 Chr.5: 139817506-139817506 on Build GRCh38
rs150955322 Chr.5: 139817505-139817505 on Build GRCh38
rs774551744 Chr.5: 139817607-139817607 on Build GRCh38
rs1055171442 Chr.5: 139817514-139817514 on Build GRCh38
rs1006189923 Chr.5: 139817778-139817778 on Build GRCh38
rs910942889 Chr.5: 139817695-139817695 on Build GRCh38
rs768303780 Chr.5: 139817485-139817485 on Build GRCh38
rs771141876 Chr.5: 139817673-139817673 on Build GRCh38
rs780792648 Chr.5: 139817575-139817575 on Build GRCh38
rs201797899 Chr.5: 139817595-139817595 on Build GRCh38
rs552313681 Chr.5: 139817422-139817422 on Build GRCh38
rs144267818 Chr.5: 139817688-139817688 on Build GRCh38
rs376308900 Chr.5: 139817466-139817466 on Build GRCh38
rs767761922 Chr.5: 139817534-139817534 on Build GRCh38
rs149567975 Chr.5: 139817573-139817573 on Build GRCh38
rs759622754 Chr.5: 139817608-139817608 on Build GRCh38
rs774410580 Chr.5: 139817526-139817526 on Build GRCh38
rs770092758 Chr.5: 139817612-139817612 on Build GRCh38
rs756576186 Chr.5: 139817766-139817767 on Build GRCh38
rs763047342 Chr.5: 139817516-139817516 on Build GRCh38
rs985244592 Chr.5: 139817394-139817394 on Build GRCh38
rs781655084 Chr.5: 139817432-139817432 on Build GRCh38
rs1013638324 Chr.5: 139817735-139817735 on Build GRCh38
rs537283609 Chr.5: 139817674-139817674 on Build GRCh38
rs749270947 Chr.5: 139817597-139817597 on Build GRCh38
rs769675466 Chr.5: 139817589-139817589 on Build GRCh38
rs75029641 Chr.5: 139817393-139817393 on Build GRCh38
rs568901457 Chr.5: 139817658-139817658 on Build GRCh38
rs570447589 Chr.5: 139817439-139817439 on Build GRCh38
rs371810809 Chr.5: 139817610-139817610 on Build GRCh38
rs924939204 Chr.5: 139817677-139817677 on Build GRCh38
rs972396920 Chr.5: 139817653-139817653 on Build GRCh38
rs1047761071 Chr.5: 139817742-139817742 on Build GRCh38
rs952195261 Chr.5: 139817618-139817618 on Build GRCh38
rs1054593704 Chr.5: 139817486-139817486 on Build GRCh38
rs199518668 Chr.5: 139817467-139817467 on Build GRCh38
rs753143508 Chr.5: 139817433-139817433 on Build GRCh38
rs936411522 Chr.5: 139817477-139817477 on Build GRCh38
rs142589356 Chr.5: 139817484-139817484 on Build GRCh38
rs181882384 Chr.5: 139817395-139817395 on Build GRCh38
rs530255820 Chr.5: 139817371-139817371 on Build GRCh38
rs748022427 Chr.5: 139817645-139817645 on Build GRCh38
rs926434078 Chr.5: 139817380-139817380 on Build GRCh38
rs1042360611 Chr.5: 139817743-139817743 on Build GRCh38
rs140081828 Chr.5: 139817756-139817756 on Build GRCh38
rs917858264 Chr.5: 139817663-139817663 on Build GRCh38
rs368556780 Chr.5: 139817527-139817527 on Build GRCh38
rs376441675 Chr.5: 139817436-139817436 on Build GRCh38
rs959312575 Chr.5: 139817392-139817392 on Build GRCh38
rs780497261 Chr.5: 139817538-139817538 on Build GRCh38
rs1033776628 Chr.5: 139817786-139817786 on Build GRCh38
rs776553648 Chr.5: 139817487-139817487 on Build GRCh38
rs749904642 Chr.5: 139817543-139817543 on Build GRCh38
rs541965198 Chr.5: 139817378-139817378 on Build GRCh38
rs553660179 Chr.5: 139817732-139817732 on Build GRCh38

View Full Product Details