Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140703792-140704313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMAT2
Entrez Gene ID
153527
Gene Name
zinc finger matrin-type 2
Gene Aliases
Ptg-12, Snu23
UniGene
Hs.350194
Gene Chromosome Location
Chr.5: 140700360-140706681 on Build GRCh38
Transcripts
NM_144723.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1433661 c.248A>G p.N83S Chr.5: 140703929-140703929 on Build GRCh38
1270971 c.293A>G p.H98R Chr.5: 140703974-140703974 on Build GRCh38
1495980 c.237-2A>T p.? Chr.5: 140703916-140703916 on Build GRCh38
1061765 c.306G>T p.K102N Chr.5: 140703987-140703987 on Build GRCh38
6241518 c.280A>G p.N94D Chr.5: 140703961-140703961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190582056 Chr.5: 140704177-140704177 on Build GRCh38
rs550560249 Chr.5: 140704101-140704101 on Build GRCh38
rs185030174 Chr.5: 140704169-140704169 on Build GRCh38
rs765902018 Chr.5: 140703948-140703948 on Build GRCh38
rs752011347 Chr.5: 140704033-140704033 on Build GRCh38
rs994688652 Chr.5: 140704163-140704163 on Build GRCh38
rs545879916 Chr.5: 140703885-140703885 on Build GRCh38
rs746189880 Chr.5: 140703887-140703887 on Build GRCh38
rs947566151 Chr.5: 140703891-140703891 on Build GRCh38
rs752668827 Chr.5: 140704007-140704007 on Build GRCh38
rs940577374 Chr.5: 140704155-140704155 on Build GRCh38
rs771995144 Chr.5: 140704278-140704278 on Build GRCh38
rs1025770732 Chr.5: 140704204-140704204 on Build GRCh38
rs761224177 Chr.5: 140703909-140703909 on Build GRCh38
rs779073953 Chr.5: 140704051-140704051 on Build GRCh38
rs1003213047 Chr.5: 140704145-140704145 on Build GRCh38
rs756073335 Chr.5: 140704023-140704023 on Build GRCh38
rs768671778 Chr.5: 140704160-140704160 on Build GRCh38
rs972870126 Chr.5: 140703825-140703825 on Build GRCh38
rs932084899 Chr.5: 140704028-140704028 on Build GRCh38
rs17286731 Chr.5: 140704159-140704159 on Build GRCh38
rs749389905 Chr.5: 140703874-140703874 on Build GRCh38
rs73791755 Chr.5: 140704103-140704103 on Build GRCh38
rs753811871 Chr.5: 140704041-140704041 on Build GRCh38
rs1003736420 Chr.5: 140704183-140704183 on Build GRCh38
rs777779611 Chr.5: 140704029-140704029 on Build GRCh38
rs779128755 Chr.5: 140703886-140703886 on Build GRCh38
rs759374873 Chr.5: 140703966-140703966 on Build GRCh38
rs777773269 Chr.5: 140703872-140703872 on Build GRCh38
rs918657326 Chr.5: 140703854-140703854 on Build GRCh38
rs769339565 Chr.5: 140703911-140703911 on Build GRCh38
rs879147244 Chr.5: 140704106-140704113 on Build GRCh38
rs1017251747 Chr.5: 140704166-140704166 on Build GRCh38
rs1035127036 Chr.5: 140703868-140703868 on Build GRCh38
rs942970076 Chr.5: 140704061-140704061 on Build GRCh38
rs913364514 Chr.5: 140703889-140703889 on Build GRCh38
rs1055086015 Chr.5: 140704082-140704082 on Build GRCh38
rs376719415 Chr.5: 140703912-140703912 on Build GRCh38
rs955450679 Chr.5: 140704211-140704211 on Build GRCh38
rs570182778 Chr.5: 140704253-140704253 on Build GRCh38
rs763123829 Chr.5: 140703907-140703907 on Build GRCh38
rs771129982 Chr.5: 140703866-140703866 on Build GRCh38
rs986845807 Chr.5: 140704254-140704254 on Build GRCh38
rs1044942275 Chr.5: 140704176-140704176 on Build GRCh38
rs117798152 Chr.5: 140703898-140703898 on Build GRCh38
rs960310008 Chr.5: 140703883-140703883 on Build GRCh38
rs772462598 Chr.5: 140703894-140703894 on Build GRCh38
rs191426383 Chr.5: 140703930-140703930 on Build GRCh38
rs140024135 Chr.5: 140703960-140703960 on Build GRCh38
rs902262757 Chr.5: 140704140-140704140 on Build GRCh38
rs767470626 Chr.5: 140703975-140703975 on Build GRCh38

View Full Product Details