Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141051709-141052214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101926905
Entrez Gene ID
101926905
Gene Name
uncharacterized LOC101926905
Gene Aliases
-
UniGene
Hs.570900
Gene Chromosome Location
Chr.5: 141046205-141078031 on Build GRCh38
Transcripts
NR_105056.2 NM_013340.3

Gene Symbol
PCDHB1
Entrez Gene ID
29930
Gene Name
protocadherin beta 1
Gene Aliases
PCDH-BETA1
UniGene
Hs.278950
Gene Chromosome Location
Chr.5: 141051376-141053962 on Build GRCh38
Transcripts
NR_105056.2 NM_013340.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347300 c.722A>T p.Q241L Chr.5: 141052192-141052192 on Build GRCh38
3851346 c.348C>A p.S116S Chr.5: 141051818-141051818 on Build GRCh38
3851344 c.297C>T p.A99A Chr.5: 141051767-141051767 on Build GRCh38
205113 c.423G>T p.K141N Chr.5: 141051893-141051893 on Build GRCh38
4964245 c.537C>T p.H179H Chr.5: 141052007-141052007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782214766 Chr.5: 141051807-141051807 on Build GRCh38
rs782596192 Chr.5: 141052041-141052042 on Build GRCh38
rs782121736 Chr.5: 141051850-141051850 on Build GRCh38
rs782809746 Chr.5: 141052003-141052003 on Build GRCh38
rs782712715 Chr.5: 141051884-141051884 on Build GRCh38
rs782104351 Chr.5: 141052130-141052130 on Build GRCh38
rs140555922 Chr.5: 141051917-141051917 on Build GRCh38
rs989886186 Chr.5: 141051802-141051802 on Build GRCh38
rs924398578 Chr.5: 141051768-141051768 on Build GRCh38
rs377751088 Chr.5: 141051994-141051994 on Build GRCh38
rs1022924332 Chr.5: 141051959-141051959 on Build GRCh38
rs781951598 Chr.5: 141052129-141052129 on Build GRCh38
rs1031520472 Chr.5: 141051995-141051995 on Build GRCh38
rs200399731 Chr.5: 141052172-141052172 on Build GRCh38
rs782456574 Chr.5: 141052025-141052025 on Build GRCh38
rs782636936 Chr.5: 141051899-141051899 on Build GRCh38
rs782466235 Chr.5: 141052177-141052177 on Build GRCh38
rs782150315 Chr.5: 141052163-141052163 on Build GRCh38
rs782180084 Chr.5: 141051767-141051767 on Build GRCh38
rs782642423 Chr.5: 141052083-141052083 on Build GRCh38
rs541462514 Chr.5: 141051890-141051890 on Build GRCh38
rs782694210 Chr.5: 141052154-141052154 on Build GRCh38
rs147316398 Chr.5: 141052080-141052080 on Build GRCh38
rs782245962 Chr.5: 141052001-141052001 on Build GRCh38
rs147085881 Chr.5: 141051930-141051930 on Build GRCh38
rs751636276 Chr.5: 141051866-141051866 on Build GRCh38
rs371875057 Chr.5: 141051979-141051980 on Build GRCh38
rs138595607 Chr.5: 141051932-141051932 on Build GRCh38
rs199852101 Chr.5: 141051785-141051785 on Build GRCh38
rs781893392 Chr.5: 141052069-141052069 on Build GRCh38
rs148313450 Chr.5: 141051881-141051881 on Build GRCh38
rs782390972 Chr.5: 141052119-141052119 on Build GRCh38
rs781944168 Chr.5: 141052157-141052157 on Build GRCh38
rs974376855 Chr.5: 141052192-141052192 on Build GRCh38
rs782820854 Chr.5: 141052167-141052167 on Build GRCh38
rs369721348 Chr.5: 141051846-141051846 on Build GRCh38
rs782208975 Chr.5: 141052087-141052087 on Build GRCh38
rs782739946 Chr.5: 141052179-141052179 on Build GRCh38
rs782000074 Chr.5: 141051788-141051788 on Build GRCh38
rs782302687 Chr.5: 141051988-141051988 on Build GRCh38
rs782466746 Chr.5: 141051918-141051918 on Build GRCh38
rs782451639 Chr.5: 141052193-141052193 on Build GRCh38
rs200874801 Chr.5: 141052011-141052011 on Build GRCh38
rs782290189 Chr.5: 141051783-141051783 on Build GRCh38
rs376473997 Chr.5: 141051843-141051843 on Build GRCh38
rs782030201 Chr.5: 141052108-141052108 on Build GRCh38
rs781838912 Chr.5: 141052016-141052016 on Build GRCh38
rs782415579 Chr.5: 141052106-141052106 on Build GRCh38
rs782664044 Chr.5: 141051833-141051834 on Build GRCh38
rs964766459 Chr.5: 141052109-141052109 on Build GRCh38
rs144072178 Chr.5: 141052043-141052043 on Build GRCh38
rs192224343 Chr.5: 141051997-141051997 on Build GRCh38
rs782384804 Chr.5: 141051944-141051944 on Build GRCh38
rs550698671 Chr.5: 141051982-141051982 on Build GRCh38
rs782534096 Chr.5: 141051977-141051978 on Build GRCh38
rs782116875 Chr.5: 141052114-141052114 on Build GRCh38
rs971250790 Chr.5: 141051739-141051739 on Build GRCh38
rs782459294 Chr.5: 141051738-141051738 on Build GRCh38
rs1039961746 Chr.5: 141051844-141051844 on Build GRCh38
rs143302058 Chr.5: 141051830-141051830 on Build GRCh38
rs782667672 Chr.5: 141051776-141051776 on Build GRCh38
rs782732872 Chr.5: 141052015-141052015 on Build GRCh38
rs782573947 Chr.5: 141052091-141052091 on Build GRCh38
rs782188780 Chr.5: 141052097-141052097 on Build GRCh38
rs781890750 Chr.5: 141051889-141051889 on Build GRCh38
rs781786602 Chr.5: 141052191-141052191 on Build GRCh38
rs563916297 Chr.5: 141051969-141051969 on Build GRCh38
rs782363571 Chr.5: 141051822-141051822 on Build GRCh38
rs554057381 Chr.5: 141052088-141052088 on Build GRCh38
rs782391922 Chr.5: 141052007-141052007 on Build GRCh38
rs782180561 Chr.5: 141051927-141051927 on Build GRCh38
rs371869618 Chr.5: 141051839-141051839 on Build GRCh38
rs782629233 Chr.5: 141051924-141051924 on Build GRCh38
rs782453777 Chr.5: 141052027-141052027 on Build GRCh38
rs782613909 Chr.5: 141051745-141051745 on Build GRCh38
rs781898157 Chr.5: 141051835-141051836 on Build GRCh38
rs782305661 Chr.5: 141051938-141051938 on Build GRCh38
rs149129886 Chr.5: 141051771-141051771 on Build GRCh38
rs144200490 Chr.5: 141052111-141052112 on Build GRCh38

View Full Product Details