Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3850436-3850944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50B
Entrez Gene ID
26240
Gene Name
family with sequence similarity 50 member B
Gene Aliases
D6S2654E, X5L
UniGene
Hs.140944
Gene Chromosome Location
Chr.6: 3831901-3851320 on Build GRCh38
Transcripts
NM_012135.2 XM_017010729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553260 c.931G>A p.E311K Chr.6: 3850742-3850742 on Build GRCh38
237367 c.894C>T p.Y298Y Chr.6: 3850705-3850705 on Build GRCh38
6659812 c.890G>A p.W297* Chr.6: 3850701-3850701 on Build GRCh38
3873637 c.735G>A p.P245P Chr.6: 3850546-3850546 on Build GRCh38
5405166 c.718G>A p.E240K Chr.6: 3850529-3850529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751038763 Chr.6: 3850814-3850814 on Build GRCh38
rs778172840 Chr.6: 3850734-3850734 on Build GRCh38
rs990351853 Chr.6: 3850911-3850911 on Build GRCh38
rs989432033 Chr.6: 3850501-3850501 on Build GRCh38
rs771755028 Chr.6: 3850572-3850572 on Build GRCh38
rs754171595 Chr.6: 3850847-3850847 on Build GRCh38
rs777316197 Chr.6: 3850480-3850480 on Build GRCh38
rs757035966 Chr.6: 3850695-3850695 on Build GRCh38
rs750156055 Chr.6: 3850706-3850706 on Build GRCh38
rs778322705 Chr.6: 3850819-3850819 on Build GRCh38
rs145301744 Chr.6: 3850648-3850648 on Build GRCh38
rs915186229 Chr.6: 3850532-3850532 on Build GRCh38
rs1050316779 Chr.6: 3850732-3850732 on Build GRCh38
rs751588535 Chr.6: 3850500-3850500 on Build GRCh38
rs1022214145 Chr.6: 3850857-3850857 on Build GRCh38
rs757827580 Chr.6: 3850856-3850856 on Build GRCh38
rs34105433 Chr.6: 3850814-3850814 on Build GRCh38
rs777690143 Chr.6: 3850535-3850535 on Build GRCh38
rs866284085 Chr.6: 3850714-3850714 on Build GRCh38
rs981356308 Chr.6: 3850558-3850560 on Build GRCh38
rs752627352 Chr.6: 3850675-3850675 on Build GRCh38
rs75036841 Chr.6: 3850771-3850771 on Build GRCh38
rs765188202 Chr.6: 3850804-3850804 on Build GRCh38
rs906259732 Chr.6: 3850833-3850833 on Build GRCh38
rs774545077 Chr.6: 3850762-3850762 on Build GRCh38
rs772287009 Chr.6: 3850774-3850774 on Build GRCh38
rs369071179 Chr.6: 3850721-3850721 on Build GRCh38
rs1003309115 Chr.6: 3850845-3850845 on Build GRCh38
rs763248399 Chr.6: 3850618-3850618 on Build GRCh38
rs749859761 Chr.6: 3850465-3850465 on Build GRCh38
rs776475846 Chr.6: 3850790-3850790 on Build GRCh38
rs763956101 Chr.6: 3850678-3850678 on Build GRCh38
rs760695140 Chr.6: 3850778-3850778 on Build GRCh38
rs767412879 Chr.6: 3850509-3850509 on Build GRCh38
rs941583529 Chr.6: 3850757-3850757 on Build GRCh38
rs766522202 Chr.6: 3850499-3850499 on Build GRCh38
rs560092614 Chr.6: 3850910-3850910 on Build GRCh38
rs990794672 Chr.6: 3850460-3850460 on Build GRCh38
rs760598905 Chr.6: 3850676-3850676 on Build GRCh38
rs753583616 Chr.6: 3850544-3850544 on Build GRCh38
rs80012120 Chr.6: 3850496-3850496 on Build GRCh38
rs372713736 Chr.6: 3850573-3850573 on Build GRCh38
rs774951383 Chr.6: 3850819-3850820 on Build GRCh38
rs779634503 Chr.6: 3850726-3850726 on Build GRCh38
rs375092204 Chr.6: 3850615-3850615 on Build GRCh38
rs748727501 Chr.6: 3850584-3850584 on Build GRCh38
rs761653588 Chr.6: 3850803-3850803 on Build GRCh38
rs200299566 Chr.6: 3850539-3850539 on Build GRCh38
rs763167959 Chr.6: 3850498-3850498 on Build GRCh38
rs758066771 Chr.6: 3850809-3850809 on Build GRCh38
rs774598344 Chr.6: 3850637-3850637 on Build GRCh38
rs745333853 Chr.6: 3850564-3850564 on Build GRCh38
rs759641838 Chr.6: 3850502-3850502 on Build GRCh38
rs770206837 Chr.6: 3850585-3850585 on Build GRCh38
rs746249027 Chr.6: 3850828-3850828 on Build GRCh38
rs1025105995 Chr.6: 3850829-3850829 on Build GRCh38
rs764959243 Chr.6: 3850698-3850698 on Build GRCh38
rs934477657 Chr.6: 3850600-3850601 on Build GRCh38
rs978121852 Chr.6: 3850608-3850608 on Build GRCh38
rs764055512 Chr.6: 3850784-3850784 on Build GRCh38
rs746530287 Chr.6: 3850729-3850729 on Build GRCh38
rs746157049 Chr.6: 3850772-3850772 on Build GRCh38
rs756594284 Chr.6: 3850730-3850730 on Build GRCh38
rs771143310 Chr.6: 3850625-3850625 on Build GRCh38
rs574756557 Chr.6: 3850820-3850820 on Build GRCh38
rs1031979872 Chr.6: 3850882-3850883 on Build GRCh38
rs149589937 Chr.6: 3850756-3850756 on Build GRCh38
rs770248206 Chr.6: 3850485-3850485 on Build GRCh38
rs901508182 Chr.6: 3850745-3850745 on Build GRCh38
rs762243354 Chr.6: 3850483-3850483 on Build GRCh38
rs993124285 Chr.6: 3850781-3850781 on Build GRCh38
rs778422915 Chr.6: 3850471-3850471 on Build GRCh38
rs770347730 Chr.6: 3850461-3850461 on Build GRCh38
rs750244023 Chr.6: 3850807-3850807 on Build GRCh38
rs779380964 Chr.6: 3850889-3850889 on Build GRCh38
rs34696251 Chr.6: 3850708-3850709 on Build GRCh38
rs978048116 Chr.6: 3850909-3850909 on Build GRCh38
rs754485246 Chr.6: 3850818-3850818 on Build GRCh38
rs554746167 Chr.6: 3850742-3850742 on Build GRCh38
rs766151029 Chr.6: 3850813-3850813 on Build GRCh38
rs767643533 Chr.6: 3850669-3850669 on Build GRCh38
rs771251395 Chr.6: 3850761-3850761 on Build GRCh38
rs978148422 Chr.6: 3850869-3850869 on Build GRCh38
rs540546419 Chr.6: 3850823-3850823 on Build GRCh38
rs775494340 Chr.6: 3850776-3850776 on Build GRCh38
rs934092798 Chr.6: 3850663-3850663 on Build GRCh38
rs756879550 Chr.6: 3850545-3850545 on Build GRCh38
rs959218747 Chr.6: 3850852-3850852 on Build GRCh38
rs769254765 Chr.6: 3850778-3850779 on Build GRCh38
rs1012343350 Chr.6: 3850825-3850825 on Build GRCh38
rs1033528223 Chr.6: 3850494-3850494 on Build GRCh38
rs753623397 Chr.6: 3850692-3850692 on Build GRCh38
rs778455041 Chr.6: 3850547-3850547 on Build GRCh38
rs773566808 Chr.6: 3850603-3850603 on Build GRCh38
rs547607174 Chr.6: 3850510-3850510 on Build GRCh38
rs369953711 Chr.6: 3850473-3850473 on Build GRCh38
rs755890303 Chr.6: 3850520-3850520 on Build GRCh38

View Full Product Details