Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 6167304-6167849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F13A1
Entrez Gene ID
2162
Gene Name
coagulation factor XIII A chain
Gene Aliases
F13A
UniGene
Hs.335513
Gene Chromosome Location
Chr.6: 6144078-6320691 on Build GRCh38
Transcripts
NM_000129.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3875046 c.1862C>A p.A621D Chr.6: 6167504-6167504 on Build GRCh38
3159993 c.1753A>G p.K585E Chr.6: 6167613-6167613 on Build GRCh38
3159992 c.1779C>T p.G593G Chr.6: 6167587-6167587 on Build GRCh38
6563681 c.1792C>T p.Q598* Chr.6: 6167574-6167574 on Build GRCh38
6107279 c.1904T>A p.I635N Chr.6: 6167462-6167462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756227463 Chr.6: 6167481-6167481 on Build GRCh38
rs965914813 Chr.6: 6167823-6167823 on Build GRCh38
rs967946370 Chr.6: 6167411-6167411 on Build GRCh38
rs894061773 Chr.6: 6167748-6167748 on Build GRCh38
rs75972743 Chr.6: 6167343-6167347 on Build GRCh38
rs764336459 Chr.6: 6167642-6167642 on Build GRCh38
rs1032273355 Chr.6: 6167733-6167733 on Build GRCh38
rs201173378 Chr.6: 6167576-6167576 on Build GRCh38
rs5983 Chr.6: 6167600-6167600 on Build GRCh38
rs185994460 Chr.6: 6167458-6167458 on Build GRCh38
rs765199468 Chr.6: 6167664-6167664 on Build GRCh38
rs1008080038 Chr.6: 6167693-6167693 on Build GRCh38
rs762000974 Chr.6: 6167659-6167659 on Build GRCh38
rs746283829 Chr.6: 6167767-6167767 on Build GRCh38
rs769242418 Chr.6: 6167342-6167347 on Build GRCh38
rs780614198 Chr.6: 6167328-6167331 on Build GRCh38
rs200830173 Chr.6: 6167487-6167487 on Build GRCh38
rs1024989526 Chr.6: 6167404-6167404 on Build GRCh38
rs202099966 Chr.6: 6167641-6167641 on Build GRCh38
rs377479323 Chr.6: 6167605-6167605 on Build GRCh38
rs376162615 Chr.6: 6167590-6167590 on Build GRCh38
rs1036886798 Chr.6: 6167379-6167379 on Build GRCh38
rs570888570 Chr.6: 6167745-6167745 on Build GRCh38
rs145180358 Chr.6: 6167505-6167505 on Build GRCh38
rs747665925 Chr.6: 6167697-6167697 on Build GRCh38
rs138754417 Chr.6: 6167589-6167589 on Build GRCh38
rs368920004 Chr.6: 6167421-6167421 on Build GRCh38
rs148207995 Chr.6: 6167558-6167558 on Build GRCh38
rs138943514 Chr.6: 6167506-6167506 on Build GRCh38
rs768255836 Chr.6: 6167439-6167439 on Build GRCh38
rs773350962 Chr.6: 6167343-6167348 on Build GRCh38
rs1002154590 Chr.6: 6167699-6167699 on Build GRCh38
rs972825823 Chr.6: 6167757-6167757 on Build GRCh38
rs141416839 Chr.6: 6167502-6167502 on Build GRCh38
rs145219717 Chr.6: 6167493-6167493 on Build GRCh38
rs57207647 Chr.6: 6167341-6167346 on Build GRCh38
rs143769071 Chr.6: 6167533-6167533 on Build GRCh38
rs746371632 Chr.6: 6167625-6167625 on Build GRCh38
rs149970442 Chr.6: 6167488-6167488 on Build GRCh38
rs373723380 Chr.6: 6167550-6167550 on Build GRCh38
rs776595899 Chr.6: 6167647-6167647 on Build GRCh38
rs749034615 Chr.6: 6167327-6167331 on Build GRCh38
rs775774306 Chr.6: 6167636-6167636 on Build GRCh38
rs768343908 Chr.6: 6167527-6167527 on Build GRCh38
rs781302959 Chr.6: 6167425-6167425 on Build GRCh38
rs764095434 Chr.6: 6167557-6167557 on Build GRCh38
rs778756088 Chr.6: 6167416-6167416 on Build GRCh38
rs1021135018 Chr.6: 6167809-6167809 on Build GRCh38
rs939898712 Chr.6: 6167582-6167582 on Build GRCh38
rs146018037 Chr.6: 6167763-6167763 on Build GRCh38
rs149534714 Chr.6: 6167674-6167674 on Build GRCh38
rs529985469 Chr.6: 6167586-6167586 on Build GRCh38
rs372738301 Chr.6: 6167462-6167462 on Build GRCh38
rs773629601 Chr.6: 6167445-6167445 on Build GRCh38
rs775757965 Chr.6: 6167549-6167549 on Build GRCh38
rs752897176 Chr.6: 6167472-6167472 on Build GRCh38
rs140646752 Chr.6: 6167529-6167529 on Build GRCh38
rs1014064013 Chr.6: 6167390-6167390 on Build GRCh38
rs774882469 Chr.6: 6167618-6167618 on Build GRCh38
rs113778852 Chr.6: 6167619-6167619 on Build GRCh38
rs435048 Chr.6: 6167388-6167388 on Build GRCh38
rs766708117 Chr.6: 6167448-6167448 on Build GRCh38
rs1056217577 Chr.6: 6167369-6167369 on Build GRCh38
rs376690763 Chr.6: 6167559-6167559 on Build GRCh38
rs542437967 Chr.6: 6167355-6167355 on Build GRCh38
rs961470447 Chr.6: 6167753-6167753 on Build GRCh38
rs747650542 Chr.6: 6167602-6167602 on Build GRCh38
rs781376571 Chr.6: 6167342-6167348 on Build GRCh38
rs375508103 Chr.6: 6167417-6167417 on Build GRCh38
rs750450399 Chr.6: 6167668-6167668 on Build GRCh38
rs763508477 Chr.6: 6167446-6167446 on Build GRCh38
rs868771084 Chr.6: 6167643-6167643 on Build GRCh38
rs779898586 Chr.6: 6167508-6167508 on Build GRCh38
rs777280256 Chr.6: 6167534-6167534 on Build GRCh38
rs765171405 Chr.6: 6167566-6167566 on Build GRCh38
rs771433623 Chr.6: 6167532-6167532 on Build GRCh38
rs199801179 Chr.6: 6167473-6167473 on Build GRCh38
rs1019889513 Chr.6: 6167746-6167746 on Build GRCh38
rs759789208 Chr.6: 6167461-6167461 on Build GRCh38
rs369187276 Chr.6: 6167531-6167531 on Build GRCh38
rs958236716 Chr.6: 6167503-6167503 on Build GRCh38
rs746626360 Chr.6: 6167516-6167516 on Build GRCh38
rs772486160 Chr.6: 6167635-6167635 on Build GRCh38
rs1000746313 Chr.6: 6167606-6167606 on Build GRCh38
rs1009245289 Chr.6: 6167397-6167397 on Build GRCh38
rs767838396 Chr.6: 6167536-6167536 on Build GRCh38
rs757172838 Chr.6: 6167562-6167562 on Build GRCh38

View Full Product Details