Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17759200-17759723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
UniGene
Hs.94499
Gene Chromosome Location
Chr.6: 17759183-17987623 on Build GRCh38
Transcripts
NM_001105568.2 XM_017011188.1 XM_017011192.1

SNP Detail

SNP ID Chromosome Location
rs1036138083 Chr.6: 17759338-17759338 on Build GRCh38
rs772191660 Chr.6: 17759287-17759288 on Build GRCh38
rs141233146 Chr.6: 17759253-17759253 on Build GRCh38
rs575460322 Chr.6: 17759285-17759285 on Build GRCh38
rs919058911 Chr.6: 17759672-17759672 on Build GRCh38
rs1011701463 Chr.6: 17759493-17759493 on Build GRCh38
rs951254129 Chr.6: 17759361-17759364 on Build GRCh38
rs1051640398 Chr.6: 17759381-17759381 on Build GRCh38
rs2038744 Chr.6: 17759236-17759236 on Build GRCh38
rs891089261 Chr.6: 17759399-17759399 on Build GRCh38
rs915397644 Chr.6: 17759624-17759624 on Build GRCh38
rs991550729 Chr.6: 17759645-17759645 on Build GRCh38
rs1003085194 Chr.6: 17759530-17759530 on Build GRCh38
rs541598091 Chr.6: 17759254-17759254 on Build GRCh38
rs1045117203 Chr.6: 17759286-17759291 on Build GRCh38
rs981724609 Chr.6: 17759293-17759293 on Build GRCh38
rs1035308051 Chr.6: 17759351-17759352 on Build GRCh38
rs182985131 Chr.6: 17759455-17759455 on Build GRCh38
rs933230993 Chr.6: 17759379-17759379 on Build GRCh38
rs919132808 Chr.6: 17759349-17759349 on Build GRCh38
rs530412498 Chr.6: 17759667-17759667 on Build GRCh38
rs369635810 Chr.6: 17759286-17759287 on Build GRCh38
rs971825150 Chr.6: 17759670-17759670 on Build GRCh38
rs1084 Chr.6: 17759506-17759506 on Build GRCh38
rs760618976 Chr.6: 17759277-17759277 on Build GRCh38
rs770911508 Chr.6: 17759301-17759301 on Build GRCh38
rs776544318 Chr.6: 17759308-17759308 on Build GRCh38
rs1039734963 Chr.6: 17759484-17759484 on Build GRCh38
rs369165731 Chr.6: 17759320-17759320 on Build GRCh38
rs542555402 Chr.6: 17759315-17759315 on Build GRCh38
rs145428957 Chr.6: 17759285-17759286 on Build GRCh38
rs1016766337 Chr.6: 17759390-17759390 on Build GRCh38
rs908903187 Chr.6: 17759647-17759647 on Build GRCh38
rs769359074 Chr.6: 17759516-17759516 on Build GRCh38
rs967544398 Chr.6: 17759523-17759523 on Build GRCh38
rs962459566 Chr.6: 17759432-17759432 on Build GRCh38
rs908966735 Chr.6: 17759316-17759316 on Build GRCh38
rs530326680 Chr.6: 17759469-17759469 on Build GRCh38
rs565253585 Chr.6: 17759542-17759542 on Build GRCh38
rs920944431 Chr.6: 17759446-17759447 on Build GRCh38
rs569473547 Chr.6: 17759646-17759646 on Build GRCh38
rs1053055690 Chr.6: 17759586-17759586 on Build GRCh38
rs1035961903 Chr.6: 17759560-17759560 on Build GRCh38
rs560945600 Chr.6: 17759382-17759382 on Build GRCh38
rs376770783 Chr.6: 17759563-17759563 on Build GRCh38
rs975120155 Chr.6: 17759436-17759438 on Build GRCh38
rs146946922 Chr.6: 17759689-17759689 on Build GRCh38
rs893191158 Chr.6: 17759487-17759487 on Build GRCh38
rs187005958 Chr.6: 17759616-17759616 on Build GRCh38
rs958616221 Chr.6: 17759305-17759305 on Build GRCh38

View Full Product Details