Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17773277-17773778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
UniGene
Hs.94499
Gene Chromosome Location
Chr.6: 17759183-17987623 on Build GRCh38
Transcripts
NM_001105566.2 NM_001105567.2 NM_001105568.2 NM_022113.5 XM_017011188.1 XM_017011189.1 XM_017011190.1 XM_017011191.1 XM_017011192.1 XM_017011193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3860635 c.4260G>T p.R1420S Chr.6: 17773503-17773503 on Build GRCh38
4813150 c.4258A>G p.R1420G Chr.6: 17773505-17773505 on Build GRCh38
3623083 c.4240G>A p.D1414N Chr.6: 17773562-17773562 on Build GRCh38
5433345 c.4298G>A p.R1433K Chr.6: 17773504-17773504 on Build GRCh38
5433344 c.4259G>A p.R1420K Chr.6: 17773504-17773504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150226905 Chr.6: 17773612-17773612 on Build GRCh38
rs567303399 Chr.6: 17773601-17773601 on Build GRCh38
rs190339616 Chr.6: 17773717-17773717 on Build GRCh38
rs756696738 Chr.6: 17773524-17773524 on Build GRCh38
rs1015855641 Chr.6: 17773629-17773629 on Build GRCh38
rs377233808 Chr.6: 17773473-17773473 on Build GRCh38
rs755445981 Chr.6: 17773507-17773507 on Build GRCh38
rs773637052 Chr.6: 17773561-17773561 on Build GRCh38
rs565597769 Chr.6: 17773446-17773446 on Build GRCh38
rs866549736 Chr.6: 17773577-17773577 on Build GRCh38
rs756895730 Chr.6: 17773435-17773435 on Build GRCh38
rs577501049 Chr.6: 17773643-17773643 on Build GRCh38
rs184662378 Chr.6: 17773591-17773591 on Build GRCh38
rs1040961206 Chr.6: 17773416-17773416 on Build GRCh38
rs960318040 Chr.6: 17773311-17773311 on Build GRCh38
rs10560011 Chr.6: 17773692-17773694 on Build GRCh38
rs777478517 Chr.6: 17773741-17773741 on Build GRCh38
rs535396225 Chr.6: 17773662-17773663 on Build GRCh38
rs911742254 Chr.6: 17773696-17773696 on Build GRCh38
rs111902485 Chr.6: 17773433-17773433 on Build GRCh38
rs919662363 Chr.6: 17773329-17773329 on Build GRCh38
rs754313942 Chr.6: 17773496-17773496 on Build GRCh38
rs868349133 Chr.6: 17773369-17773369 on Build GRCh38
rs973093956 Chr.6: 17773665-17773665 on Build GRCh38
rs771426174 Chr.6: 17773567-17773567 on Build GRCh38
rs528691373 Chr.6: 17773384-17773384 on Build GRCh38
rs912206811 Chr.6: 17773701-17773701 on Build GRCh38
rs921874644 Chr.6: 17773740-17773740 on Build GRCh38
rs983250601 Chr.6: 17773691-17773691 on Build GRCh38
rs973883651 Chr.6: 17773666-17773666 on Build GRCh38
rs920334114 Chr.6: 17773684-17773684 on Build GRCh38
rs960533233 Chr.6: 17773608-17773608 on Build GRCh38
rs778884124 Chr.6: 17773554-17773554 on Build GRCh38
rs780376033 Chr.6: 17773607-17773607 on Build GRCh38
rs969731803 Chr.6: 17773637-17773637 on Build GRCh38
rs765819929 Chr.6: 17773605-17773605 on Build GRCh38
rs17689215 Chr.6: 17773559-17773559 on Build GRCh38
rs116328220 Chr.6: 17773472-17773472 on Build GRCh38
rs1026770351 Chr.6: 17773689-17773692 on Build GRCh38
rs780989870 Chr.6: 17773442-17773442 on Build GRCh38
rs538573941 Chr.6: 17773701-17773701 on Build GRCh38
rs370060319 Chr.6: 17773477-17773477 on Build GRCh38
rs915585696 Chr.6: 17773349-17773349 on Build GRCh38
rs373092082 Chr.6: 17773525-17773525 on Build GRCh38
rs763542054 Chr.6: 17773510-17773510 on Build GRCh38
rs944939540 Chr.6: 17773406-17773406 on Build GRCh38
rs140293391 Chr.6: 17773565-17773565 on Build GRCh38
rs944598180 Chr.6: 17773715-17773715 on Build GRCh38
rs779771179 Chr.6: 17773531-17773531 on Build GRCh38
rs755957437 Chr.6: 17773526-17773526 on Build GRCh38
rs920365872 Chr.6: 17773471-17773471 on Build GRCh38
rs1047635332 Chr.6: 17773484-17773484 on Build GRCh38
rs750086012 Chr.6: 17773618-17773618 on Build GRCh38
rs373383529 Chr.6: 17773610-17773610 on Build GRCh38
rs78281954 Chr.6: 17773492-17773492 on Build GRCh38
rs767801548 Chr.6: 17773491-17773491 on Build GRCh38
rs985117719 Chr.6: 17773373-17773373 on Build GRCh38
rs779758923 Chr.6: 17773510-17773510 on Build GRCh38
rs973788900 Chr.6: 17773308-17773312 on Build GRCh38
rs749464997 Chr.6: 17773688-17773688 on Build GRCh38
rs761162305 Chr.6: 17773493-17773493 on Build GRCh38
rs929043950 Chr.6: 17773685-17773685 on Build GRCh38
rs1021653694 Chr.6: 17773563-17773563 on Build GRCh38
rs866918817 Chr.6: 17773550-17773550 on Build GRCh38
rs866912983 Chr.6: 17773650-17773650 on Build GRCh38
rs761975221 Chr.6: 17773478-17773478 on Build GRCh38
rs768601692 Chr.6: 17773549-17773549 on Build GRCh38
rs992862398 Chr.6: 17773315-17773315 on Build GRCh38
rs749190992 Chr.6: 17773545-17773545 on Build GRCh38
rs758221996 Chr.6: 17773720-17773720 on Build GRCh38
rs367717849 Chr.6: 17773458-17773458 on Build GRCh38
rs751030387 Chr.6: 17773517-17773517 on Build GRCh38

View Full Product Details