Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46129847-46130365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP4
Entrez Gene ID
22875
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative)
Gene Aliases
NPP4
UniGene
Hs.643497
Gene Chromosome Location
Chr.6: 46129964-46146699 on Build GRCh38
Transcripts
NM_014936.4

SNP Detail

SNP ID Chromosome Location
rs944162290 Chr.6: 46129871-46129872 on Build GRCh38
rs567120073 Chr.6: 46130231-46130231 on Build GRCh38
rs1055028013 Chr.6: 46129894-46129894 on Build GRCh38
rs1053988070 Chr.6: 46130337-46130337 on Build GRCh38
rs569718664 Chr.6: 46130079-46130079 on Build GRCh38
rs770429672 Chr.6: 46130126-46130126 on Build GRCh38
rs990668378 Chr.6: 46130168-46130168 on Build GRCh38
rs1003484920 Chr.6: 46130109-46130109 on Build GRCh38
rs377286278 Chr.6: 46130151-46130151 on Build GRCh38
rs985832921 Chr.6: 46130204-46130204 on Build GRCh38
rs970624222 Chr.6: 46130209-46130209 on Build GRCh38
rs890708048 Chr.6: 46130100-46130100 on Build GRCh38
rs959492450 Chr.6: 46130141-46130141 on Build GRCh38
rs893560991 Chr.6: 46130099-46130099 on Build GRCh38
rs3734209 Chr.6: 46130032-46130032 on Build GRCh38
rs534620628 Chr.6: 46130238-46130238 on Build GRCh38
rs1038280286 Chr.6: 46129971-46129971 on Build GRCh38
rs902251044 Chr.6: 46129930-46129930 on Build GRCh38
rs949428622 Chr.6: 46130049-46130049 on Build GRCh38
rs906419676 Chr.6: 46130106-46130106 on Build GRCh38
rs7775082 Chr.6: 46129892-46129892 on Build GRCh38
rs578261937 Chr.6: 46130257-46130257 on Build GRCh38
rs1052468896 Chr.6: 46130098-46130098 on Build GRCh38
rs944313592 Chr.6: 46130062-46130062 on Build GRCh38
rs1007879741 Chr.6: 46129993-46129993 on Build GRCh38
rs1014752644 Chr.6: 46130131-46130131 on Build GRCh38
rs796568369 Chr.6: 46130306-46130308 on Build GRCh38
rs917927132 Chr.6: 46129932-46129932 on Build GRCh38
rs536776508 Chr.6: 46130088-46130088 on Build GRCh38
rs1003452250 Chr.6: 46130335-46130335 on Build GRCh38
rs551201286 Chr.6: 46130056-46130056 on Build GRCh38
rs547249031 Chr.6: 46129998-46129998 on Build GRCh38
rs1024881041 Chr.6: 46130179-46130179 on Build GRCh38
rs924110662 Chr.6: 46130221-46130221 on Build GRCh38
rs1020635313 Chr.6: 46130104-46130104 on Build GRCh38
rs917825239 Chr.6: 46130045-46130045 on Build GRCh38
rs902158020 Chr.6: 46130113-46130113 on Build GRCh38
rs932155600 Chr.6: 46130090-46130090 on Build GRCh38
rs906227461 Chr.6: 46130307-46130307 on Build GRCh38
rs922021390 Chr.6: 46130251-46130251 on Build GRCh38
rs1041180321 Chr.6: 46130083-46130083 on Build GRCh38
rs966003180 Chr.6: 46130214-46130214 on Build GRCh38
rs893498513 Chr.6: 46129957-46129957 on Build GRCh38
rs1000520317 Chr.6: 46130321-46130321 on Build GRCh38
rs182692460 Chr.6: 46129955-46129955 on Build GRCh38
rs932180825 Chr.6: 46130253-46130253 on Build GRCh38
rs983406640 Chr.6: 46130053-46130053 on Build GRCh38
rs764996289 Chr.6: 46130117-46130117 on Build GRCh38
rs372039460 Chr.6: 46130017-46130017 on Build GRCh38
rs1032609306 Chr.6: 46130029-46130029 on Build GRCh38
rs890589109 Chr.6: 46130274-46130274 on Build GRCh38
rs545621682 Chr.6: 46129946-46129946 on Build GRCh38
rs999256632 Chr.6: 46129931-46129931 on Build GRCh38
rs368496718 Chr.6: 46130302-46130302 on Build GRCh38
rs561097698 Chr.6: 46129976-46129976 on Build GRCh38
rs1001036594 Chr.6: 46130114-46130114 on Build GRCh38
rs372314984 Chr.6: 46130248-46130248 on Build GRCh38
rs942120893 Chr.6: 46130068-46130068 on Build GRCh38
rs557615687 Chr.6: 46130288-46130288 on Build GRCh38
rs1041964664 Chr.6: 46130300-46130300 on Build GRCh38
rs3734208 Chr.6: 46130021-46130021 on Build GRCh38
rs796838678 Chr.6: 46129935-46129935 on Build GRCh38
rs1020751471 Chr.6: 46130007-46130007 on Build GRCh38
rs1044728543 Chr.6: 46130086-46130086 on Build GRCh38
rs959209278 Chr.6: 46130031-46130031 on Build GRCh38
rs977008431 Chr.6: 46130220-46130221 on Build GRCh38
rs952833700 Chr.6: 46130197-46130197 on Build GRCh38
rs368139350 Chr.6: 46129972-46129972 on Build GRCh38
rs555464389 Chr.6: 46130101-46130101 on Build GRCh38
rs1047827747 Chr.6: 46129948-46129948 on Build GRCh38
rs894889981 Chr.6: 46130341-46130341 on Build GRCh38
rs955053500 Chr.6: 46129981-46129981 on Build GRCh38
rs983290608 Chr.6: 46130223-46130223 on Build GRCh38
rs902211839 Chr.6: 46130306-46130306 on Build GRCh38
rs147166974 Chr.6: 46129966-46129966 on Build GRCh38
rs961435157 Chr.6: 46130024-46130024 on Build GRCh38
rs982324742 Chr.6: 46130018-46130018 on Build GRCh38
rs1053508736 Chr.6: 46130095-46130095 on Build GRCh38
rs935464276 Chr.6: 46130225-46130225 on Build GRCh38
rs1051947041 Chr.6: 46130254-46130254 on Build GRCh38
rs961905488 Chr.6: 46130145-46130145 on Build GRCh38
rs947682608 Chr.6: 46130048-46130048 on Build GRCh38
rs946299740 Chr.6: 46130290-46130290 on Build GRCh38
rs545609609 Chr.6: 46130267-46130267 on Build GRCh38
rs1006377723 Chr.6: 46129958-46129958 on Build GRCh38
rs1032080764 Chr.6: 46130116-46130116 on Build GRCh38
rs528501189 Chr.6: 46129997-46129997 on Build GRCh38
rs537598813 Chr.6: 46129974-46129974 on Build GRCh38

View Full Product Details