Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89926802-89927341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH2
Entrez Gene ID
60468
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
BTBD25
UniGene
Hs.269764
Gene Chromosome Location
Chr.6: 89926528-90296912 on Build GRCh38
Transcripts
NM_001170794.1 NM_021813.3 XM_005248758.4 XM_005248759.4 XM_017011156.1 XM_017011157.1 XM_017011158.1 XM_017011159.1 XM_017011160.1 XM_017011161.1 XM_017011162.1 XM_017011163.1 XM_017011164.1 XM_017011166.1

SNP Detail

SNP ID Chromosome Location
rs564201419 Chr.6: 89926861-89926861 on Build GRCh38
rs768314248 Chr.6: 89927089-89927089 on Build GRCh38
rs904794103 Chr.6: 89927138-89927138 on Build GRCh38
rs945986985 Chr.6: 89926882-89926882 on Build GRCh38
rs540179180 Chr.6: 89926976-89926976 on Build GRCh38
rs749010081 Chr.6: 89926880-89926880 on Build GRCh38
rs973365452 Chr.6: 89927270-89927270 on Build GRCh38
rs907852108 Chr.6: 89926906-89926906 on Build GRCh38
rs917432932 Chr.6: 89927084-89927084 on Build GRCh38
rs57331601 Chr.6: 89927077-89927077 on Build GRCh38
rs1028732608 Chr.6: 89927169-89927169 on Build GRCh38
rs1039347802 Chr.6: 89927162-89927162 on Build GRCh38
rs927320307 Chr.6: 89927291-89927291 on Build GRCh38
rs748095358 Chr.6: 89927254-89927254 on Build GRCh38
rs901537533 Chr.6: 89927159-89927159 on Build GRCh38
rs895275713 Chr.6: 89926947-89926947 on Build GRCh38
rs45610136 Chr.6: 89926926-89926926 on Build GRCh38
rs983135316 Chr.6: 89926905-89926905 on Build GRCh38
rs80159519 Chr.6: 89926832-89926832 on Build GRCh38
rs559630386 Chr.6: 89926977-89926977 on Build GRCh38
rs886687475 Chr.6: 89927095-89927095 on Build GRCh38
rs1029486431 Chr.6: 89927212-89927212 on Build GRCh38
rs946299458 Chr.6: 89927096-89927096 on Build GRCh38
rs973414751 Chr.6: 89926929-89926929 on Build GRCh38
rs1004067812 Chr.6: 89927225-89927225 on Build GRCh38
rs899589234 Chr.6: 89927187-89927187 on Build GRCh38
rs755675627 Chr.6: 89926844-89926844 on Build GRCh38
rs927330356 Chr.6: 89926961-89926961 on Build GRCh38
rs981359684 Chr.6: 89927269-89927269 on Build GRCh38
rs993343798 Chr.6: 89927316-89927316 on Build GRCh38
rs866077603 Chr.6: 89926885-89926885 on Build GRCh38
rs781118999 Chr.6: 89927277-89927277 on Build GRCh38
rs868692526 Chr.6: 89926848-89926848 on Build GRCh38
rs779764647 Chr.6: 89927275-89927275 on Build GRCh38
rs58265329 Chr.6: 89926918-89926918 on Build GRCh38
rs998376755 Chr.6: 89927205-89927205 on Build GRCh38
rs913130188 Chr.6: 89926865-89926865 on Build GRCh38
rs548449825 Chr.6: 89927198-89927198 on Build GRCh38
rs772065905 Chr.6: 89927294-89927294 on Build GRCh38
rs995002973 Chr.6: 89926960-89926961 on Build GRCh38
rs545505164 Chr.6: 89927241-89927241 on Build GRCh38
rs182150964 Chr.6: 89926842-89926842 on Build GRCh38
rs999783092 Chr.6: 89927153-89927153 on Build GRCh38
rs1042336310 Chr.6: 89927118-89927118 on Build GRCh38
rs3798791 Chr.6: 89927220-89927220 on Build GRCh38
rs60636991 Chr.6: 89927273-89927273 on Build GRCh38
rs1049716904 Chr.6: 89926963-89926963 on Build GRCh38

View Full Product Details