Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89930405-89930914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH2
Entrez Gene ID
60468
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
BTBD25
UniGene
Hs.269764
Gene Chromosome Location
Chr.6: 89926528-90296912 on Build GRCh38
Transcripts
NM_001170794.1 NM_021813.3 XM_005248758.4 XM_005248759.4 XM_017011156.1 XM_017011157.1 XM_017011158.1 XM_017011159.1 XM_017011160.1 XM_017011161.1 XM_017011162.1 XM_017011163.1 XM_017011164.1 XM_017011166.1

SNP Detail

SNP ID Chromosome Location
rs952669001 Chr.6: 89930603-89930603 on Build GRCh38
rs968299697 Chr.6: 89930463-89930463 on Build GRCh38
rs1052035287 Chr.6: 89930457-89930457 on Build GRCh38
rs980610591 Chr.6: 89930546-89930546 on Build GRCh38
rs984136872 Chr.6: 89930612-89930612 on Build GRCh38
rs928181212 Chr.6: 89930688-89930688 on Build GRCh38
rs927352148 Chr.6: 89930850-89930850 on Build GRCh38
rs778003219 Chr.6: 89930723-89930723 on Build GRCh38
rs1038526263 Chr.6: 89930574-89930574 on Build GRCh38
rs956649836 Chr.6: 89930508-89930508 on Build GRCh38
rs752372222 Chr.6: 89930724-89930724 on Build GRCh38
rs946350679 Chr.6: 89930736-89930736 on Build GRCh38
rs915506669 Chr.6: 89930735-89930735 on Build GRCh38
rs887703735 Chr.6: 89930888-89930888 on Build GRCh38
rs2289577 Chr.6: 89930772-89930772 on Build GRCh38
rs145987745 Chr.6: 89930867-89930867 on Build GRCh38
rs1027545589 Chr.6: 89930880-89930880 on Build GRCh38
rs534526306 Chr.6: 89930501-89930501 on Build GRCh38
rs946403152 Chr.6: 89930524-89930524 on Build GRCh38
rs993443890 Chr.6: 89930580-89930580 on Build GRCh38
rs1040597892 Chr.6: 89930791-89930791 on Build GRCh38
rs972857365 Chr.6: 89930632-89930657 on Build GRCh38
rs888860350 Chr.6: 89930768-89930768 on Build GRCh38
rs930237331 Chr.6: 89930594-89930594 on Build GRCh38
rs982677883 Chr.6: 89930643-89930643 on Build GRCh38
rs979290558 Chr.6: 89930483-89930483 on Build GRCh38
rs567904349 Chr.6: 89930539-89930539 on Build GRCh38
rs927453982 Chr.6: 89930556-89930556 on Build GRCh38
rs993350496 Chr.6: 89930873-89930873 on Build GRCh38
rs1050098434 Chr.6: 89930744-89930744 on Build GRCh38
rs956573801 Chr.6: 89930745-89930745 on Build GRCh38
rs567193988 Chr.6: 89930500-89930500 on Build GRCh38
rs1030906764 Chr.6: 89930485-89930485 on Build GRCh38
rs888879586 Chr.6: 89930541-89930541 on Build GRCh38
rs903631219 Chr.6: 89930734-89930735 on Build GRCh38
rs897687065 Chr.6: 89930836-89930836 on Build GRCh38
rs1024108563 Chr.6: 89930625-89930625 on Build GRCh38
rs763500619 Chr.6: 89930502-89930502 on Build GRCh38
rs1018083450 Chr.6: 89930446-89930446 on Build GRCh38
rs1009106657 Chr.6: 89930542-89930542 on Build GRCh38
rs1013509910 Chr.6: 89930748-89930748 on Build GRCh38
rs538007584 Chr.6: 89930561-89930561 on Build GRCh38
rs1039849889 Chr.6: 89930659-89930659 on Build GRCh38
rs764879626 Chr.6: 89930629-89930629 on Build GRCh38
rs1055683 Chr.6: 89930525-89930525 on Build GRCh38
rs1025633344 Chr.6: 89930589-89930589 on Build GRCh38
rs1018819064 Chr.6: 89930550-89930550 on Build GRCh38
rs969504367 Chr.6: 89930642-89930642 on Build GRCh38
rs556627588 Chr.6: 89930564-89930564 on Build GRCh38
rs988327224 Chr.6: 89930733-89930733 on Build GRCh38
rs142567155 Chr.6: 89930445-89930445 on Build GRCh38

View Full Product Details