Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138725431-138725977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GVQW2
Entrez Gene ID
100507462
Gene Name
GVQW motif containing 2
Gene Aliases
-
UniGene
Hs.571054
Gene Chromosome Location
Chr.6: 138725211-138773679 on Build GRCh38
Transcripts
NM_001242740.1

SNP Detail

SNP ID Chromosome Location
rs1044684832 Chr.6: 138725473-138725473 on Build GRCh38
rs137932681 Chr.6: 138725669-138725669 on Build GRCh38
rs947753393 Chr.6: 138725464-138725464 on Build GRCh38
rs982209489 Chr.6: 138725747-138725747 on Build GRCh38
rs574856593 Chr.6: 138725846-138725846 on Build GRCh38
rs1052708926 Chr.6: 138725490-138725490 on Build GRCh38
rs770112168 Chr.6: 138725754-138725754 on Build GRCh38
rs1018248142 Chr.6: 138725576-138725576 on Build GRCh38
rs930843828 Chr.6: 138725807-138725807 on Build GRCh38
rs945436837 Chr.6: 138725477-138725477 on Build GRCh38
rs775321797 Chr.6: 138725845-138725845 on Build GRCh38
rs1007448349 Chr.6: 138725876-138725876 on Build GRCh38
rs920560913 Chr.6: 138725929-138725929 on Build GRCh38
rs1040479238 Chr.6: 138725618-138725618 on Build GRCh38
rs184024631 Chr.6: 138725794-138725794 on Build GRCh38
rs568447677 Chr.6: 138725653-138725653 on Build GRCh38
rs150305035 Chr.6: 138725608-138725608 on Build GRCh38
rs1035097979 Chr.6: 138725910-138725910 on Build GRCh38
rs143386765 Chr.6: 138725894-138725894 on Build GRCh38
rs1000584044 Chr.6: 138725486-138725486 on Build GRCh38
rs944495254 Chr.6: 138725834-138725834 on Build GRCh38
rs529597028 Chr.6: 138725538-138725538 on Build GRCh38
rs918664736 Chr.6: 138725614-138725614 on Build GRCh38
rs997738647 Chr.6: 138725504-138725504 on Build GRCh38
rs992723754 Chr.6: 138725716-138725716 on Build GRCh38
rs770067870 Chr.6: 138725926-138725926 on Build GRCh38
rs1004058653 Chr.6: 138725896-138725896 on Build GRCh38
rs796701338 Chr.6: 138725915-138725915 on Build GRCh38
rs1020034930 Chr.6: 138725698-138725698 on Build GRCh38
rs186732169 Chr.6: 138725497-138725497 on Build GRCh38
rs962795094 Chr.6: 138725583-138725583 on Build GRCh38
rs1052212903 Chr.6: 138725832-138725832 on Build GRCh38
rs889038106 Chr.6: 138725856-138725856 on Build GRCh38
rs553867803 Chr.6: 138725835-138725835 on Build GRCh38
rs746669528 Chr.6: 138725921-138725921 on Build GRCh38
rs536673299 Chr.6: 138725891-138725893 on Build GRCh38
rs140184874 Chr.6: 138725537-138725537 on Build GRCh38
rs1006913460 Chr.6: 138725566-138725566 on Build GRCh38
rs1010073286 Chr.6: 138725859-138725859 on Build GRCh38
rs569202495 Chr.6: 138725759-138725759 on Build GRCh38
rs191497948 Chr.6: 138725791-138725791 on Build GRCh38
rs898535101 Chr.6: 138725940-138725940 on Build GRCh38
rs936329688 Chr.6: 138725820-138725820 on Build GRCh38
rs566866465 Chr.6: 138725580-138725580 on Build GRCh38
rs1055376867 Chr.6: 138725512-138725512 on Build GRCh38
rs1047935205 Chr.6: 138725814-138725814 on Build GRCh38
rs971251592 Chr.6: 138725892-138725892 on Build GRCh38
rs1010040704 Chr.6: 138725686-138725687 on Build GRCh38
rs957254053 Chr.6: 138725684-138725684 on Build GRCh38
rs903632452 Chr.6: 138725481-138725481 on Build GRCh38
rs201722783 Chr.6: 138725454-138725456 on Build GRCh38
rs1047902671 Chr.6: 138725571-138725571 on Build GRCh38
rs900164606 Chr.6: 138725635-138725635 on Build GRCh38
rs918183316 Chr.6: 138725720-138725720 on Build GRCh38

View Full Product Details