Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138726263-138726775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GVQW2
Entrez Gene ID
100507462
Gene Name
GVQW motif containing 2
Gene Aliases
-
UniGene
Hs.571054
Gene Chromosome Location
Chr.6: 138725211-138773679 on Build GRCh38
Transcripts
NM_001242740.1

SNP Detail

SNP ID Chromosome Location
rs929176799 Chr.6: 138726601-138726601 on Build GRCh38
rs943213187 Chr.6: 138726543-138726543 on Build GRCh38
rs78431885 Chr.6: 138726490-138726490 on Build GRCh38
rs1013590987 Chr.6: 138726305-138726305 on Build GRCh38
rs546200754 Chr.6: 138726494-138726494 on Build GRCh38
rs77639083 Chr.6: 138726297-138726297 on Build GRCh38
rs887134203 Chr.6: 138726608-138726608 on Build GRCh38
rs568612244 Chr.6: 138726453-138726453 on Build GRCh38
rs938564672 Chr.6: 138726616-138726616 on Build GRCh38
rs931783361 Chr.6: 138726536-138726536 on Build GRCh38
rs144543660 Chr.6: 138726640-138726640 on Build GRCh38
rs966674118 Chr.6: 138726473-138726473 on Build GRCh38
rs980890680 Chr.6: 138726411-138726411 on Build GRCh38
rs78165675 Chr.6: 138726341-138726341 on Build GRCh38
rs549091364 Chr.6: 138726471-138726471 on Build GRCh38
rs181961365 Chr.6: 138726508-138726508 on Build GRCh38
rs887203292 Chr.6: 138726298-138726298 on Build GRCh38
rs1002332915 Chr.6: 138726680-138726680 on Build GRCh38
rs1032606780 Chr.6: 138726433-138726433 on Build GRCh38
rs913523944 Chr.6: 138726455-138726455 on Build GRCh38
rs945701663 Chr.6: 138726575-138726575 on Build GRCh38
rs914262213 Chr.6: 138726565-138726565 on Build GRCh38
rs989854145 Chr.6: 138726544-138726544 on Build GRCh38
rs538086984 Chr.6: 138726324-138726324 on Build GRCh38
rs1032901820 Chr.6: 138726705-138726705 on Build GRCh38
rs773766539 Chr.6: 138726298-138726298 on Build GRCh38
rs905255151 Chr.6: 138726642-138726642 on Build GRCh38
rs555144126 Chr.6: 138726371-138726371 on Build GRCh38
rs527437998 Chr.6: 138726661-138726661 on Build GRCh38
rs753703646 Chr.6: 138726684-138726684 on Build GRCh38
rs867614969 Chr.6: 138726357-138726357 on Build GRCh38
rs149310240 Chr.6: 138726602-138726602 on Build GRCh38
rs553022925 Chr.6: 138726297-138726297 on Build GRCh38
rs541697461 Chr.6: 138726631-138726631 on Build GRCh38
rs1037510910 Chr.6: 138726550-138726550 on Build GRCh38
rs1012362785 Chr.6: 138726647-138726647 on Build GRCh38
rs763935385 Chr.6: 138726678-138726678 on Build GRCh38
rs958506426 Chr.6: 138726451-138726451 on Build GRCh38
rs752368342 Chr.6: 138726492-138726492 on Build GRCh38
rs138881088 Chr.6: 138726470-138726470 on Build GRCh38
rs920541804 Chr.6: 138726570-138726570 on Build GRCh38
rs1027728890 Chr.6: 138726648-138726648 on Build GRCh38

View Full Product Details