Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161347583-161348129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

SNP Detail

SNP ID Chromosome Location
rs564009047 Chr.6: 161348030-161348030 on Build GRCh38
rs886180554 Chr.6: 161348051-161348051 on Build GRCh38
rs911904884 Chr.6: 161347913-161347913 on Build GRCh38
rs868117815 Chr.6: 161347624-161347624 on Build GRCh38
rs1122470 Chr.6: 161348078-161348078 on Build GRCh38
rs371405899 Chr.6: 161347648-161347648 on Build GRCh38
rs886061229 Chr.6: 161347656-161347656 on Build GRCh38
rs149239597 Chr.6: 161347905-161347905 on Build GRCh38
rs117341007 Chr.6: 161347607-161347607 on Build GRCh38
rs986053451 Chr.6: 161347775-161347775 on Build GRCh38
rs1036578009 Chr.6: 161347614-161347623 on Build GRCh38
rs541101637 Chr.6: 161347704-161347705 on Build GRCh38
rs12215397 Chr.6: 161347669-161347669 on Build GRCh38
rs903812654 Chr.6: 161348100-161348100 on Build GRCh38
rs541584413 Chr.6: 161347710-161347710 on Build GRCh38
rs966472244 Chr.6: 161347852-161347852 on Build GRCh38
rs563061113 Chr.6: 161348018-161348018 on Build GRCh38
rs920730915 Chr.6: 161347993-161347993 on Build GRCh38
rs934683487 Chr.6: 161347620-161347620 on Build GRCh38
rs967552813 Chr.6: 161347696-161347696 on Build GRCh38
rs527357513 Chr.6: 161347800-161347800 on Build GRCh38
rs1012194436 Chr.6: 161347879-161347879 on Build GRCh38
rs763588687 Chr.6: 161347613-161347614 on Build GRCh38
rs768016642 Chr.6: 161348065-161348065 on Build GRCh38
rs191130749 Chr.6: 161347745-161347745 on Build GRCh38
rs939124630 Chr.6: 161348068-161348068 on Build GRCh38
rs759912804 Chr.6: 161347926-161347926 on Build GRCh38
rs919086054 Chr.6: 161347615-161347619 on Build GRCh38
rs138660139 Chr.6: 161347882-161347882 on Build GRCh38
rs200119552 Chr.6: 161347707-161347707 on Build GRCh38
rs764413910 Chr.6: 161347865-161347865 on Build GRCh38
rs903465113 Chr.6: 161348042-161348042 on Build GRCh38
rs886061228 Chr.6: 161347625-161347625 on Build GRCh38
rs912139846 Chr.6: 161347622-161347622 on Build GRCh38
rs1031281346 Chr.6: 161348103-161348103 on Build GRCh38
rs1052237818 Chr.6: 161347621-161347621 on Build GRCh38
rs12215447 Chr.6: 161347736-161347736 on Build GRCh38
rs943653651 Chr.6: 161347690-161347690 on Build GRCh38
rs1022228804 Chr.6: 161348085-161348085 on Build GRCh38
rs774851231 Chr.6: 161347920-161347920 on Build GRCh38
rs1009670442 Chr.6: 161347966-161347966 on Build GRCh38
rs752358219 Chr.6: 161348076-161348076 on Build GRCh38
rs68121389 Chr.6: 161347619-161347619 on Build GRCh38
rs1048399777 Chr.6: 161348035-161348035 on Build GRCh38
rs575823392 Chr.6: 161347687-161347687 on Build GRCh38
rs1047483841 Chr.6: 161347807-161347807 on Build GRCh38
rs200780698 Chr.6: 161347811-161347812 on Build GRCh38
rs553992232 Chr.6: 161347820-161347820 on Build GRCh38
rs35851380 Chr.6: 161347810-161347811 on Build GRCh38
rs1035130429 Chr.6: 161347842-161347842 on Build GRCh38
rs886064547 Chr.6: 161347810-161347810 on Build GRCh38
rs896140352 Chr.6: 161347921-161347921 on Build GRCh38
rs978805894 Chr.6: 161347890-161347890 on Build GRCh38
rs896046959 Chr.6: 161347660-161347660 on Build GRCh38
rs543147935 Chr.6: 161347908-161347908 on Build GRCh38
rs75529362 Chr.6: 161348102-161348102 on Build GRCh38
rs886061230 Chr.6: 161348077-161348077 on Build GRCh38
rs1003206040 Chr.6: 161347825-161347825 on Build GRCh38
rs1049376713 Chr.6: 161347900-161347900 on Build GRCh38
rs867195540 Chr.6: 161347613-161347613 on Build GRCh38
rs924717939 Chr.6: 161347898-161347898 on Build GRCh38
rs930817992 Chr.6: 161347997-161347997 on Build GRCh38
rs556470130 Chr.6: 161347678-161347678 on Build GRCh38
rs1056115186 Chr.6: 161348069-161348069 on Build GRCh38
rs185919757 Chr.6: 161347709-161347709 on Build GRCh38
rs930497988 Chr.6: 161347615-161347615 on Build GRCh38
rs899360537 Chr.6: 161347753-161347753 on Build GRCh38
rs924577572 Chr.6: 161347611-161347611 on Build GRCh38
rs994519081 Chr.6: 161347806-161347806 on Build GRCh38
rs545759459 Chr.6: 161347623-161347623 on Build GRCh38
rs953437558 Chr.6: 161348076-161348076 on Build GRCh38
rs919209009 Chr.6: 161347884-161347884 on Build GRCh38
rs116309008 Chr.6: 161347912-161347912 on Build GRCh38
rs972509346 Chr.6: 161348101-161348101 on Build GRCh38
rs967885779 Chr.6: 161347885-161347885 on Build GRCh38
rs1048801792 Chr.6: 161347618-161347618 on Build GRCh38
rs1056144823 Chr.6: 161347625-161347625 on Build GRCh38
rs937762581 Chr.6: 161347906-161347906 on Build GRCh38
rs541135564 Chr.6: 161347867-161347867 on Build GRCh38

View Full Product Details