Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161348430-161348939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

SNP Detail

SNP ID Chromosome Location
rs555269742 Chr.6: 161348817-161348817 on Build GRCh38
rs538716074 Chr.6: 161348903-161348903 on Build GRCh38
rs779851186 Chr.6: 161348498-161348498 on Build GRCh38
rs371797205 Chr.6: 161348637-161348637 on Build GRCh38
rs191142442 Chr.6: 161348742-161348742 on Build GRCh38
rs991951798 Chr.6: 161348794-161348794 on Build GRCh38
rs143810946 Chr.6: 161348734-161348734 on Build GRCh38
rs977759553 Chr.6: 161348486-161348486 on Build GRCh38
rs559966988 Chr.6: 161348666-161348666 on Build GRCh38
rs529580837 Chr.6: 161348567-161348567 on Build GRCh38
rs534857414 Chr.6: 161348831-161348831 on Build GRCh38
rs543363389 Chr.6: 161348496-161348496 on Build GRCh38
rs567890129 Chr.6: 161348549-161348549 on Build GRCh38
rs182568156 Chr.6: 161348752-161348752 on Build GRCh38
rs567338994 Chr.6: 161348775-161348775 on Build GRCh38
rs924952261 Chr.6: 161348489-161348489 on Build GRCh38
rs577554278 Chr.6: 161348487-161348487 on Build GRCh38
rs200695402 Chr.6: 161348889-161348890 on Build GRCh38
rs748008270 Chr.6: 161348517-161348517 on Build GRCh38
rs564250931 Chr.6: 161348477-161348477 on Build GRCh38
rs994003191 Chr.6: 161348629-161348629 on Build GRCh38
rs985101238 Chr.6: 161348849-161348849 on Build GRCh38
rs886061231 Chr.6: 161348629-161348629 on Build GRCh38
rs11961229 Chr.6: 161348637-161348637 on Build GRCh38
rs1040517912 Chr.6: 161348874-161348874 on Build GRCh38
rs889401575 Chr.6: 161348499-161348499 on Build GRCh38
rs947816105 Chr.6: 161348578-161348578 on Build GRCh38
rs966455491 Chr.6: 161348811-161348811 on Build GRCh38
rs563296835 Chr.6: 161348521-161348521 on Build GRCh38
rs557674255 Chr.6: 161348838-161348838 on Build GRCh38
rs77926621 Chr.6: 161348847-161348847 on Build GRCh38
rs952153860 Chr.6: 161348843-161348843 on Build GRCh38
rs1015592643 Chr.6: 161348853-161348853 on Build GRCh38
rs537153723 Chr.6: 161348746-161348746 on Build GRCh38
rs991883389 Chr.6: 161348518-161348518 on Build GRCh38
rs748283584 Chr.6: 161348678-161348678 on Build GRCh38
rs1039544349 Chr.6: 161348753-161348753 on Build GRCh38
rs140107485 Chr.6: 161348713-161348713 on Build GRCh38
rs961026516 Chr.6: 161348856-161348856 on Build GRCh38
rs571904443 Chr.6: 161348818-161348818 on Build GRCh38
rs3734464 Chr.6: 161348803-161348803 on Build GRCh38
rs887756626 Chr.6: 161348647-161348647 on Build GRCh38
rs893845769 Chr.6: 161348645-161348645 on Build GRCh38
rs11961237 Chr.6: 161348692-161348692 on Build GRCh38

View Full Product Details