Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161386665-161387078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6514613 c.1167G>T p.Q389H Chr.6: 161386794-161386794 on Build GRCh38
6952225 c.1092C>A p.F364L Chr.6: 161386869-161386869 on Build GRCh38
3622659 c.1111G>A p.A371T Chr.6: 161386850-161386850 on Build GRCh38
3622658 c.1132A>G p.S378G Chr.6: 161386829-161386829 on Build GRCh38
4998786 c.1154G>A p.G385E Chr.6: 161386807-161386807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954879263 Chr.6: 161386713-161386713 on Build GRCh38
rs750834152 Chr.6: 161386759-161386759 on Build GRCh38
rs138360641 Chr.6: 161386935-161386935 on Build GRCh38
rs982012304 Chr.6: 161386764-161386764 on Build GRCh38
rs56092260 Chr.6: 161386865-161386865 on Build GRCh38
rs765372014 Chr.6: 161386895-161386895 on Build GRCh38
rs754463648 Chr.6: 161386870-161386870 on Build GRCh38
rs1022551831 Chr.6: 161386992-161386992 on Build GRCh38
rs564189761 Chr.6: 161386702-161386702 on Build GRCh38
rs562644112 Chr.6: 161386732-161386732 on Build GRCh38
rs766523228 Chr.6: 161386775-161386775 on Build GRCh38
rs753025419 Chr.6: 161386802-161386802 on Build GRCh38
rs780804768 Chr.6: 161386919-161386919 on Build GRCh38
rs370271614 Chr.6: 161386837-161386837 on Build GRCh38
rs539788992 Chr.6: 161387000-161387000 on Build GRCh38
rs576718166 Chr.6: 161386714-161386714 on Build GRCh38
rs756949723 Chr.6: 161386803-161386803 on Build GRCh38
rs761813150 Chr.6: 161386824-161386824 on Build GRCh38
rs762546735 Chr.6: 161386745-161386745 on Build GRCh38
rs996990326 Chr.6: 161387018-161387018 on Build GRCh38
rs1007722672 Chr.6: 161387004-161387004 on Build GRCh38
rs773654456 Chr.6: 161386861-161386861 on Build GRCh38
rs890742500 Chr.6: 161386996-161386996 on Build GRCh38
rs543017610 Chr.6: 161386721-161386721 on Build GRCh38
rs774091145 Chr.6: 161386747-161386747 on Build GRCh38
rs753986748 Chr.6: 161386776-161386776 on Build GRCh38
rs927762047 Chr.6: 161386768-161386768 on Build GRCh38
rs113690635 Chr.6: 161386703-161386703 on Build GRCh38
rs908566232 Chr.6: 161386734-161386734 on Build GRCh38
rs755151346 Chr.6: 161386791-161386791 on Build GRCh38
rs1011276006 Chr.6: 161386974-161386974 on Build GRCh38
rs200194930 Chr.6: 161386898-161386898 on Build GRCh38
rs760179604 Chr.6: 161387043-161387043 on Build GRCh38
rs775043892 Chr.6: 161386922-161386922 on Build GRCh38
rs1801582 Chr.6: 161386823-161386823 on Build GRCh38
rs181818478 Chr.6: 161386781-161386781 on Build GRCh38
rs561808120 Chr.6: 161386756-161386756 on Build GRCh38
rs759676993 Chr.6: 161386873-161386873 on Build GRCh38
rs1007279877 Chr.6: 161386718-161386718 on Build GRCh38
rs185908514 Chr.6: 161386701-161386701 on Build GRCh38
rs1052022463 Chr.6: 161386972-161386972 on Build GRCh38
rs748763421 Chr.6: 161386829-161386829 on Build GRCh38
rs756753053 Chr.6: 161386724-161386724 on Build GRCh38
rs551388732 Chr.6: 161386730-161386730 on Build GRCh38
rs773860689 Chr.6: 161386845-161386845 on Build GRCh38
rs548103938 Chr.6: 161386746-161386746 on Build GRCh38
rs771353933 Chr.6: 161386785-161386785 on Build GRCh38
rs761213043 Chr.6: 161386864-161386864 on Build GRCh38
rs1052749492 Chr.6: 161386860-161386860 on Build GRCh38
rs1030256291 Chr.6: 161387053-161387053 on Build GRCh38
rs377235301 Chr.6: 161386760-161386760 on Build GRCh38
rs373905109 Chr.6: 161386755-161386755 on Build GRCh38
rs761535398 Chr.6: 161386848-161386848 on Build GRCh38
rs775013357 Chr.6: 161386743-161386743 on Build GRCh38
rs191114756 Chr.6: 161386733-161386733 on Build GRCh38
rs781046528 Chr.6: 161386805-161386805 on Build GRCh38
rs201427980 Chr.6: 161387046-161387046 on Build GRCh38
rs375036403 Chr.6: 161386849-161386849 on Build GRCh38
rs547404732 Chr.6: 161386798-161386798 on Build GRCh38
rs758682179 Chr.6: 161386917-161386917 on Build GRCh38
rs771555178 Chr.6: 161386878-161386878 on Build GRCh38
rs948581056 Chr.6: 161386799-161386799 on Build GRCh38

View Full Product Details