Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 162053986-162054527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6979315 c.535-2_535-1insA p.? Chr.6: 162054175-162054176 on Build GRCh38
6940929 c.587G>C p.C196S Chr.6: 162054122-162054122 on Build GRCh38
257666 c.603C>T p.C201C Chr.6: 162054106-162054106 on Build GRCh38
4859145 c.556G>A p.V186I Chr.6: 162054153-162054153 on Build GRCh38
4006197 c.587G>A p.C196Y Chr.6: 162054122-162054122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151305042 Chr.6: 162054163-162054163 on Build GRCh38
rs977940711 Chr.6: 162054181-162054182 on Build GRCh38
rs758593834 Chr.6: 162054186-162054188 on Build GRCh38
rs112448533 Chr.6: 162054019-162054019 on Build GRCh38
rs72480421 Chr.6: 162054109-162054109 on Build GRCh38
rs765360163 Chr.6: 162054063-162054067 on Build GRCh38
rs913971177 Chr.6: 162054247-162054247 on Build GRCh38
rs56274447 Chr.6: 162054174-162054174 on Build GRCh38
rs761430731 Chr.6: 162054144-162054144 on Build GRCh38
rs976607110 Chr.6: 162054279-162054279 on Build GRCh38
rs377344238 Chr.6: 162054154-162054154 on Build GRCh38
rs527346336 Chr.6: 162054424-162054424 on Build GRCh38
rs941005559 Chr.6: 162054367-162054367 on Build GRCh38
rs544944835 Chr.6: 162054389-162054389 on Build GRCh38
rs368945837 Chr.6: 162054198-162054198 on Build GRCh38
rs747262712 Chr.6: 162054195-162054195 on Build GRCh38
rs1050072208 Chr.6: 162054338-162054338 on Build GRCh38
rs745637077 Chr.6: 162054130-162054130 on Build GRCh38
rs768628018 Chr.6: 162054486-162054486 on Build GRCh38
rs994342567 Chr.6: 162054027-162054027 on Build GRCh38
rs555771711 Chr.6: 162054290-162054290 on Build GRCh38
rs199810018 Chr.6: 162054147-162054147 on Build GRCh38
rs920181545 Chr.6: 162054358-162054358 on Build GRCh38
rs530632014 Chr.6: 162054137-162054137 on Build GRCh38
rs748352072 Chr.6: 162054113-162054113 on Build GRCh38
rs370479735 Chr.6: 162054173-162054173 on Build GRCh38
rs745916231 Chr.6: 162054412-162054412 on Build GRCh38
rs200985148 Chr.6: 162054095-162054095 on Build GRCh38
rs903752654 Chr.6: 162054452-162054452 on Build GRCh38
rs992213627 Chr.6: 162054204-162054204 on Build GRCh38
rs9456735 Chr.6: 162054135-162054135 on Build GRCh38
rs375005083 Chr.6: 162054224-162054224 on Build GRCh38
rs201039350 Chr.6: 162054183-162054183 on Build GRCh38
rs756411170 Chr.6: 162054155-162054155 on Build GRCh38
rs143737665 Chr.6: 162054326-162054326 on Build GRCh38
rs1022408570 Chr.6: 162054444-162054444 on Build GRCh38
rs777801875 Chr.6: 162054193-162054193 on Build GRCh38
rs958049428 Chr.6: 162054441-162054441 on Build GRCh38
rs888068912 Chr.6: 162054106-162054106 on Build GRCh38
rs981392307 Chr.6: 162054194-162054194 on Build GRCh38
rs994106388 Chr.6: 162054382-162054382 on Build GRCh38
rs755495887 Chr.6: 162054168-162054168 on Build GRCh38
rs768030209 Chr.6: 162054066-162054066 on Build GRCh38
rs201206485 Chr.6: 162054118-162054118 on Build GRCh38
rs774409433 Chr.6: 162054216-162054216 on Build GRCh38
rs763650901 Chr.6: 162054138-162054138 on Build GRCh38
rs9365364 Chr.6: 162054402-162054402 on Build GRCh38
rs1002473274 Chr.6: 162054433-162054433 on Build GRCh38
rs775450886 Chr.6: 162054199-162054199 on Build GRCh38
rs758565015 Chr.6: 162054185-162054185 on Build GRCh38
rs1034325356 Chr.6: 162054440-162054440 on Build GRCh38
rs1033693594 Chr.6: 162054196-162054196 on Build GRCh38
rs1021085395 Chr.6: 162054477-162054477 on Build GRCh38
rs944404536 Chr.6: 162054411-162054411 on Build GRCh38
rs773550500 Chr.6: 162054128-162054128 on Build GRCh38
rs201748755 Chr.6: 162054063-162054063 on Build GRCh38
rs1030807963 Chr.6: 162054492-162054492 on Build GRCh38
rs768688516 Chr.6: 162054205-162054205 on Build GRCh38
rs778993899 Chr.6: 162054099-162054099 on Build GRCh38
rs1010901016 Chr.6: 162054448-162054448 on Build GRCh38
rs756273262 Chr.6: 162054489-162054489 on Build GRCh38
rs149060668 Chr.6: 162054141-162054141 on Build GRCh38
rs142383136 Chr.6: 162054158-162054158 on Build GRCh38
rs370785453 Chr.6: 162054201-162054201 on Build GRCh38
rs762612415 Chr.6: 162054381-162054381 on Build GRCh38
rs766731978 Chr.6: 162054156-162054156 on Build GRCh38
rs528221295 Chr.6: 162054078-162054078 on Build GRCh38
rs752857056 Chr.6: 162054093-162054097 on Build GRCh38
rs773158383 Chr.6: 162054366-162054366 on Build GRCh38
rs922552029 Chr.6: 162054325-162054325 on Build GRCh38
rs1018598457 Chr.6: 162054131-162054131 on Build GRCh38
rs41268561 Chr.6: 162054251-162054251 on Build GRCh38
rs970996521 Chr.6: 162054187-162054187 on Build GRCh38
rs776248562 Chr.6: 162054015-162054015 on Build GRCh38
rs780523707 Chr.6: 162054074-162054074 on Build GRCh38
rs1047209996 Chr.6: 162054388-162054388 on Build GRCh38
rs113199678 Chr.6: 162054030-162054030 on Build GRCh38
rs756563827 Chr.6: 162054067-162054067 on Build GRCh38
rs1036764995 Chr.6: 162054372-162054372 on Build GRCh38
rs746588181 Chr.6: 162054071-162054071 on Build GRCh38
rs896896022 Chr.6: 162054375-162054375 on Build GRCh38

View Full Product Details