Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20140206-20140737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927668
Entrez Gene ID
101927668
Gene Name
uncharacterized LOC101927668
Gene Aliases
-
UniGene
Hs.715900
Gene Chromosome Location
Chr.7: 19918981-20140426 on Build GRCh38
Transcripts
NR_110114.1 NM_182762.3

Gene Symbol
MACC1
Entrez Gene ID
346389
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
7A5, SH3BP4L
UniGene
Hs.598388
Gene Chromosome Location
Chr.7: 20134655-20217390 on Build GRCh38
Transcripts
NR_110114.1 NM_182762.3

SNP Detail

SNP ID Chromosome Location
rs543307854 Chr.7: 20140674-20140675 on Build GRCh38
rs1039623109 Chr.7: 20140378-20140378 on Build GRCh38
rs886300067 Chr.7: 20140434-20140434 on Build GRCh38
rs1037897924 Chr.7: 20140674-20140674 on Build GRCh38
rs1038679801 Chr.7: 20140288-20140288 on Build GRCh38
rs141250478 Chr.7: 20140235-20140235 on Build GRCh38
rs942346282 Chr.7: 20140715-20140715 on Build GRCh38
rs936389664 Chr.7: 20140282-20140282 on Build GRCh38
rs6944126 Chr.7: 20140669-20140669 on Build GRCh38
rs780710411 Chr.7: 20140663-20140663 on Build GRCh38
rs537403398 Chr.7: 20140573-20140573 on Build GRCh38
rs761983236 Chr.7: 20140274-20140274 on Build GRCh38
rs182732375 Chr.7: 20140661-20140661 on Build GRCh38
rs1001490613 Chr.7: 20140479-20140479 on Build GRCh38
rs751787261 Chr.7: 20140251-20140251 on Build GRCh38
rs532775305 Chr.7: 20140464-20140464 on Build GRCh38
rs374550850 Chr.7: 20140433-20140433 on Build GRCh38
rs563133191 Chr.7: 20140347-20140347 on Build GRCh38
rs1020306250 Chr.7: 20140511-20140511 on Build GRCh38
rs963620409 Chr.7: 20140474-20140474 on Build GRCh38
rs552410274 Chr.7: 20140594-20140594 on Build GRCh38
rs909663215 Chr.7: 20140680-20140680 on Build GRCh38
rs899087576 Chr.7: 20140686-20140686 on Build GRCh38
rs767955474 Chr.7: 20140441-20140441 on Build GRCh38
rs560125991 Chr.7: 20140507-20140507 on Build GRCh38
rs112992060 Chr.7: 20140383-20140383 on Build GRCh38
rs926184219 Chr.7: 20140262-20140262 on Build GRCh38
rs557544941 Chr.7: 20140370-20140370 on Build GRCh38
rs926229085 Chr.7: 20140565-20140565 on Build GRCh38
rs756632598 Chr.7: 20140566-20140566 on Build GRCh38
rs17142482 Chr.7: 20140508-20140508 on Build GRCh38
rs375689668 Chr.7: 20140442-20140442 on Build GRCh38
rs997239087 Chr.7: 20140364-20140364 on Build GRCh38
rs191792550 Chr.7: 20140513-20140513 on Build GRCh38
rs1035246820 Chr.7: 20140512-20140512 on Build GRCh38
rs972214759 Chr.7: 20140316-20140316 on Build GRCh38
rs919408967 Chr.7: 20140607-20140607 on Build GRCh38
rs187399585 Chr.7: 20140506-20140506 on Build GRCh38
rs932956317 Chr.7: 20140610-20140610 on Build GRCh38
rs900262068 Chr.7: 20140343-20140343 on Build GRCh38
rs574374286 Chr.7: 20140234-20140234 on Build GRCh38
rs150858751 Chr.7: 20140549-20140549 on Build GRCh38
rs6964413 Chr.7: 20140632-20140632 on Build GRCh38
rs371590849 Chr.7: 20140330-20140330 on Build GRCh38
rs888106454 Chr.7: 20140358-20140358 on Build GRCh38
rs1051541861 Chr.7: 20140621-20140621 on Build GRCh38
rs1016218975 Chr.7: 20140457-20140457 on Build GRCh38
rs182288834 Chr.7: 20140417-20140417 on Build GRCh38
rs537217388 Chr.7: 20140359-20140359 on Build GRCh38
rs534541271 Chr.7: 20140648-20140648 on Build GRCh38
rs939484095 Chr.7: 20140303-20140303 on Build GRCh38
rs535375247 Chr.7: 20140676-20140676 on Build GRCh38
rs901092189 Chr.7: 20140422-20140422 on Build GRCh38
rs928242518 Chr.7: 20140264-20140264 on Build GRCh38
rs1001376055 Chr.7: 20140704-20140704 on Build GRCh38
rs764350621 Chr.7: 20140459-20140459 on Build GRCh38

View Full Product Details