Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23644386-23644923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC126
Entrez Gene ID
90693
Gene Name
coiled-coil domain containing 126
Gene Aliases
-
UniGene
Hs.232296
Gene Chromosome Location
Chr.7: 23597333-23644708 on Build GRCh38
Transcripts
NM_138771.3 XM_017012775.1 XM_017012776.1

SNP Detail

SNP ID Chromosome Location
rs144640248 Chr.7: 23644618-23644618 on Build GRCh38
rs558215134 Chr.7: 23644459-23644459 on Build GRCh38
rs138615757 Chr.7: 23644722-23644722 on Build GRCh38
rs994320944 Chr.7: 23644849-23644849 on Build GRCh38
rs953076528 Chr.7: 23644442-23644442 on Build GRCh38
rs949732596 Chr.7: 23644741-23644741 on Build GRCh38
rs1025894703 Chr.7: 23644599-23644599 on Build GRCh38
rs1040788657 Chr.7: 23644760-23644760 on Build GRCh38
rs189063298 Chr.7: 23644887-23644887 on Build GRCh38
rs979544394 Chr.7: 23644675-23644675 on Build GRCh38
rs1028549573 Chr.7: 23644436-23644436 on Build GRCh38
rs1016644495 Chr.7: 23644832-23644832 on Build GRCh38
rs184562874 Chr.7: 23644449-23644449 on Build GRCh38
rs770392599 Chr.7: 23644707-23644707 on Build GRCh38
rs984217963 Chr.7: 23644448-23644448 on Build GRCh38
rs550498190 Chr.7: 23644504-23644507 on Build GRCh38
rs961438812 Chr.7: 23644472-23644472 on Build GRCh38
rs955385334 Chr.7: 23644613-23644613 on Build GRCh38
rs948725977 Chr.7: 23644548-23644548 on Build GRCh38
rs567162943 Chr.7: 23644781-23644781 on Build GRCh38
rs942756905 Chr.7: 23644626-23644626 on Build GRCh38
rs185812282 Chr.7: 23644812-23644812 on Build GRCh38
rs972923354 Chr.7: 23644527-23644527 on Build GRCh38
rs936618758 Chr.7: 23644671-23644671 on Build GRCh38
rs543884141 Chr.7: 23644874-23644874 on Build GRCh38
rs1053020602 Chr.7: 23644704-23644704 on Build GRCh38
rs1017032195 Chr.7: 23644451-23644451 on Build GRCh38
rs898667676 Chr.7: 23644846-23644846 on Build GRCh38
rs376764800 Chr.7: 23644792-23644792 on Build GRCh38
rs552293390 Chr.7: 23644817-23644818 on Build GRCh38
rs935853721 Chr.7: 23644698-23644698 on Build GRCh38
rs559561182 Chr.7: 23644661-23644661 on Build GRCh38
rs189430640 Chr.7: 23644694-23644694 on Build GRCh38
rs992895354 Chr.7: 23644504-23644504 on Build GRCh38
rs181202835 Chr.7: 23644793-23644793 on Build GRCh38
rs578054084 Chr.7: 23644534-23644534 on Build GRCh38
rs561463554 Chr.7: 23644799-23644799 on Build GRCh38
rs980514092 Chr.7: 23644565-23644565 on Build GRCh38
rs142120220 Chr.7: 23644897-23644897 on Build GRCh38
rs575779151 Chr.7: 23644422-23644422 on Build GRCh38
rs911239877 Chr.7: 23644625-23644625 on Build GRCh38

View Full Product Details