Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27182638-27183138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA11
Entrez Gene ID
3207
Gene Name
homeobox A11
Gene Aliases
HOX1, HOX1I, RUSAT1
UniGene
Hs.249171
Gene Chromosome Location
Chr.7: 27181157-27185216 on Build GRCh38
Transcripts
NM_005523.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110125 c.898_899AA>CT p.N300L Chr.7: 27182839-27182840 on Build GRCh38
3832586 c.853G>T p.V285F Chr.7: 27182885-27182885 on Build GRCh38
370915 c.765G>A p.E255E Chr.7: 27182973-27182973 on Build GRCh38
6028062 c.731delA p.K244fs*51 Chr.7: 27183007-27183007 on Build GRCh38
7163772 c.798T>G p.I266M Chr.7: 27182940-27182940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766487741 Chr.7: 27182756-27182756 on Build GRCh38
rs1048342326 Chr.7: 27182939-27182939 on Build GRCh38
rs760114301 Chr.7: 27183061-27183061 on Build GRCh38
rs184605433 Chr.7: 27182791-27182791 on Build GRCh38
rs985474969 Chr.7: 27182865-27182865 on Build GRCh38
rs749179017 Chr.7: 27182876-27182876 on Build GRCh38
rs766983697 Chr.7: 27182901-27182901 on Build GRCh38
rs747567030 Chr.7: 27182814-27182814 on Build GRCh38
rs747431045 Chr.7: 27182985-27182985 on Build GRCh38
rs533563771 Chr.7: 27183033-27183033 on Build GRCh38
rs758467910 Chr.7: 27182833-27182834 on Build GRCh38
rs377744532 Chr.7: 27182747-27182747 on Build GRCh38
rs774243650 Chr.7: 27183044-27183044 on Build GRCh38
rs759857446 Chr.7: 27182765-27182765 on Build GRCh38
rs754825702 Chr.7: 27182766-27182767 on Build GRCh38
rs911846678 Chr.7: 27182902-27182902 on Build GRCh38
rs149018886 Chr.7: 27183026-27183026 on Build GRCh38
rs1038754200 Chr.7: 27182686-27182686 on Build GRCh38
rs557441185 Chr.7: 27182942-27182942 on Build GRCh38
rs761718543 Chr.7: 27183046-27183046 on Build GRCh38
rs749932187 Chr.7: 27183053-27183053 on Build GRCh38
rs115808703 Chr.7: 27182898-27182898 on Build GRCh38
rs368315894 Chr.7: 27182949-27182949 on Build GRCh38
rs755750728 Chr.7: 27182916-27182916 on Build GRCh38
rs763248001 Chr.7: 27183034-27183034 on Build GRCh38
rs891453900 Chr.7: 27183057-27183057 on Build GRCh38
rs760737881 Chr.7: 27182752-27182752 on Build GRCh38
rs974627662 Chr.7: 27182966-27182966 on Build GRCh38
rs765082062 Chr.7: 27182775-27182775 on Build GRCh38
rs767500718 Chr.7: 27183052-27183052 on Build GRCh38
rs770470576 Chr.7: 27182687-27182687 on Build GRCh38
rs372505902 Chr.7: 27182965-27182965 on Build GRCh38
rs763955016 Chr.7: 27182883-27182883 on Build GRCh38
rs944521903 Chr.7: 27182913-27182913 on Build GRCh38
rs371730328 Chr.7: 27182816-27182816 on Build GRCh38
rs778514371 Chr.7: 27182779-27182779 on Build GRCh38
rs747952678 Chr.7: 27182826-27182826 on Build GRCh38
rs776938360 Chr.7: 27182757-27182757 on Build GRCh38
rs775407962 Chr.7: 27182851-27182851 on Build GRCh38
rs759798705 Chr.7: 27182830-27182830 on Build GRCh38
rs1032649522 Chr.7: 27182712-27182712 on Build GRCh38
rs777829525 Chr.7: 27182843-27182843 on Build GRCh38
rs553465836 Chr.7: 27183068-27183068 on Build GRCh38
rs864321666 Chr.7: 27182866-27182866 on Build GRCh38
rs1045410394 Chr.7: 27182718-27182718 on Build GRCh38
rs566789760 Chr.7: 27182710-27182710 on Build GRCh38
rs1008718199 Chr.7: 27182673-27182673 on Build GRCh38
rs60368266 Chr.7: 27183069-27183069 on Build GRCh38
rs749987350 Chr.7: 27182910-27182910 on Build GRCh38
rs17427861 Chr.7: 27182790-27182790 on Build GRCh38
rs367981250 Chr.7: 27182802-27182802 on Build GRCh38
rs751392924 Chr.7: 27182890-27182890 on Build GRCh38
rs1055593858 Chr.7: 27183107-27183107 on Build GRCh38
rs765601556 Chr.7: 27182839-27182839 on Build GRCh38
rs754693094 Chr.7: 27183075-27183075 on Build GRCh38
rs777965862 Chr.7: 27183077-27183077 on Build GRCh38
rs201081293 Chr.7: 27182787-27182787 on Build GRCh38
rs1010447674 Chr.7: 27182781-27182781 on Build GRCh38
rs974583690 Chr.7: 27182826-27182826 on Build GRCh38
rs199875419 Chr.7: 27182997-27182997 on Build GRCh38
rs900357733 Chr.7: 27182977-27182977 on Build GRCh38
rs748520497 Chr.7: 27182798-27182798 on Build GRCh38
rs755805936 Chr.7: 27182793-27182793 on Build GRCh38
rs375071728 Chr.7: 27182823-27182823 on Build GRCh38
rs906971936 Chr.7: 27182744-27182744 on Build GRCh38
rs770522667 Chr.7: 27183050-27183050 on Build GRCh38
rs1025013154 Chr.7: 27182786-27182786 on Build GRCh38
rs758635540 Chr.7: 27182964-27182964 on Build GRCh38
rs779227542 Chr.7: 27182797-27182797 on Build GRCh38
rs377202514 Chr.7: 27183024-27183024 on Build GRCh38
rs918830277 Chr.7: 27182679-27182679 on Build GRCh38
rs751075694 Chr.7: 27182788-27182788 on Build GRCh38
rs1010357318 Chr.7: 27183066-27183066 on Build GRCh38
rs768285430 Chr.7: 27183043-27183043 on Build GRCh38
rs752576612 Chr.7: 27182782-27182782 on Build GRCh38
rs1038871223 Chr.7: 27182929-27182929 on Build GRCh38
rs746556562 Chr.7: 27183037-27183038 on Build GRCh38
rs748995496 Chr.7: 27182665-27182665 on Build GRCh38
rs536684764 Chr.7: 27182728-27182728 on Build GRCh38
rs773529344 Chr.7: 27182812-27182812 on Build GRCh38
rs762722718 Chr.7: 27182853-27182853 on Build GRCh38
rs868538518 Chr.7: 27182769-27182769 on Build GRCh38

View Full Product Details