Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27542853-27543356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIBADH
Entrez Gene ID
11112
Gene Name
3-hydroxyisobutyrate dehydrogenase
Gene Aliases
NS5ATP1
UniGene
Hs.406758
Gene Chromosome Location
Chr.7: 27525440-27663001 on Build GRCh38
Transcripts
NM_152740.3 XM_017011719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574794 c.485-7G>A p.? Chr.7: 27543107-27543107 on Build GRCh38
1088901 c.500G>A p.R167Q Chr.7: 27543085-27543085 on Build GRCh38
3995532 c.588G>C p.V196V Chr.7: 27542997-27542997 on Build GRCh38
1673079 c.527G>T p.G176V Chr.7: 27543058-27543058 on Build GRCh38
7256904 c.492A>G p.G164G Chr.7: 27543093-27543093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746083478 Chr.7: 27543005-27543005 on Build GRCh38
rs750072729 Chr.7: 27542933-27542933 on Build GRCh38
rs753518865 Chr.7: 27543064-27543064 on Build GRCh38
rs752045760 Chr.7: 27542999-27542999 on Build GRCh38
rs375377576 Chr.7: 27543140-27543140 on Build GRCh38
rs760187776 Chr.7: 27543056-27543056 on Build GRCh38
rs765964510 Chr.7: 27543062-27543062 on Build GRCh38
rs954948405 Chr.7: 27543283-27543283 on Build GRCh38
rs1003598905 Chr.7: 27542908-27542909 on Build GRCh38
rs937992328 Chr.7: 27543319-27543319 on Build GRCh38
rs200680648 Chr.7: 27543001-27543001 on Build GRCh38
rs752997445 Chr.7: 27542962-27542962 on Build GRCh38
rs186668305 Chr.7: 27543069-27543069 on Build GRCh38
rs191127256 Chr.7: 27543324-27543324 on Build GRCh38
rs756256591 Chr.7: 27542960-27542960 on Build GRCh38
rs760381893 Chr.7: 27542936-27542936 on Build GRCh38
rs748013886 Chr.7: 27542896-27542896 on Build GRCh38
rs1029524362 Chr.7: 27543252-27543252 on Build GRCh38
rs768604832 Chr.7: 27543021-27543021 on Build GRCh38
rs750473162 Chr.7: 27543076-27543076 on Build GRCh38
rs530335121 Chr.7: 27543229-27543229 on Build GRCh38
rs370742965 Chr.7: 27543081-27543081 on Build GRCh38
rs766617202 Chr.7: 27543148-27543148 on Build GRCh38
rs773571533 Chr.7: 27543092-27543092 on Build GRCh38
rs748116774 Chr.7: 27543090-27543090 on Build GRCh38
rs112747080 Chr.7: 27543304-27543304 on Build GRCh38
rs988603091 Chr.7: 27543147-27543147 on Build GRCh38
rs949939066 Chr.7: 27543178-27543178 on Build GRCh38
rs565454335 Chr.7: 27543123-27543123 on Build GRCh38
rs966912353 Chr.7: 27542983-27542983 on Build GRCh38
rs781724865 Chr.7: 27543074-27543074 on Build GRCh38
rs767864211 Chr.7: 27543130-27543130 on Build GRCh38
rs938082226 Chr.7: 27542915-27542915 on Build GRCh38
rs764514229 Chr.7: 27543118-27543118 on Build GRCh38
rs761349197 Chr.7: 27542921-27542921 on Build GRCh38
rs150435784 Chr.7: 27543060-27543060 on Build GRCh38
rs377597338 Chr.7: 27543036-27543036 on Build GRCh38
rs555463296 Chr.7: 27543107-27543107 on Build GRCh38
rs200702367 Chr.7: 27543014-27543014 on Build GRCh38
rs774317331 Chr.7: 27543027-27543027 on Build GRCh38
rs1033298999 Chr.7: 27543141-27543141 on Build GRCh38
rs373825338 Chr.7: 27543011-27543011 on Build GRCh38
rs199965096 Chr.7: 27542994-27542994 on Build GRCh38
rs978309739 Chr.7: 27543234-27543234 on Build GRCh38
rs562814983 Chr.7: 27543201-27543201 on Build GRCh38
rs770992261 Chr.7: 27542968-27542968 on Build GRCh38
rs766978826 Chr.7: 27542930-27542930 on Build GRCh38
rs970518102 Chr.7: 27542901-27542901 on Build GRCh38
rs377399312 Chr.7: 27542956-27542956 on Build GRCh38
rs762252127 Chr.7: 27543125-27543125 on Build GRCh38
rs763271452 Chr.7: 27543065-27543065 on Build GRCh38
rs761707613 Chr.7: 27543031-27543031 on Build GRCh38
rs199960454 Chr.7: 27543108-27543108 on Build GRCh38
rs780213428 Chr.7: 27543070-27543070 on Build GRCh38
rs768479109 Chr.7: 27543085-27543085 on Build GRCh38
rs758781605 Chr.7: 27542985-27542985 on Build GRCh38
rs759243257 Chr.7: 27543117-27543117 on Build GRCh38
rs749564467 Chr.7: 27543082-27543082 on Build GRCh38
rs777003772 Chr.7: 27543027-27543028 on Build GRCh38
rs561279065 Chr.7: 27543098-27543098 on Build GRCh38
rs772525885 Chr.7: 27543055-27543055 on Build GRCh38
rs943597664 Chr.7: 27543029-27543029 on Build GRCh38
rs575630511 Chr.7: 27543013-27543013 on Build GRCh38
rs774179352 Chr.7: 27542920-27542920 on Build GRCh38
rs764414539 Chr.7: 27543066-27543066 on Build GRCh38
rs113063115 Chr.7: 27542918-27542918 on Build GRCh38
rs372529743 Chr.7: 27543112-27543112 on Build GRCh38
rs532631126 Chr.7: 27543132-27543132 on Build GRCh38
rs978447 Chr.7: 27543164-27543164 on Build GRCh38
rs770465936 Chr.7: 27543110-27543110 on Build GRCh38
rs980003031 Chr.7: 27543198-27543198 on Build GRCh38
rs756286958 Chr.7: 27543078-27543078 on Build GRCh38
rs571243282 Chr.7: 27542878-27542880 on Build GRCh38

View Full Product Details