Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31517902-31518438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC129
Entrez Gene ID
223075
Gene Name
coiled-coil domain containing 129
Gene Aliases
-
UniGene
Hs.224269
Gene Chromosome Location
Chr.7: 31514071-31658720 on Build GRCh38
Transcripts
NM_001257967.1 NR_047565.1 XM_011515213.1 XM_017011871.1 XM_017011872.1

SNP Detail

SNP ID Chromosome Location
rs923550191 Chr.7: 31518221-31518221 on Build GRCh38
rs73305474 Chr.7: 31518061-31518061 on Build GRCh38
rs891977290 Chr.7: 31518294-31518294 on Build GRCh38
rs994835044 Chr.7: 31518123-31518123 on Build GRCh38
rs1011711604 Chr.7: 31518315-31518315 on Build GRCh38
rs909267856 Chr.7: 31518173-31518173 on Build GRCh38
rs556652176 Chr.7: 31518261-31518261 on Build GRCh38
rs1053196863 Chr.7: 31518008-31518008 on Build GRCh38
rs922177274 Chr.7: 31518011-31518011 on Build GRCh38
rs975032054 Chr.7: 31518280-31518280 on Build GRCh38
rs953731826 Chr.7: 31518312-31518312 on Build GRCh38
rs891573279 Chr.7: 31518012-31518012 on Build GRCh38
rs748446158 Chr.7: 31517955-31517955 on Build GRCh38
rs542102315 Chr.7: 31517951-31517951 on Build GRCh38
rs972588258 Chr.7: 31518135-31518135 on Build GRCh38
rs566088143 Chr.7: 31518157-31518157 on Build GRCh38
rs182067798 Chr.7: 31518201-31518201 on Build GRCh38
rs527876010 Chr.7: 31517966-31517966 on Build GRCh38
rs987785453 Chr.7: 31517945-31517945 on Build GRCh38
rs1021351201 Chr.7: 31518033-31518033 on Build GRCh38
rs188188823 Chr.7: 31518059-31518059 on Build GRCh38
rs959664444 Chr.7: 31518379-31518379 on Build GRCh38
rs764973097 Chr.7: 31518340-31518340 on Build GRCh38
rs1011755886 Chr.7: 31518029-31518029 on Build GRCh38
rs535502417 Chr.7: 31518376-31518376 on Build GRCh38
rs922239779 Chr.7: 31518302-31518302 on Build GRCh38
rs1023579839 Chr.7: 31518124-31518124 on Build GRCh38
rs1043231207 Chr.7: 31518320-31518320 on Build GRCh38
rs570878715 Chr.7: 31518212-31518212 on Build GRCh38
rs969287823 Chr.7: 31517926-31517926 on Build GRCh38
rs1039267329 Chr.7: 31517969-31517969 on Build GRCh38
rs12701106 Chr.7: 31518038-31518038 on Build GRCh38
rs919365447 Chr.7: 31517929-31517929 on Build GRCh38
rs1034805489 Chr.7: 31517943-31517943 on Build GRCh38
rs912010895 Chr.7: 31518332-31518332 on Build GRCh38
rs186281416 Chr.7: 31518349-31518349 on Build GRCh38
rs180736658 Chr.7: 31518077-31518077 on Build GRCh38
rs537977015 Chr.7: 31518234-31518234 on Build GRCh38
rs78623479 Chr.7: 31518183-31518183 on Build GRCh38
rs190773785 Chr.7: 31518152-31518152 on Build GRCh38
rs1053271661 Chr.7: 31518290-31518290 on Build GRCh38
rs150418649 Chr.7: 31517968-31517968 on Build GRCh38
rs967727723 Chr.7: 31518249-31518249 on Build GRCh38
rs902093856 Chr.7: 31517993-31517993 on Build GRCh38
rs1049118443 Chr.7: 31517958-31517958 on Build GRCh38
rs932194588 Chr.7: 31518022-31518022 on Build GRCh38
rs1035340891 Chr.7: 31518370-31518370 on Build GRCh38
rs933562593 Chr.7: 31518226-31518226 on Build GRCh38
rs187303262 Chr.7: 31518138-31518138 on Build GRCh38
rs145196814 Chr.7: 31518000-31518000 on Build GRCh38
rs929371025 Chr.7: 31517938-31517938 on Build GRCh38
rs545653561 Chr.7: 31518397-31518397 on Build GRCh38
rs12701107 Chr.7: 31518111-31518111 on Build GRCh38
rs905608804 Chr.7: 31518356-31518356 on Build GRCh38
rs956629572 Chr.7: 31517944-31517944 on Build GRCh38
rs560439405 Chr.7: 31517952-31517952 on Build GRCh38
rs556926947 Chr.7: 31518149-31518149 on Build GRCh38
rs1001223595 Chr.7: 31518361-31518361 on Build GRCh38
rs909226087 Chr.7: 31517962-31517962 on Build GRCh38
rs930659040 Chr.7: 31518413-31518413 on Build GRCh38
rs997677807 Chr.7: 31518002-31518002 on Build GRCh38

View Full Product Details