Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71420826-71421322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WBSCR17
Entrez Gene ID
64409
Gene Name
Williams-Beuren syndrome chromosome region 17
Gene Aliases
GALNACT17, GALNT16, GALNT20, GALNTL3, GalNAc-T5L
UniGene
Hs.488591
Gene Chromosome Location
Chr.7: 71132414-71713601 on Build GRCh38
Transcripts
NM_022479.2 XM_011516467.2 XM_011516469.2 XM_017012521.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5541677 c.778C>T p.L260L Chr.7: 71420921-71420921 on Build GRCh38
4856839 c.793G>A p.E265K Chr.7: 71420936-71420936 on Build GRCh38
3950734 c.790C>A p.Q264K Chr.7: 71420933-71420933 on Build GRCh38
4572664 c.824T>C p.I275T Chr.7: 71420967-71420967 on Build GRCh38
6821386 c.779T>C p.L260P Chr.7: 71420922-71420922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761079507 Chr.7: 71421003-71421003 on Build GRCh38
rs376285148 Chr.7: 71421109-71421109 on Build GRCh38
rs144050734 Chr.7: 71421097-71421097 on Build GRCh38
rs73363757 Chr.7: 71420877-71420877 on Build GRCh38
rs749428973 Chr.7: 71420966-71420966 on Build GRCh38
rs765278202 Chr.7: 71421045-71421045 on Build GRCh38
rs751565301 Chr.7: 71420943-71420943 on Build GRCh38
rs1056597341 Chr.7: 71420923-71420923 on Build GRCh38
rs752472906 Chr.7: 71421080-71421080 on Build GRCh38
rs372792355 Chr.7: 71421095-71421095 on Build GRCh38
rs112759984 Chr.7: 71421145-71421145 on Build GRCh38
rs777772965 Chr.7: 71420893-71420893 on Build GRCh38
rs771465510 Chr.7: 71420899-71420899 on Build GRCh38
rs769088187 Chr.7: 71421116-71421116 on Build GRCh38
rs866584899 Chr.7: 71421030-71421030 on Build GRCh38
rs754896515 Chr.7: 71420947-71420947 on Build GRCh38
rs866308070 Chr.7: 71421008-71421008 on Build GRCh38
rs866747641 Chr.7: 71420926-71420926 on Build GRCh38
rs780119747 Chr.7: 71420865-71420865 on Build GRCh38
rs397936866 Chr.7: 71420898-71420899 on Build GRCh38
rs576659383 Chr.7: 71421233-71421233 on Build GRCh38
rs765990737 Chr.7: 71421055-71421055 on Build GRCh38
rs1007058782 Chr.7: 71421234-71421234 on Build GRCh38
rs1030487275 Chr.7: 71421274-71421274 on Build GRCh38
rs199903990 Chr.7: 71421128-71421128 on Build GRCh38
rs948149446 Chr.7: 71421164-71421164 on Build GRCh38
rs765425954 Chr.7: 71421151-71421151 on Build GRCh38
rs532538143 Chr.7: 71421252-71421252 on Build GRCh38
rs775779137 Chr.7: 71421002-71421002 on Build GRCh38
rs200703759 Chr.7: 71421114-71421114 on Build GRCh38
rs142659982 Chr.7: 71421165-71421165 on Build GRCh38
rs11455717 Chr.7: 71420897-71420898 on Build GRCh38
rs759603442 Chr.7: 71421064-71421064 on Build GRCh38
rs973013551 Chr.7: 71421118-71421118 on Build GRCh38
rs376148000 Chr.7: 71421016-71421016 on Build GRCh38
rs772443852 Chr.7: 71420905-71420905 on Build GRCh38
rs759983467 Chr.7: 71420900-71420900 on Build GRCh38
rs749951505 Chr.7: 71420940-71420940 on Build GRCh38
rs553454349 Chr.7: 71421129-71421129 on Build GRCh38
rs78727355 Chr.7: 71420891-71420891 on Build GRCh38
rs772948920 Chr.7: 71421022-71421022 on Build GRCh38
rs771017589 Chr.7: 71420970-71420970 on Build GRCh38
rs972981881 Chr.7: 71421132-71421132 on Build GRCh38
rs138385323 Chr.7: 71420917-71420917 on Build GRCh38
rs191215830 Chr.7: 71420866-71420866 on Build GRCh38
rs904359007 Chr.7: 71421174-71421174 on Build GRCh38
rs764931976 Chr.7: 71420927-71420927 on Build GRCh38
rs891699101 Chr.7: 71421214-71421214 on Build GRCh38
rs142670632 Chr.7: 71420921-71420921 on Build GRCh38
rs868863031 Chr.7: 71421093-71421093 on Build GRCh38
rs869129433 Chr.7: 71421125-71421126 on Build GRCh38
rs776804275 Chr.7: 71421126-71421126 on Build GRCh38
rs756483138 Chr.7: 71421218-71421218 on Build GRCh38
rs759085286 Chr.7: 71421134-71421134 on Build GRCh38
rs778903809 Chr.7: 71420973-71420973 on Build GRCh38
rs758504621 Chr.7: 71421157-71421157 on Build GRCh38
rs200403024 Chr.7: 71420858-71420858 on Build GRCh38
rs555884344 Chr.7: 71421083-71421083 on Build GRCh38
rs200177349 Chr.7: 71421125-71421125 on Build GRCh38
rs760467373 Chr.7: 71421141-71421141 on Build GRCh38
rs1050734113 Chr.7: 71421221-71421221 on Build GRCh38
rs770049757 Chr.7: 71420892-71420892 on Build GRCh38
rs781199608 Chr.7: 71420948-71420948 on Build GRCh38
rs184060184 Chr.7: 71420949-71420949 on Build GRCh38
rs368686888 Chr.7: 71421136-71421136 on Build GRCh38
rs763951131 Chr.7: 71421143-71421143 on Build GRCh38
rs960967075 Chr.7: 71421284-71421284 on Build GRCh38
rs749363045 Chr.7: 71420897-71420897 on Build GRCh38
rs754100590 Chr.7: 71421089-71421089 on Build GRCh38
rs756339180 Chr.7: 71420959-71420959 on Build GRCh38
rs745937045 Chr.7: 71421112-71421112 on Build GRCh38
rs758955349 Chr.7: 71421113-71421113 on Build GRCh38
rs762495552 Chr.7: 71420942-71420942 on Build GRCh38
rs974898586 Chr.7: 71420928-71420928 on Build GRCh38
rs868631473 Chr.7: 71420957-71420957 on Build GRCh38
rs935848337 Chr.7: 71421176-71421176 on Build GRCh38
rs547880827 Chr.7: 71421263-71421263 on Build GRCh38
rs1025926158 Chr.7: 71421260-71421260 on Build GRCh38
rs61737661 Chr.7: 71421007-71421007 on Build GRCh38
rs766894940 Chr.7: 71421135-71421135 on Build GRCh38
rs750545617 Chr.7: 71421153-71421153 on Build GRCh38
rs202102121 Chr.7: 71420988-71420988 on Build GRCh38
rs778022057 Chr.7: 71420964-71420964 on Build GRCh38
rs397790672 Chr.7: 71421124-71421125 on Build GRCh38
rs199628058 Chr.7: 71421031-71421031 on Build GRCh38
rs746382123 Chr.7: 71420995-71420995 on Build GRCh38
rs755966198 Chr.7: 71421082-71421082 on Build GRCh38
rs201636538 Chr.7: 71421079-71421079 on Build GRCh38
rs910157454 Chr.7: 71420886-71420886 on Build GRCh38
rs985200950 Chr.7: 71421122-71421122 on Build GRCh38
rs992384558 Chr.7: 71421288-71421288 on Build GRCh38
rs368529766 Chr.7: 71420914-71420914 on Build GRCh38
rs1006191529 Chr.7: 71421201-71421201 on Build GRCh38
rs139549051 Chr.7: 71421062-71421062 on Build GRCh38
rs772568078 Chr.7: 71421000-71421000 on Build GRCh38
rs11981404 Chr.7: 71421236-71421236 on Build GRCh38
rs368231359 Chr.7: 71420860-71420860 on Build GRCh38
rs565089948 Chr.7: 71421237-71421237 on Build GRCh38
rs74524505 Chr.7: 71421200-71421200 on Build GRCh38

View Full Product Details