Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71787418-71787959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
UniGene
Hs.592750
Gene Chromosome Location
Chr.7: 71779491-72504271 on Build GRCh38
Transcripts
NM_001017440.2 NM_031468.3 XM_011516594.2 XM_011516595.1 XM_011516596.2 XM_011516597.2 XM_011516599.1 XM_017012675.1 XM_017012676.1 XM_017012677.1 XM_017012678.1 XM_017012679.1 XM_017012680.1 XM_017012681.1 XM_017012682.1 XM_017012683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5594349 c.655G>A p.E219K Chr.7: 71787780-71787780 on Build GRCh38
1150739 c.747G>T p.L249L Chr.7: 71787814-71787814 on Build GRCh38
3833126 c.727T>C p.F243L Chr.7: 71787834-71787834 on Build GRCh38
6619127 c.568C>T p.R190W Chr.7: 71787867-71787867 on Build GRCh38
3882178 c.711C>T p.C237C Chr.7: 71787850-71787850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768444471 Chr.7: 71787913-71787913 on Build GRCh38
rs200056778 Chr.7: 71787724-71787724 on Build GRCh38
rs757130825 Chr.7: 71787771-71787771 on Build GRCh38
rs369065730 Chr.7: 71787664-71787664 on Build GRCh38
rs555912494 Chr.7: 71787648-71787648 on Build GRCh38
rs140495509 Chr.7: 71787841-71787841 on Build GRCh38
rs756678308 Chr.7: 71787472-71787472 on Build GRCh38
rs922556161 Chr.7: 71787807-71787807 on Build GRCh38
rs765184032 Chr.7: 71787922-71787922 on Build GRCh38
rs370771019 Chr.7: 71787791-71787791 on Build GRCh38
rs773397212 Chr.7: 71787719-71787719 on Build GRCh38
rs763977683 Chr.7: 71787738-71787738 on Build GRCh38
rs894090997 Chr.7: 71787570-71787570 on Build GRCh38
rs574086430 Chr.7: 71787652-71787652 on Build GRCh38
rs755350805 Chr.7: 71787934-71787934 on Build GRCh38
rs956959501 Chr.7: 71787808-71787808 on Build GRCh38
rs575385473 Chr.7: 71787665-71787665 on Build GRCh38
rs779689666 Chr.7: 71787556-71787556 on Build GRCh38
rs921217848 Chr.7: 71787461-71787461 on Build GRCh38
rs935484694 Chr.7: 71787509-71787509 on Build GRCh38
rs114333930 Chr.7: 71787680-71787680 on Build GRCh38
rs539805217 Chr.7: 71787683-71787683 on Build GRCh38
rs377124333 Chr.7: 71787549-71787549 on Build GRCh38
rs775710059 Chr.7: 71787732-71787732 on Build GRCh38
rs753817354 Chr.7: 71787885-71787885 on Build GRCh38
rs772021684 Chr.7: 71787699-71787699 on Build GRCh38
rs904830305 Chr.7: 71787591-71787591 on Build GRCh38
rs144352678 Chr.7: 71787870-71787870 on Build GRCh38
rs370660878 Chr.7: 71787931-71787931 on Build GRCh38
rs755358796 Chr.7: 71787651-71787651 on Build GRCh38
rs796793798 Chr.7: 71787718-71787718 on Build GRCh38
rs911238188 Chr.7: 71787661-71787661 on Build GRCh38
rs759232948 Chr.7: 71787856-71787856 on Build GRCh38
rs996166169 Chr.7: 71787660-71787660 on Build GRCh38
rs780355828 Chr.7: 71787935-71787935 on Build GRCh38
rs937731364 Chr.7: 71787701-71787701 on Build GRCh38
rs746157759 Chr.7: 71787781-71787781 on Build GRCh38
rs1044863557 Chr.7: 71787875-71787878 on Build GRCh38
rs189711219 Chr.7: 71787529-71787529 on Build GRCh38
rs926360336 Chr.7: 71787899-71787899 on Build GRCh38
rs927666149 Chr.7: 71787535-71787535 on Build GRCh38
rs865962050 Chr.7: 71787896-71787896 on Build GRCh38
rs773565061 Chr.7: 71787850-71787850 on Build GRCh38
rs774961183 Chr.7: 71787867-71787867 on Build GRCh38
rs747080980 Chr.7: 71787814-71787814 on Build GRCh38
rs566877402 Chr.7: 71787530-71787530 on Build GRCh38
rs777220852 Chr.7: 71787840-71787840 on Build GRCh38
rs747229209 Chr.7: 71787720-71787720 on Build GRCh38
rs952652920 Chr.7: 71787674-71787674 on Build GRCh38
rs1003157654 Chr.7: 71787722-71787722 on Build GRCh38
rs772280579 Chr.7: 71787731-71787731 on Build GRCh38
rs1011034904 Chr.7: 71787531-71787531 on Build GRCh38
rs111497562 Chr.7: 71787736-71787736 on Build GRCh38
rs767107071 Chr.7: 71787504-71787504 on Build GRCh38
rs747120402 Chr.7: 71787672-71787672 on Build GRCh38
rs574140528 Chr.7: 71787685-71787685 on Build GRCh38
rs746028273 Chr.7: 71787728-71787728 on Build GRCh38
rs376575825 Chr.7: 71787760-71787760 on Build GRCh38
rs200374847 Chr.7: 71787769-71787769 on Build GRCh38
rs768579672 Chr.7: 71787835-71787835 on Build GRCh38
rs546913908 Chr.7: 71787657-71787657 on Build GRCh38
rs1043008881 Chr.7: 71787618-71787618 on Build GRCh38
rs375705532 Chr.7: 71787765-71787765 on Build GRCh38
rs765540825 Chr.7: 71787757-71787757 on Build GRCh38
rs1005254909 Chr.7: 71787829-71787829 on Build GRCh38
rs200207748 Chr.7: 71787766-71787766 on Build GRCh38
rs201522077 Chr.7: 71787739-71787739 on Build GRCh38
rs529005167 Chr.7: 71787714-71787714 on Build GRCh38
rs1054789739 Chr.7: 71787566-71787566 on Build GRCh38
rs777793983 Chr.7: 71787768-71787768 on Build GRCh38
rs377414004 Chr.7: 71787904-71787904 on Build GRCh38
rs543237863 Chr.7: 71787690-71787690 on Build GRCh38
rs545701116 Chr.7: 71787675-71787675 on Build GRCh38
rs569432917 Chr.7: 71787462-71787462 on Build GRCh38
rs563992207 Chr.7: 71787676-71787676 on Build GRCh38
rs532063545 Chr.7: 71787737-71787737 on Build GRCh38
rs983946451 Chr.7: 71787466-71787466 on Build GRCh38
rs80291804 Chr.7: 71787689-71787689 on Build GRCh38
rs767122514 Chr.7: 71787866-71787866 on Build GRCh38
rs901365933 Chr.7: 71787505-71787505 on Build GRCh38
rs748599942 Chr.7: 71787846-71787846 on Build GRCh38
rs763453907 Chr.7: 71787880-71787880 on Build GRCh38
rs770151080 Chr.7: 71787849-71787849 on Build GRCh38
rs747370593 Chr.7: 71787463-71787468 on Build GRCh38
rs539787937 Chr.7: 71787508-71787508 on Build GRCh38
rs749597576 Chr.7: 71787596-71787596 on Build GRCh38
rs952828961 Chr.7: 71787658-71787658 on Build GRCh38
rs113126722 Chr.7: 71787723-71787723 on Build GRCh38
rs902692670 Chr.7: 71787626-71787626 on Build GRCh38
rs772412135 Chr.7: 71787787-71787787 on Build GRCh38
rs1056062398 Chr.7: 71787918-71787918 on Build GRCh38

View Full Product Details