Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73328627-73329147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP6
Entrez Gene ID
8468
Gene Name
FK506 binding protein 6
Gene Aliases
FKBP36
UniGene
Hs.661266
Gene Chromosome Location
Chr.7: 73328152-73358637 on Build GRCh38
Transcripts
NM_001135211.2 NM_001281304.1 NM_003602.4 XM_006716153.2 XM_017012741.1 XM_017012742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5894426 c.136G>A p.G46R Chr.7: 73328653-73328653 on Build GRCh38
6449389 c.157C>G p.P53A Chr.7: 73328674-73328674 on Build GRCh38
5531289 c.152C>T p.S51L Chr.7: 73328684-73328684 on Build GRCh38
6449387 c.157C>G p.P53A Chr.7: 73328674-73328674 on Build GRCh38
5894428 c.136G>A p.G46R Chr.7: 73328653-73328653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782539241 Chr.7: 73328684-73328684 on Build GRCh38
rs781991123 Chr.7: 73328733-73328733 on Build GRCh38
rs782101452 Chr.7: 73328681-73328681 on Build GRCh38
rs372557515 Chr.7: 73328700-73328700 on Build GRCh38
rs543376453 Chr.7: 73328961-73328961 on Build GRCh38
rs782029346 Chr.7: 73328665-73328665 on Build GRCh38
rs781198883 Chr.7: 73329045-73329045 on Build GRCh38
rs375149793 Chr.7: 73328706-73328706 on Build GRCh38
rs201938784 Chr.7: 73328742-73328742 on Build GRCh38
rs781930877 Chr.7: 73329101-73329101 on Build GRCh38
rs541642662 Chr.7: 73328774-73328774 on Build GRCh38
rs782470191 Chr.7: 73328697-73328697 on Build GRCh38
rs199994623 Chr.7: 73328715-73328715 on Build GRCh38
rs782306134 Chr.7: 73328710-73328710 on Build GRCh38
rs376231085 Chr.7: 73328756-73328756 on Build GRCh38
rs189818801 Chr.7: 73328951-73328951 on Build GRCh38
rs560616857 Chr.7: 73329124-73329124 on Build GRCh38
rs369763024 Chr.7: 73328737-73328737 on Build GRCh38
rs782366580 Chr.7: 73328732-73328732 on Build GRCh38
rs781796844 Chr.7: 73328690-73328690 on Build GRCh38
rs184753552 Chr.7: 73328929-73328929 on Build GRCh38
rs530329798 Chr.7: 73328899-73328899 on Build GRCh38
rs549775269 Chr.7: 73328782-73328782 on Build GRCh38
rs570929250 Chr.7: 73328831-73328831 on Build GRCh38
rs564032345 Chr.7: 73329046-73329046 on Build GRCh38
rs782129593 Chr.7: 73328735-73328735 on Build GRCh38
rs576316627 Chr.7: 73328953-73328953 on Build GRCh38
rs766599013 Chr.7: 73328747-73328748 on Build GRCh38
rs782399413 Chr.7: 73328648-73328648 on Build GRCh38
rs561421587 Chr.7: 73328896-73328896 on Build GRCh38
rs117566868 Chr.7: 73328967-73328967 on Build GRCh38
rs782283718 Chr.7: 73328707-73328707 on Build GRCh38
rs578077191 Chr.7: 73329044-73329044 on Build GRCh38
rs782036857 Chr.7: 73328739-73328739 on Build GRCh38
rs781960689 Chr.7: 73328673-73328673 on Build GRCh38
rs755078916 Chr.7: 73328931-73328931 on Build GRCh38
rs781869532 Chr.7: 73328682-73328682 on Build GRCh38
rs782218431 Chr.7: 73328716-73328716 on Build GRCh38
rs115819412 Chr.7: 73328835-73328835 on Build GRCh38
rs782718896 Chr.7: 73328672-73328672 on Build GRCh38
rs181980616 Chr.7: 73328921-73328921 on Build GRCh38
rs545472664 Chr.7: 73329092-73329092 on Build GRCh38
rs782815391 Chr.7: 73328687-73328687 on Build GRCh38

View Full Product Details