Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100181999-100182501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAG3
Entrez Gene ID
10734
Gene Name
stromal antigen 3
Gene Aliases
-
UniGene
Hs.592283
Gene Chromosome Location
Chr.7: 100177563-100219334 on Build GRCh38
Transcripts
NM_001282716.1 NM_001282717.1 NM_001282718.1 NM_012447.3 XM_011515742.1 XM_017011683.1 XM_017011684.1 XM_017011685.1 XM_017011686.1 XM_017011687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227731 c.218C>T p.P73L Chr.7: 100182191-100182191 on Build GRCh38
748236 c.219G>A p.P73P Chr.7: 100182192-100182192 on Build GRCh38
6583530 c.154T>A p.F52I Chr.7: 100182127-100182127 on Build GRCh38
118172 c.133T>C p.L45L Chr.7: 100182106-100182106 on Build GRCh38
5765360 c.219+5G>A p.? Chr.7: 100182197-100182197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372543441 Chr.7: 100182162-100182162 on Build GRCh38
rs557088096 Chr.7: 100182426-100182426 on Build GRCh38
rs761565485 Chr.7: 100182039-100182039 on Build GRCh38
rs925061660 Chr.7: 100182472-100182476 on Build GRCh38
rs368831215 Chr.7: 100182124-100182124 on Build GRCh38
rs543606133 Chr.7: 100182396-100182396 on Build GRCh38
rs1013819547 Chr.7: 100182284-100182284 on Build GRCh38
rs767628834 Chr.7: 100182179-100182179 on Build GRCh38
rs985424358 Chr.7: 100182074-100182074 on Build GRCh38
rs551667300 Chr.7: 100182256-100182256 on Build GRCh38
rs146224381 Chr.7: 100182404-100182404 on Build GRCh38
rs756236220 Chr.7: 100182187-100182187 on Build GRCh38
rs200232685 Chr.7: 100182145-100182145 on Build GRCh38
rs1006855053 Chr.7: 100182378-100182378 on Build GRCh38
rs778737784 Chr.7: 100182121-100182121 on Build GRCh38
rs187538572 Chr.7: 100182275-100182275 on Build GRCh38
rs369173165 Chr.7: 100182230-100182230 on Build GRCh38
rs752704145 Chr.7: 100182062-100182062 on Build GRCh38
rs200800005 Chr.7: 100182149-100182149 on Build GRCh38
rs757697214 Chr.7: 100182209-100182209 on Build GRCh38
rs370022253 Chr.7: 100182191-100182191 on Build GRCh38
rs6465764 Chr.7: 100182263-100182263 on Build GRCh38
rs975891897 Chr.7: 100182146-100182148 on Build GRCh38
rs778392586 Chr.7: 100182068-100182068 on Build GRCh38
rs966608212 Chr.7: 100182456-100182457 on Build GRCh38
rs759858941 Chr.7: 100182178-100182178 on Build GRCh38
rs192812211 Chr.7: 100182307-100182307 on Build GRCh38
rs978319539 Chr.7: 100182461-100182461 on Build GRCh38
rs182967242 Chr.7: 100182416-100182416 on Build GRCh38
rs534244395 Chr.7: 100182272-100182272 on Build GRCh38
rs764978239 Chr.7: 100182172-100182172 on Build GRCh38
rs754354994 Chr.7: 100182192-100182192 on Build GRCh38
rs749716573 Chr.7: 100182088-100182088 on Build GRCh38
rs879843533 Chr.7: 100182352-100182352 on Build GRCh38
rs755046326 Chr.7: 100182057-100182057 on Build GRCh38
rs751758656 Chr.7: 100182054-100182054 on Build GRCh38
rs746995023 Chr.7: 100182083-100182083 on Build GRCh38
rs377148930 Chr.7: 100182065-100182065 on Build GRCh38
rs76153486 Chr.7: 100182358-100182358 on Build GRCh38
rs533315054 Chr.7: 100182248-100182248 on Build GRCh38
rs766637930 Chr.7: 100182052-100182052 on Build GRCh38
rs775358985 Chr.7: 100182173-100182173 on Build GRCh38
rs374987093 Chr.7: 100182047-100182047 on Build GRCh38
rs746500893 Chr.7: 100182148-100182148 on Build GRCh38
rs548211372 Chr.7: 100182358-100182359 on Build GRCh38
rs761426606 Chr.7: 100182276-100182276 on Build GRCh38
rs770580653 Chr.7: 100182099-100182100 on Build GRCh38
rs752861781 Chr.7: 100182180-100182180 on Build GRCh38
rs936364766 Chr.7: 100182474-100182476 on Build GRCh38
rs145456455 Chr.7: 100182150-100182150 on Build GRCh38
rs916223085 Chr.7: 100182477-100182481 on Build GRCh38
rs376805911 Chr.7: 100182210-100182210 on Build GRCh38
rs1046616204 Chr.7: 100182316-100182316 on Build GRCh38
rs1027111692 Chr.7: 100182356-100182356 on Build GRCh38
rs781274050 Chr.7: 100182060-100182060 on Build GRCh38
rs779282242 Chr.7: 100182216-100182216 on Build GRCh38
rs758141419 Chr.7: 100182241-100182241 on Build GRCh38
rs1054812295 Chr.7: 100182474-100182474 on Build GRCh38
rs761620488 Chr.7: 100182171-100182171 on Build GRCh38
rs562791783 Chr.7: 100182393-100182393 on Build GRCh38
rs775128180 Chr.7: 100182146-100182146 on Build GRCh38
rs952900319 Chr.7: 100182359-100182359 on Build GRCh38
rs776527887 Chr.7: 100182155-100182155 on Build GRCh38
rs551184556 Chr.7: 100182024-100182024 on Build GRCh38
rs1018192173 Chr.7: 100182387-100182387 on Build GRCh38
rs757643690 Chr.7: 100182096-100182096 on Build GRCh38
rs573167362 Chr.7: 100182428-100182428 on Build GRCh38
rs141509086 Chr.7: 100182345-100182345 on Build GRCh38

View Full Product Details