Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100889343-100889835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFSP1
Entrez Gene ID
402682
Gene Name
UFM1 specific peptidase 1 (inactive)
Gene Aliases
UFSP
UniGene
Hs.534845
Gene Chromosome Location
Chr.7: 100888723-100889718 on Build GRCh38
Transcripts
NM_001015072.3

SNP Detail

SNP ID Chromosome Location
rs147414076 Chr.7: 100889792-100889792 on Build GRCh38
rs1005230944 Chr.7: 100889715-100889715 on Build GRCh38
rs1045467331 Chr.7: 100889640-100889640 on Build GRCh38
rs2733918 Chr.7: 100889514-100889514 on Build GRCh38
rs796904396 Chr.7: 100889737-100889761 on Build GRCh38
rs2251631 Chr.7: 100889741-100889741 on Build GRCh38
rs1013920404 Chr.7: 100889458-100889458 on Build GRCh38
rs954583973 Chr.7: 100889756-100889756 on Build GRCh38
rs1005924961 Chr.7: 100889736-100889736 on Build GRCh38
rs530368519 Chr.7: 100889407-100889407 on Build GRCh38
rs985700213 Chr.7: 100889707-100889707 on Build GRCh38
rs1053915064 Chr.7: 100889441-100889441 on Build GRCh38
rs923494745 Chr.7: 100889560-100889560 on Build GRCh38
rs945455516 Chr.7: 100889781-100889781 on Build GRCh38
rs879791787 Chr.7: 100889751-100889751 on Build GRCh38
rs943341978 Chr.7: 100889539-100889539 on Build GRCh38
rs961290411 Chr.7: 100889581-100889581 on Build GRCh38
rs936725120 Chr.7: 100889665-100889665 on Build GRCh38
rs867713267 Chr.7: 100889778-100889778 on Build GRCh38
rs537404942 Chr.7: 100889502-100889502 on Build GRCh38
rs1028106279 Chr.7: 100889509-100889509 on Build GRCh38
rs952534498 Chr.7: 100889556-100889556 on Build GRCh38
rs945647838 Chr.7: 100889565-100889565 on Build GRCh38
rs926728330 Chr.7: 100889649-100889649 on Build GRCh38
rs903588048 Chr.7: 100889406-100889414 on Build GRCh38
rs989962830 Chr.7: 100889766-100889766 on Build GRCh38
rs2733920 Chr.7: 100889526-100889526 on Build GRCh38
rs570580442 Chr.7: 100889646-100889646 on Build GRCh38
rs570086495 Chr.7: 100889501-100889501 on Build GRCh38
rs994112774 Chr.7: 100889685-100889685 on Build GRCh38
rs537227711 Chr.7: 100889586-100889586 on Build GRCh38
rs17880700 Chr.7: 100889696-100889696 on Build GRCh38
rs913952056 Chr.7: 100889771-100889771 on Build GRCh38
rs984926141 Chr.7: 100889517-100889517 on Build GRCh38
rs980812950 Chr.7: 100889799-100889799 on Build GRCh38
rs543235565 Chr.7: 100889777-100889777 on Build GRCh38
rs60031054 Chr.7: 100889741-100889765 on Build GRCh38
rs4996164 Chr.7: 100889753-100889753 on Build GRCh38
rs1041357997 Chr.7: 100889691-100889691 on Build GRCh38
rs1017651953 Chr.7: 100889708-100889708 on Build GRCh38
rs949455206 Chr.7: 100889611-100889611 on Build GRCh38
rs888221319 Chr.7: 100889714-100889714 on Build GRCh38
rs576212907 Chr.7: 100889773-100889773 on Build GRCh38
rs554563565 Chr.7: 100889768-100889768 on Build GRCh38
rs1026810762 Chr.7: 100889688-100889688 on Build GRCh38
rs1053969073 Chr.7: 100889670-100889670 on Build GRCh38
rs62482512 Chr.7: 100889758-100889758 on Build GRCh38
rs992734734 Chr.7: 100889583-100889583 on Build GRCh38
rs1042544900 Chr.7: 100889399-100889399 on Build GRCh38
rs908360582 Chr.7: 100889574-100889574 on Build GRCh38
rs976494873 Chr.7: 100889551-100889551 on Build GRCh38
rs917148423 Chr.7: 100889592-100889592 on Build GRCh38
rs996532795 Chr.7: 100889700-100889700 on Build GRCh38
rs1015285564 Chr.7: 100889725-100889725 on Build GRCh38
rs902093342 Chr.7: 100889363-100889363 on Build GRCh38
rs112427842 Chr.7: 100889674-100889674 on Build GRCh38
rs936419404 Chr.7: 100889589-100889589 on Build GRCh38
rs867932174 Chr.7: 100889379-100889379 on Build GRCh38
rs2733919 Chr.7: 100889519-100889519 on Build GRCh38
rs17885823 Chr.7: 100889680-100889680 on Build GRCh38
rs869096849 Chr.7: 100889778-100889787 on Build GRCh38
rs377312174 Chr.7: 100889746-100889746 on Build GRCh38
rs910540673 Chr.7: 100889535-100889535 on Build GRCh38
rs926586151 Chr.7: 100889803-100889803 on Build GRCh38
rs892739231 Chr.7: 100889478-100889478 on Build GRCh38
rs532178082 Chr.7: 100889365-100889365 on Build GRCh38
rs570279154 Chr.7: 100889568-100889568 on Build GRCh38
rs560107719 Chr.7: 100889401-100889401 on Build GRCh38
rs936670698 Chr.7: 100889816-100889816 on Build GRCh38
rs996914252 Chr.7: 100889507-100889507 on Build GRCh38
rs552902607 Chr.7: 100889678-100889678 on Build GRCh38
rs1021732730 Chr.7: 100889682-100889691 on Build GRCh38
rs983973473 Chr.7: 100889569-100889569 on Build GRCh38
rs970347303 Chr.7: 100889797-100889797 on Build GRCh38
rs1054924561 Chr.7: 100889618-100889618 on Build GRCh38
rs945511923 Chr.7: 100889388-100889388 on Build GRCh38
rs555030402 Chr.7: 100889459-100889459 on Build GRCh38
rs547380547 Chr.7: 100889366-100889366 on Build GRCh38
rs894847485 Chr.7: 100889452-100889452 on Build GRCh38
rs142793647 Chr.7: 100889761-100889761 on Build GRCh38
rs965168094 Chr.7: 100889738-100889738 on Build GRCh38
rs756005843 Chr.7: 100889549-100889549 on Build GRCh38
rs894169602 Chr.7: 100889731-100889731 on Build GRCh38
rs200182652 Chr.7: 100889742-100889761 on Build GRCh38
rs552677066 Chr.7: 100889639-100889639 on Build GRCh38
rs1009791576 Chr.7: 100889493-100889493 on Build GRCh38
rs1028368334 Chr.7: 100889706-100889706 on Build GRCh38
rs28379036 Chr.7: 100889763-100889763 on Build GRCh38

View Full Product Details