Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161160528-161161023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP21
Entrez Gene ID
27005
Gene Name
ubiquitin specific peptidase 21
Gene Aliases
USP16, USP23
UniGene
Hs.8015
Gene Chromosome Location
Chr.1: 161159458-161165726 on Build GRCh38
Transcripts
NM_001014443.2 NM_001319848.1 NM_012475.4 XM_011509414.1 XM_017001036.1 XM_017001037.1 XM_017001038.1 XM_017001039.1 XM_017001040.1 XM_017001041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6122167 c.230G>T p.R77L Chr.1: 161160870-161160870 on Build GRCh38
5605303 c.267C>T p.P89P Chr.1: 161160907-161160907 on Build GRCh38
3477180 c.11C>T p.A4V Chr.1: 161160651-161160651 on Build GRCh38
1295354 c.366G>A p.M122I Chr.1: 161161006-161161006 on Build GRCh38
3477182 c.256C>T p.H86Y Chr.1: 161160896-161160896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373250780 Chr.1: 161160626-161160626 on Build GRCh38
rs773739033 Chr.1: 161160766-161160766 on Build GRCh38
rs763201934 Chr.1: 161160969-161160969 on Build GRCh38
rs566177531 Chr.1: 161160580-161160580 on Build GRCh38
rs370790417 Chr.1: 161160859-161160859 on Build GRCh38
rs779429015 Chr.1: 161160870-161160870 on Build GRCh38
rs764540139 Chr.1: 161160616-161160616 on Build GRCh38
rs749685785 Chr.1: 161160909-161160909 on Build GRCh38
rs372425350 Chr.1: 161160965-161160965 on Build GRCh38
rs770632791 Chr.1: 161160740-161160740 on Build GRCh38
rs149985789 Chr.1: 161160671-161160671 on Build GRCh38
rs745980122 Chr.1: 161160631-161160631 on Build GRCh38
rs544093165 Chr.1: 161160752-161160752 on Build GRCh38
rs375476473 Chr.1: 161160967-161160967 on Build GRCh38
rs780900838 Chr.1: 161160728-161160728 on Build GRCh38
rs751053740 Chr.1: 161160966-161160966 on Build GRCh38
rs777424873 Chr.1: 161160781-161160781 on Build GRCh38
rs758436746 Chr.1: 161160638-161160638 on Build GRCh38
rs554785282 Chr.1: 161160767-161160767 on Build GRCh38
rs976976354 Chr.1: 161160927-161160927 on Build GRCh38
rs749224489 Chr.1: 161160739-161160739 on Build GRCh38
rs367894011 Chr.1: 161160961-161160961 on Build GRCh38
rs369560591 Chr.1: 161160741-161160741 on Build GRCh38
rs137880709 Chr.1: 161160910-161160910 on Build GRCh38
rs377369830 Chr.1: 161160804-161160804 on Build GRCh38
rs191180673 Chr.1: 161160998-161160998 on Build GRCh38
rs893606035 Chr.1: 161160575-161160575 on Build GRCh38
rs560488704 Chr.1: 161160832-161160832 on Build GRCh38
rs147394083 Chr.1: 161160913-161160913 on Build GRCh38
rs140487526 Chr.1: 161160658-161160658 on Build GRCh38
rs753029463 Chr.1: 161160854-161160854 on Build GRCh38
rs764589289 Chr.1: 161160677-161160677 on Build GRCh38
rs1014931023 Chr.1: 161160881-161160881 on Build GRCh38
rs34779722 Chr.1: 161160911-161160911 on Build GRCh38
rs754044354 Chr.1: 161160923-161160923 on Build GRCh38
rs765801629 Chr.1: 161160688-161160688 on Build GRCh38
rs754366733 Chr.1: 161160619-161160619 on Build GRCh38
rs755162566 Chr.1: 161160723-161160723 on Build GRCh38
rs573665408 Chr.1: 161160587-161160587 on Build GRCh38
rs762032221 Chr.1: 161160680-161160680 on Build GRCh38
rs780099574 Chr.1: 161160707-161160707 on Build GRCh38
rs750456510 Chr.1: 161160944-161160944 on Build GRCh38
rs746631922 Chr.1: 161160803-161160803 on Build GRCh38
rs760755383 Chr.1: 161160918-161160918 on Build GRCh38
rs750832234 Chr.1: 161160705-161160705 on Build GRCh38
rs115968198 Chr.1: 161160789-161160789 on Build GRCh38
rs771414801 Chr.1: 161160792-161160792 on Build GRCh38
rs767479799 Chr.1: 161160851-161160851 on Build GRCh38
rs759623543 Chr.1: 161160849-161160849 on Build GRCh38
rs749277844 Chr.1: 161160788-161160788 on Build GRCh38
rs778995626 Chr.1: 161160863-161160863 on Build GRCh38
rs748000177 Chr.1: 161160729-161160729 on Build GRCh38
rs779217194 Chr.1: 161160999-161160999 on Build GRCh38
rs771976856 Chr.1: 161160603-161160604 on Build GRCh38
rs953519657 Chr.1: 161160694-161160694 on Build GRCh38
rs751000774 Chr.1: 161160765-161160765 on Build GRCh38
rs202128032 Chr.1: 161160805-161160805 on Build GRCh38
rs1027833821 Chr.1: 161160681-161160681 on Build GRCh38
rs373004398 Chr.1: 161160888-161160888 on Build GRCh38
rs1048478827 Chr.1: 161160761-161160761 on Build GRCh38
rs947870525 Chr.1: 161160825-161160825 on Build GRCh38
rs201630346 Chr.1: 161160848-161160848 on Build GRCh38
rs775409507 Chr.1: 161160915-161160915 on Build GRCh38
rs745399364 Chr.1: 161160791-161160791 on Build GRCh38
rs756560989 Chr.1: 161160597-161160597 on Build GRCh38
rs758241325 Chr.1: 161160953-161160953 on Build GRCh38
rs747180723 Chr.1: 161160656-161160656 on Build GRCh38
rs145921869 Chr.1: 161160858-161160858 on Build GRCh38
rs747118690 Chr.1: 161160687-161160687 on Build GRCh38
rs757988113 Chr.1: 161160869-161160869 on Build GRCh38
rs751846238 Chr.1: 161160718-161160718 on Build GRCh38
rs780262251 Chr.1: 161160642-161160642 on Build GRCh38
rs186717254 Chr.1: 161160920-161160920 on Build GRCh38
rs370862481 Chr.1: 161160919-161160919 on Build GRCh38
rs773081315 Chr.1: 161160610-161160612 on Build GRCh38
rs773418847 Chr.1: 161160811-161160811 on Build GRCh38
rs199737344 Chr.1: 161160663-161160663 on Build GRCh38
rs138981004 Chr.1: 161160835-161160835 on Build GRCh38
rs769907409 Chr.1: 161160946-161160946 on Build GRCh38
rs780111343 Chr.1: 161160672-161160672 on Build GRCh38
rs765114499 Chr.1: 161160926-161160926 on Build GRCh38
rs745744328 Chr.1: 161160867-161160867 on Build GRCh38
rs765465616 Chr.1: 161160759-161160759 on Build GRCh38
rs757075486 Chr.1: 161160861-161160861 on Build GRCh38
rs994956917 Chr.1: 161160917-161160917 on Build GRCh38
rs780961535 Chr.1: 161160662-161160662 on Build GRCh38
rs776218601 Chr.1: 161160900-161160900 on Build GRCh38
rs147848740 Chr.1: 161160665-161160665 on Build GRCh38
rs1027738645 Chr.1: 161160935-161160935 on Build GRCh38
rs759307340 Chr.1: 161160743-161160743 on Build GRCh38
rs752802166 Chr.1: 161160780-161160780 on Build GRCh38
rs769407463 Chr.1: 161160732-161160732 on Build GRCh38
rs575048074 Chr.1: 161160734-161160734 on Build GRCh38
rs772975124 Chr.1: 161160664-161160664 on Build GRCh38
rs976462968 Chr.1: 161160783-161160783 on Build GRCh38
rs755003081 Chr.1: 161160770-161160770 on Build GRCh38
rs372804422 Chr.1: 161160753-161160753 on Build GRCh38
rs751188861 Chr.1: 161160590-161160590 on Build GRCh38
rs746718961 Chr.1: 161160874-161160874 on Build GRCh38
rs775224335 Chr.1: 161160800-161160800 on Build GRCh38
rs770506309 Chr.1: 161160670-161160670 on Build GRCh38
rs372271090 Chr.1: 161160979-161160979 on Build GRCh38
rs149171020 Chr.1: 161160751-161160751 on Build GRCh38
rs116557343 Chr.1: 161160678-161160678 on Build GRCh38
rs935089193 Chr.1: 161160795-161160795 on Build GRCh38
rs781510108 Chr.1: 161160778-161160778 on Build GRCh38

View Full Product Details