Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119423503-119424009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOV
Entrez Gene ID
4856
Gene Name
nephroblastoma overexpressed
Gene Aliases
CCN3, IBP-9, IGFBP-9, IGFBP9, NOVh
UniGene
Hs.235935
Gene Chromosome Location
Chr.8: 119416312-119424438 on Build GRCh38
Transcripts
NM_002514.3

SNP Detail

SNP ID Chromosome Location
rs890693960 Chr.8: 119423982-119423982 on Build GRCh38
rs1051321209 Chr.8: 119423783-119423783 on Build GRCh38
rs1057722 Chr.8: 119423722-119423722 on Build GRCh38
rs569964568 Chr.8: 119423923-119423923 on Build GRCh38
rs574724230 Chr.8: 119423573-119423573 on Build GRCh38
rs566178584 Chr.8: 119423647-119423647 on Build GRCh38
rs187308086 Chr.8: 119423589-119423589 on Build GRCh38
rs1010424225 Chr.8: 119423576-119423576 on Build GRCh38
rs962913011 Chr.8: 119423532-119423532 on Build GRCh38
rs12547326 Chr.8: 119423857-119423857 on Build GRCh38
rs974086585 Chr.8: 119423570-119423570 on Build GRCh38
rs1054065698 Chr.8: 119423715-119423715 on Build GRCh38
rs918007010 Chr.8: 119423642-119423642 on Build GRCh38
rs535001147 Chr.8: 119423562-119423562 on Build GRCh38
rs969344232 Chr.8: 119423740-119423740 on Build GRCh38
rs1025882132 Chr.8: 119423895-119423895 on Build GRCh38
rs559204595 Chr.8: 119423534-119423534 on Build GRCh38
rs945806456 Chr.8: 119423838-119423838 on Build GRCh38
rs1007470739 Chr.8: 119423785-119423785 on Build GRCh38
rs557009367 Chr.8: 119423625-119423625 on Build GRCh38
rs938595023 Chr.8: 119423791-119423795 on Build GRCh38
rs77725825 Chr.8: 119423710-119423710 on Build GRCh38
rs1008990057 Chr.8: 119423648-119423648 on Build GRCh38
rs1043163206 Chr.8: 119423872-119423872 on Build GRCh38
rs774939690 Chr.8: 119423843-119423843 on Build GRCh38
rs1057380136 Chr.8: 119423571-119423571 on Build GRCh38
rs761909253 Chr.8: 119423792-119423792 on Build GRCh38
rs1056955236 Chr.8: 119423802-119423802 on Build GRCh38
rs386729238 Chr.8: 119423572-119423573 on Build GRCh38
rs927173284 Chr.8: 119423789-119423789 on Build GRCh38
rs904228880 Chr.8: 119423588-119423588 on Build GRCh38
rs995787471 Chr.8: 119423892-119423892 on Build GRCh38
rs905789350 Chr.8: 119423557-119423557 on Build GRCh38
rs951398688 Chr.8: 119423918-119423918 on Build GRCh38
rs890844640 Chr.8: 119423784-119423784 on Build GRCh38
rs938458654 Chr.8: 119423559-119423559 on Build GRCh38
rs115999559 Chr.8: 119423714-119423714 on Build GRCh38
rs545555874 Chr.8: 119423686-119423686 on Build GRCh38
rs775916517 Chr.8: 119423665-119423665 on Build GRCh38
rs945698590 Chr.8: 119423755-119423755 on Build GRCh38
rs191846263 Chr.8: 119423973-119423973 on Build GRCh38
rs1042725526 Chr.8: 119423579-119423579 on Build GRCh38
rs904281169 Chr.8: 119423888-119423888 on Build GRCh38
rs760644424 Chr.8: 119423804-119423804 on Build GRCh38
rs11786312 Chr.8: 119423845-119423845 on Build GRCh38
rs913095251 Chr.8: 119423832-119423832 on Build GRCh38
rs995774792 Chr.8: 119423891-119423891 on Build GRCh38
rs2071518 Chr.8: 119423572-119423572 on Build GRCh38
rs1050064089 Chr.8: 119423977-119423977 on Build GRCh38
rs571077753 Chr.8: 119423561-119423561 on Build GRCh38
rs992674317 Chr.8: 119423603-119423603 on Build GRCh38
rs2071519 Chr.8: 119423668-119423668 on Build GRCh38

View Full Product Details