Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123441533-123442065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDYHV1
Entrez Gene ID
55093
Gene Name
WDYHV motif containing 1
Gene Aliases
C8orf32
UniGene
Hs.18029
Gene Chromosome Location
Chr.8: 123416725-123467527 on Build GRCh38
Transcripts
NM_001283024.1 NM_001283027.1 NM_018024.2 NR_133926.1 XM_011517147.1 XM_011517151.2 XM_017013601.1 XM_017013602.1 XM_017013603.1

SNP Detail

SNP ID Chromosome Location
rs1025335928 Chr.8: 123441719-123441719 on Build GRCh38
rs547767687 Chr.8: 123441632-123441632 on Build GRCh38
rs1007026298 Chr.8: 123441657-123441657 on Build GRCh38
rs763776182 Chr.8: 123442009-123442009 on Build GRCh38
rs927888724 Chr.8: 123441832-123441832 on Build GRCh38
rs760295167 Chr.8: 123441980-123441980 on Build GRCh38
rs761655988 Chr.8: 123442031-123442031 on Build GRCh38
rs756383492 Chr.8: 123441972-123441972 on Build GRCh38
rs151337341 Chr.8: 123441824-123441824 on Build GRCh38
rs562096564 Chr.8: 123441954-123441954 on Build GRCh38
rs767885869 Chr.8: 123441836-123441836 on Build GRCh38
rs955222909 Chr.8: 123441576-123441576 on Build GRCh38
rs935286567 Chr.8: 123441841-123441841 on Build GRCh38
rs1025961436 Chr.8: 123441567-123441567 on Build GRCh38
rs1045284 Chr.8: 123441739-123441739 on Build GRCh38
rs143588946 Chr.8: 123441688-123441688 on Build GRCh38
rs994806709 Chr.8: 123441693-123441693 on Build GRCh38
rs16898316 Chr.8: 123441626-123441626 on Build GRCh38
rs1041238907 Chr.8: 123442019-123442019 on Build GRCh38
rs994212296 Chr.8: 123441819-123441819 on Build GRCh38
rs947369664 Chr.8: 123441929-123441929 on Build GRCh38
rs140037202 Chr.8: 123441687-123441687 on Build GRCh38
rs534122751 Chr.8: 123441844-123441844 on Build GRCh38
rs978461880 Chr.8: 123441930-123441930 on Build GRCh38
rs915288467 Chr.8: 123441897-123441897 on Build GRCh38
rs916775364 Chr.8: 123441982-123441982 on Build GRCh38
rs1045270 Chr.8: 123441559-123441559 on Build GRCh38
rs969202111 Chr.8: 123441807-123441807 on Build GRCh38
rs974877433 Chr.8: 123441877-123441877 on Build GRCh38
rs1038138358 Chr.8: 123442018-123442018 on Build GRCh38
rs916291996 Chr.8: 123441784-123441784 on Build GRCh38
rs939621204 Chr.8: 123441956-123441956 on Build GRCh38
rs1043167312 Chr.8: 123441951-123441951 on Build GRCh38
rs967369818 Chr.8: 123441661-123441669 on Build GRCh38
rs536629401 Chr.8: 123441667-123441667 on Build GRCh38
rs139416705 Chr.8: 123441855-123441855 on Build GRCh38
rs575741160 Chr.8: 123442017-123442017 on Build GRCh38
rs986562618 Chr.8: 123441579-123441579 on Build GRCh38
rs553768982 Chr.8: 123441847-123441847 on Build GRCh38
rs113151923 Chr.8: 123441857-123441857 on Build GRCh38
rs867490090 Chr.8: 123441644-123441644 on Build GRCh38
rs565296081 Chr.8: 123441600-123441600 on Build GRCh38
rs902736935 Chr.8: 123441668-123441668 on Build GRCh38
rs544692133 Chr.8: 123442022-123442022 on Build GRCh38
rs567677517 Chr.8: 123441648-123441648 on Build GRCh38
rs898181514 Chr.8: 123441792-123441792 on Build GRCh38
rs1045291 Chr.8: 123441767-123441767 on Build GRCh38
rs968089101 Chr.8: 123441920-123441920 on Build GRCh38
rs946689423 Chr.8: 123441660-123441661 on Build GRCh38

View Full Product Details