Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24364953-24365450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT20HL1
Entrez Gene ID
100130302
Gene Name
SPT20 homolog, SAGA complex component-like 1
Gene Aliases
FAM48B1, SPT20L
UniGene
Hs.717243
Gene Chromosome Location
Chr.X: 24362761-24365424 on Build GRCh38
Transcripts
NM_001136234.1

SNP Detail

SNP ID Chromosome Location
rs376801277 Chr.X: 24365262-24365262 on Build GRCh38
rs908550206 Chr.X: 24365395-24365395 on Build GRCh38
rs1038908340 Chr.X: 24365139-24365139 on Build GRCh38
rs763434267 Chr.X: 24365077-24365077 on Build GRCh38
rs28689626 Chr.X: 24365198-24365198 on Build GRCh38
rs748218885 Chr.X: 24365012-24365012 on Build GRCh38
rs746349777 Chr.X: 24365418-24365418 on Build GRCh38
rs773777087 Chr.X: 24365226-24365226 on Build GRCh38
rs762046239 Chr.X: 24365009-24365009 on Build GRCh38
rs761865144 Chr.X: 24365329-24365329 on Build GRCh38
rs766793313 Chr.X: 24365091-24365091 on Build GRCh38
rs765423905 Chr.X: 24365187-24365187 on Build GRCh38
rs371616878 Chr.X: 24365228-24365228 on Build GRCh38
rs771365413 Chr.X: 24365270-24365270 on Build GRCh38
rs969940684 Chr.X: 24365253-24365253 on Build GRCh38
rs372971112 Chr.X: 24365388-24365388 on Build GRCh38
rs750296296 Chr.X: 24365350-24365350 on Build GRCh38
rs769729628 Chr.X: 24365020-24365020 on Build GRCh38
rs1029440219 Chr.X: 24365157-24365157 on Build GRCh38
rs766506962 Chr.X: 24365356-24365356 on Build GRCh38
rs996572957 Chr.X: 24365148-24365148 on Build GRCh38
rs749313961 Chr.X: 24365396-24365396 on Build GRCh38
rs369726215 Chr.X: 24365367-24365367 on Build GRCh38
rs755702187 Chr.X: 24365179-24365179 on Build GRCh38
rs765225343 Chr.X: 24365330-24365330 on Build GRCh38
rs749796149 Chr.X: 24365264-24365264 on Build GRCh38
rs749218449 Chr.X: 24364972-24364972 on Build GRCh38
rs747202019 Chr.X: 24365005-24365005 on Build GRCh38
rs757178044 Chr.X: 24364978-24364978 on Build GRCh38
rs374203557 Chr.X: 24365220-24365220 on Build GRCh38
rs769916325 Chr.X: 24365246-24365246 on Build GRCh38
rs866617118 Chr.X: 24365213-24365213 on Build GRCh38
rs752264632 Chr.X: 24365169-24365169 on Build GRCh38
rs756259994 Chr.X: 24365372-24365372 on Build GRCh38
rs955011439 Chr.X: 24365199-24365199 on Build GRCh38
rs939354909 Chr.X: 24365353-24365353 on Build GRCh38
rs763018510 Chr.X: 24365245-24365245 on Build GRCh38
rs769999719 Chr.X: 24365065-24365065 on Build GRCh38
rs759119802 Chr.X: 24365142-24365142 on Build GRCh38
rs770970726 Chr.X: 24365398-24365398 on Build GRCh38
rs750270209 Chr.X: 24365028-24365029 on Build GRCh38
rs755894037 Chr.X: 24365198-24365199 on Build GRCh38
rs778848458 Chr.X: 24365004-24365004 on Build GRCh38
rs779699145 Chr.X: 24365358-24365358 on Build GRCh38
rs759989696 Chr.X: 24365275-24365275 on Build GRCh38
rs185653409 Chr.X: 24365223-24365223 on Build GRCh38
rs779754818 Chr.X: 24365205-24365252 on Build GRCh38
rs767162353 Chr.X: 24365160-24365160 on Build GRCh38
rs755185514 Chr.X: 24365210-24365210 on Build GRCh38
rs775273786 Chr.X: 24365293-24365293 on Build GRCh38
rs774628805 Chr.X: 24365100-24365100 on Build GRCh38
rs776595971 Chr.X: 24365011-24365011 on Build GRCh38
rs763687328 Chr.X: 24365180-24365180 on Build GRCh38
rs373379344 Chr.X: 24365061-24365061 on Build GRCh38
rs774733244 Chr.X: 24365274-24365274 on Build GRCh38
rs779526646 Chr.X: 24365417-24365417 on Build GRCh38
rs772305819 Chr.X: 24365289-24365289 on Build GRCh38
rs763367977 Chr.X: 24365328-24365328 on Build GRCh38
rs745627149 Chr.X: 24365207-24365207 on Build GRCh38
rs763703714 Chr.X: 24365298-24365298 on Build GRCh38
rs760242312 Chr.X: 24365296-24365296 on Build GRCh38
rs199953632 Chr.X: 24365044-24365044 on Build GRCh38

View Full Product Details