Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40627155-40627692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf38
Entrez Gene ID
159013
Gene Name
chromosome X open reading frame 38
Gene Aliases
-
UniGene
Hs.495961
Gene Chromosome Location
Chr.X: 40626921-40647678 on Build GRCh38
Transcripts
NM_144970.2 XM_005272589.2 XM_006724527.3 XM_006724528.2 XM_017029302.1 XM_017029303.1 XM_017029304.1 XM_017029305.1

SNP Detail

SNP ID Chromosome Location
rs1003126594 Chr.X: 40627625-40627625 on Build GRCh38
rs764448571 Chr.X: 40627358-40627358 on Build GRCh38
rs973281840 Chr.X: 40627417-40627417 on Build GRCh38
rs2968912 Chr.X: 40627473-40627473 on Build GRCh38
rs775659236 Chr.X: 40627351-40627351 on Build GRCh38
rs1023506782 Chr.X: 40627588-40627588 on Build GRCh38
rs58287640 Chr.X: 40627184-40627187 on Build GRCh38
rs1016841911 Chr.X: 40627387-40627387 on Build GRCh38
rs867861527 Chr.X: 40627184-40627184 on Build GRCh38
rs774783161 Chr.X: 40627316-40627316 on Build GRCh38
rs760016670 Chr.X: 40627337-40627337 on Build GRCh38
rs999733033 Chr.X: 40627237-40627237 on Build GRCh38
rs771308910 Chr.X: 40627273-40627273 on Build GRCh38
rs1006412323 Chr.X: 40627205-40627205 on Build GRCh38
rs6651613 Chr.X: 40627640-40627640 on Build GRCh38
rs1011192165 Chr.X: 40627585-40627585 on Build GRCh38
rs981784720 Chr.X: 40627374-40627374 on Build GRCh38
rs752132256 Chr.X: 40627615-40627615 on Build GRCh38
rs3180277 Chr.X: 40627268-40627268 on Build GRCh38
rs761006800 Chr.X: 40627352-40627352 on Build GRCh38
rs386824921 Chr.X: 40627184-40627186 on Build GRCh38
rs758796591 Chr.X: 40627534-40627534 on Build GRCh38
rs918779404 Chr.X: 40627434-40627434 on Build GRCh38
rs778105333 Chr.X: 40627612-40627612 on Build GRCh38
rs969020916 Chr.X: 40627338-40627338 on Build GRCh38
rs761548893 Chr.X: 40627185-40627189 on Build GRCh38
rs1023537826 Chr.X: 40627315-40627315 on Build GRCh38
rs188207152 Chr.X: 40627457-40627457 on Build GRCh38
rs901230170 Chr.X: 40627546-40627546 on Build GRCh38
rs951019783 Chr.X: 40627196-40627196 on Build GRCh38
rs750282691 Chr.X: 40627378-40627379 on Build GRCh38
rs958171833 Chr.X: 40627262-40627262 on Build GRCh38
rs750332889 Chr.X: 40627471-40627471 on Build GRCh38
rs772288534 Chr.X: 40627347-40627347 on Build GRCh38
rs183480442 Chr.X: 40627388-40627388 on Build GRCh38
rs761186280 Chr.X: 40627187-40627187 on Build GRCh38
rs199559152 Chr.X: 40627185-40627188 on Build GRCh38
rs928623368 Chr.X: 40627515-40627515 on Build GRCh38
rs753055183 Chr.X: 40627483-40627483 on Build GRCh38
rs765086285 Chr.X: 40627187-40627187 on Build GRCh38
rs181087038 Chr.X: 40627558-40627558 on Build GRCh38
rs1136363 Chr.X: 40627249-40627249 on Build GRCh38
rs927675603 Chr.X: 40627378-40627378 on Build GRCh38
rs931496019 Chr.X: 40627435-40627435 on Build GRCh38
rs1026536997 Chr.X: 40627190-40627190 on Build GRCh38
rs865862420 Chr.X: 40627186-40627186 on Build GRCh38
rs761570367 Chr.X: 40627433-40627433 on Build GRCh38
rs751031878 Chr.X: 40627577-40627577 on Build GRCh38
rs911510231 Chr.X: 40627472-40627472 on Build GRCh38
rs971820750 Chr.X: 40627658-40627658 on Build GRCh38
rs3208430 Chr.X: 40627201-40627201 on Build GRCh38
rs935448667 Chr.X: 40627556-40627556 on Build GRCh38
rs766192877 Chr.X: 40627482-40627482 on Build GRCh38
rs868572647 Chr.X: 40627185-40627185 on Build GRCh38
rs1058983 Chr.X: 40627199-40627199 on Build GRCh38
rs962075614 Chr.X: 40627652-40627652 on Build GRCh38
rs893629676 Chr.X: 40627567-40627567 on Build GRCh38

View Full Product Details