Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46602988-46603491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A7
Entrez Gene ID
84679
Gene Name
solute carrier family 9 member A7
Gene Aliases
NHE-7, NHE7, SLC9A6
UniGene
Hs.91389
Gene Chromosome Location
Chr.X: 46599251-46759172 on Build GRCh38
Transcripts
NM_001257291.1 NM_032591.2 XM_005272681.4 XM_005272682.4 XM_006724564.3 XM_011543990.2 XM_017029905.1 XM_017029906.1

SNP Detail

SNP ID Chromosome Location
rs964797759 Chr.X: 46603181-46603181 on Build GRCh38
rs911715331 Chr.X: 46603429-46603429 on Build GRCh38
rs184044017 Chr.X: 46603211-46603211 on Build GRCh38
rs759968054 Chr.X: 46603313-46603313 on Build GRCh38
rs991059881 Chr.X: 46603178-46603178 on Build GRCh38
rs369904802 Chr.X: 46603186-46603186 on Build GRCh38
rs1001097806 Chr.X: 46603135-46603135 on Build GRCh38
rs188499408 Chr.X: 46603109-46603109 on Build GRCh38
rs779907936 Chr.X: 46603128-46603128 on Build GRCh38
rs978387066 Chr.X: 46603261-46603261 on Build GRCh38
rs1051972665 Chr.X: 46603407-46603407 on Build GRCh38
rs934401620 Chr.X: 46603387-46603387 on Build GRCh38
rs773364214 Chr.X: 46603177-46603177 on Build GRCh38
rs1020744581 Chr.X: 46603078-46603078 on Build GRCh38
rs180989458 Chr.X: 46603248-46603248 on Build GRCh38
rs950798555 Chr.X: 46603088-46603088 on Build GRCh38
rs1044956748 Chr.X: 46603349-46603349 on Build GRCh38
rs534543895 Chr.X: 46603015-46603015 on Build GRCh38
rs942248292 Chr.X: 46603264-46603264 on Build GRCh38
rs1057076787 Chr.X: 46603445-46603446 on Build GRCh38
rs553130650 Chr.X: 46603242-46603242 on Build GRCh38
rs924284588 Chr.X: 46603340-46603340 on Build GRCh38
rs983666113 Chr.X: 46603235-46603235 on Build GRCh38
rs868681372 Chr.X: 46603267-46603267 on Build GRCh38
rs188339317 Chr.X: 46603226-46603226 on Build GRCh38
rs772099078 Chr.X: 46603158-46603158 on Build GRCh38
rs2272822 Chr.X: 46603145-46603145 on Build GRCh38
rs959831890 Chr.X: 46603172-46603172 on Build GRCh38
rs745645793 Chr.X: 46603038-46603038 on Build GRCh38
rs770299625 Chr.X: 46603359-46603359 on Build GRCh38
rs938754023 Chr.X: 46603392-46603392 on Build GRCh38
rs775647044 Chr.X: 46603037-46603037 on Build GRCh38
rs975774613 Chr.X: 46603194-46603194 on Build GRCh38

View Full Product Details