Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48706177-48706683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUV39H1
Entrez Gene ID
6839
Gene Name
suppressor of variegation 3-9 homolog 1
Gene Aliases
H3-K9-HMTase 1, KMT1A, MG44, SUV39H
UniGene
Hs.522639
Gene Chromosome Location
Chr.X: 48695554-48709016 on Build GRCh38
Transcripts
NM_001282166.1 NM_003173.3

SNP Detail

SNP ID Chromosome Location
rs377027180 Chr.X: 48706449-48706449 on Build GRCh38
rs141546891 Chr.X: 48706354-48706354 on Build GRCh38
rs375610379 Chr.X: 48706204-48706204 on Build GRCh38
rs782656874 Chr.X: 48706227-48706227 on Build GRCh38
rs973790599 Chr.X: 48706493-48706493 on Build GRCh38
rs202042569 Chr.X: 48706424-48706424 on Build GRCh38
rs782209222 Chr.X: 48706491-48706491 on Build GRCh38
rs782340673 Chr.X: 48706289-48706289 on Build GRCh38
rs947449536 Chr.X: 48706318-48706318 on Build GRCh38
rs782568943 Chr.X: 48706258-48706258 on Build GRCh38
rs781877031 Chr.X: 48706425-48706425 on Build GRCh38
rs782037864 Chr.X: 48706608-48706608 on Build GRCh38
rs781900909 Chr.X: 48706539-48706539 on Build GRCh38
rs781965749 Chr.X: 48706446-48706446 on Build GRCh38
rs782533904 Chr.X: 48706429-48706429 on Build GRCh38
rs782051846 Chr.X: 48706364-48706364 on Build GRCh38
rs368988897 Chr.X: 48706242-48706242 on Build GRCh38
rs782111188 Chr.X: 48706210-48706210 on Build GRCh38
rs79235204 Chr.X: 48706499-48706499 on Build GRCh38
rs782312190 Chr.X: 48706466-48706466 on Build GRCh38
rs372441780 Chr.X: 48706634-48706634 on Build GRCh38
rs782355758 Chr.X: 48706524-48706524 on Build GRCh38
rs782715815 Chr.X: 48706631-48706631 on Build GRCh38
rs782286731 Chr.X: 48706228-48706228 on Build GRCh38
rs782792759 Chr.X: 48706658-48706658 on Build GRCh38
rs782133049 Chr.X: 48706463-48706464 on Build GRCh38
rs782200027 Chr.X: 48706282-48706282 on Build GRCh38
rs782767775 Chr.X: 48706659-48706659 on Build GRCh38
rs782423798 Chr.X: 48706555-48706555 on Build GRCh38
rs781974468 Chr.X: 48706297-48706297 on Build GRCh38
rs782577741 Chr.X: 48706469-48706469 on Build GRCh38
rs138513845 Chr.X: 48706306-48706306 on Build GRCh38
rs184563517 Chr.X: 48706211-48706211 on Build GRCh38
rs782654392 Chr.X: 48706452-48706452 on Build GRCh38
rs963892037 Chr.X: 48706486-48706486 on Build GRCh38
rs782700678 Chr.X: 48706447-48706447 on Build GRCh38
rs781792051 Chr.X: 48706432-48706432 on Build GRCh38
rs201867779 Chr.X: 48706310-48706310 on Build GRCh38
rs782035829 Chr.X: 48706319-48706319 on Build GRCh38
rs782735428 Chr.X: 48706423-48706423 on Build GRCh38
rs782454438 Chr.X: 48706442-48706442 on Build GRCh38
rs782043171 Chr.X: 48706605-48706605 on Build GRCh38
rs781827609 Chr.X: 48706649-48706649 on Build GRCh38
rs782817837 Chr.X: 48706430-48706430 on Build GRCh38
rs782724003 Chr.X: 48706393-48706393 on Build GRCh38
rs782287892 Chr.X: 48706454-48706454 on Build GRCh38
rs1026744699 Chr.X: 48706575-48706575 on Build GRCh38
rs373029228 Chr.X: 48706373-48706373 on Build GRCh38
rs782132107 Chr.X: 48706545-48706545 on Build GRCh38
rs782057614 Chr.X: 48706617-48706617 on Build GRCh38

View Full Product Details