Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212858068-212858563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR1
Entrez Gene ID
28982
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Gene Aliases
AXPC1, FLVCR, MFSD7B, PCA, PCARP
UniGene
Hs.7055
Gene Chromosome Location
Chr.1: 212858255-212899363 on Build GRCh38
Transcripts
NM_014053.3 XM_011509446.2 XM_011509447.2 XM_011509448.1 NR_027285.1 NR_027286.1

Gene Symbol
FLVCR1-AS1
Entrez Gene ID
642946
Gene Name
FLVCR1 antisense RNA 1 (head to head)
Gene Aliases
LQK1, NCRNA00292
UniGene
Hs.552649
Gene Chromosome Location
Chr.1: 212856604-212858138 on Build GRCh38
Transcripts
NM_014053.3 XM_011509446.2 XM_011509447.2 XM_011509448.1 NR_027285.1 NR_027286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6534205 c.71C>G p.P24R Chr.1: 212858523-212858523 on Build GRCh38
6541701 c.31G>T p.A11S Chr.1: 212858483-212858483 on Build GRCh38
6425377 c.91G>A p.V31I Chr.1: 212858543-212858543 on Build GRCh38
6428522 c.16G>T p.D6Y Chr.1: 212858468-212858468 on Build GRCh38
4956199 c.98A>G p.K33R Chr.1: 212858550-212858550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947617988 Chr.1: 212858321-212858321 on Build GRCh38
rs1043289275 Chr.1: 212858257-212858257 on Build GRCh38
rs979404283 Chr.1: 212858326-212858326 on Build GRCh38
rs1057226211 Chr.1: 212858169-212858169 on Build GRCh38
rs886045922 Chr.1: 212858417-212858417 on Build GRCh38
rs2201602 Chr.1: 212858131-212858131 on Build GRCh38
rs888276114 Chr.1: 212858417-212858417 on Build GRCh38
rs916015252 Chr.1: 212858477-212858477 on Build GRCh38
rs562153726 Chr.1: 212858523-212858523 on Build GRCh38
rs1005876471 Chr.1: 212858210-212858210 on Build GRCh38
rs768206151 Chr.1: 212858513-212858513 on Build GRCh38
rs969279368 Chr.1: 212858515-212858515 on Build GRCh38
rs964372054 Chr.1: 212858139-212858139 on Build GRCh38
rs927598088 Chr.1: 212858339-212858339 on Build GRCh38
rs1057131599 Chr.1: 212858351-212858351 on Build GRCh38
rs536673071 Chr.1: 212858270-212858270 on Build GRCh38
rs747866785 Chr.1: 212858522-212858522 on Build GRCh38
rs994272478 Chr.1: 212858178-212858178 on Build GRCh38
rs554048351 Chr.1: 212858299-212858299 on Build GRCh38
rs41296692 Chr.1: 212858366-212858366 on Build GRCh38
rs1048027270 Chr.1: 212858209-212858209 on Build GRCh38
rs41296686 Chr.1: 212858141-212858141 on Build GRCh38
rs1047179571 Chr.1: 212858368-212858368 on Build GRCh38
rs2201603 Chr.1: 212858367-212858367 on Build GRCh38
rs1043348269 Chr.1: 212858145-212858145 on Build GRCh38
rs992883082 Chr.1: 212858476-212858476 on Build GRCh38
rs41296688 Chr.1: 212858193-212858193 on Build GRCh38
rs886045924 Chr.1: 212858503-212858503 on Build GRCh38
rs776069050 Chr.1: 212858254-212858254 on Build GRCh38
rs576457204 Chr.1: 212858480-212858480 on Build GRCh38
rs765796589 Chr.1: 212858376-212858376 on Build GRCh38
rs747330928 Chr.1: 212858260-212858260 on Build GRCh38
rs1005409961 Chr.1: 212858428-212858428 on Build GRCh38
rs913383179 Chr.1: 212858315-212858315 on Build GRCh38
rs534759890 Chr.1: 212858293-212858293 on Build GRCh38
rs777071598 Chr.1: 212858531-212858531 on Build GRCh38
rs922288059 Chr.1: 212858272-212858272 on Build GRCh38
rs998058913 Chr.1: 212858459-212858459 on Build GRCh38
rs1017337739 Chr.1: 212858325-212858325 on Build GRCh38
rs772978535 Chr.1: 212858448-212858448 on Build GRCh38
rs1022067741 Chr.1: 212858503-212858503 on Build GRCh38
rs963667001 Chr.1: 212858247-212858247 on Build GRCh38
rs910846815 Chr.1: 212858100-212858100 on Build GRCh38
rs765337184 Chr.1: 212858296-212858297 on Build GRCh38
rs373452417 Chr.1: 212858297-212858297 on Build GRCh38
rs762737335 Chr.1: 212858473-212858473 on Build GRCh38
rs927762933 Chr.1: 212858397-212858397 on Build GRCh38
rs533056358 Chr.1: 212858196-212858196 on Build GRCh38
rs922382924 Chr.1: 212858109-212858109 on Build GRCh38
rs867110653 Chr.1: 212858363-212858363 on Build GRCh38
rs111734301 Chr.1: 212858492-212858492 on Build GRCh38
rs554891521 Chr.1: 212858211-212858211 on Build GRCh38
rs886045921 Chr.1: 212858272-212858272 on Build GRCh38
rs927813757 Chr.1: 212858210-212858217 on Build GRCh38
rs904774870 Chr.1: 212858267-212858267 on Build GRCh38
rs541543586 Chr.1: 212858543-212858543 on Build GRCh38
rs151032809 Chr.1: 212858298-212858299 on Build GRCh38
rs531752151 Chr.1: 212858125-212858125 on Build GRCh38
rs529044833 Chr.1: 212858154-212858154 on Build GRCh38
rs374379789 Chr.1: 212858521-212858521 on Build GRCh38
rs963783024 Chr.1: 212858105-212858105 on Build GRCh38
rs937590890 Chr.1: 212858348-212858348 on Build GRCh38
rs1053324425 Chr.1: 212858542-212858542 on Build GRCh38
rs41296690 Chr.1: 212858253-212858253 on Build GRCh38
rs746296931 Chr.1: 212858534-212858534 on Build GRCh38
rs12404942 Chr.1: 212858122-212858122 on Build GRCh38
rs34019267 Chr.1: 212858542-212858542 on Build GRCh38
rs567238059 Chr.1: 212858279-212858279 on Build GRCh38
rs371573598 Chr.1: 212858290-212858290 on Build GRCh38
rs947592568 Chr.1: 212858138-212858138 on Build GRCh38
rs17676799 Chr.1: 212858297-212858297 on Build GRCh38
rs543847452 Chr.1: 212858501-212858501 on Build GRCh38
rs555000488 Chr.1: 212858277-212858277 on Build GRCh38
rs987915815 Chr.1: 212858305-212858305 on Build GRCh38
rs1029247819 Chr.1: 212858470-212858470 on Build GRCh38
rs201384413 Chr.1: 212858438-212858438 on Build GRCh38
rs35119480 Chr.1: 212858535-212858535 on Build GRCh38
rs976357872 Chr.1: 212858271-212858271 on Build GRCh38
rs895875259 Chr.1: 212858170-212858170 on Build GRCh38
rs569277164 Chr.1: 212858244-212858244 on Build GRCh38
rs937538361 Chr.1: 212858527-212858527 on Build GRCh38
rs547266558 Chr.1: 212858180-212858180 on Build GRCh38
rs1057259959 Chr.1: 212858242-212858242 on Build GRCh38
rs2201601 Chr.1: 212858106-212858106 on Build GRCh38
rs1001560179 Chr.1: 212858163-212858163 on Build GRCh38
rs886045923 Chr.1: 212858494-212858494 on Build GRCh38
rs529365517 Chr.1: 212858540-212858540 on Build GRCh38
rs899735028 Chr.1: 212858455-212858455 on Build GRCh38
rs1013346043 Chr.1: 212858250-212858250 on Build GRCh38

View Full Product Details