Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 28677244-28677750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAMBI
Entrez Gene ID
25805
Gene Name
BMP and activin membrane bound inhibitor
Gene Aliases
NMA
UniGene
Hs.533336
Gene Chromosome Location
Chr.10: 28677495-28682939 on Build GRCh38
Transcripts
NM_012342.2

SNP Detail

SNP ID Chromosome Location
rs1041694262 Chr.10: 28677347-28677347 on Build GRCh38
rs935936798 Chr.10: 28677513-28677513 on Build GRCh38
rs984997891 Chr.10: 28677383-28677383 on Build GRCh38
rs568617902 Chr.10: 28677690-28677690 on Build GRCh38
rs952379670 Chr.10: 28677380-28677380 on Build GRCh38
rs975118118 Chr.10: 28677398-28677398 on Build GRCh38
rs534560506 Chr.10: 28677401-28677402 on Build GRCh38
rs1006661160 Chr.10: 28677601-28677601 on Build GRCh38
rs868669102 Chr.10: 28677315-28677315 on Build GRCh38
rs369774203 Chr.10: 28677289-28677289 on Build GRCh38
rs955120640 Chr.10: 28677554-28677554 on Build GRCh38
rs950947251 Chr.10: 28677649-28677649 on Build GRCh38
rs903124307 Chr.10: 28677443-28677443 on Build GRCh38
rs1047732948 Chr.10: 28677532-28677532 on Build GRCh38
rs578027365 Chr.10: 28677269-28677269 on Build GRCh38
rs1039015230 Chr.10: 28677609-28677609 on Build GRCh38
rs1054820307 Chr.10: 28677516-28677516 on Build GRCh38
rs948585238 Chr.10: 28677520-28677520 on Build GRCh38
rs997707068 Chr.10: 28677548-28677548 on Build GRCh38
rs1048314582 Chr.10: 28677589-28677589 on Build GRCh38
rs112446885 Chr.10: 28677386-28677386 on Build GRCh38
rs909658897 Chr.10: 28677324-28677324 on Build GRCh38
rs1012701424 Chr.10: 28677458-28677459 on Build GRCh38
rs189067155 Chr.10: 28677553-28677553 on Build GRCh38
rs1024262803 Chr.10: 28677359-28677359 on Build GRCh38
rs922374269 Chr.10: 28677506-28677506 on Build GRCh38
rs985351952 Chr.10: 28677319-28677319 on Build GRCh38
rs561565965 Chr.10: 28677631-28677631 on Build GRCh38
rs1016899065 Chr.10: 28677655-28677655 on Build GRCh38
rs572801800 Chr.10: 28677358-28677358 on Build GRCh38
rs935892357 Chr.10: 28677450-28677450 on Build GRCh38
rs1017772347 Chr.10: 28677501-28677501 on Build GRCh38
rs865809050 Chr.10: 28677500-28677500 on Build GRCh38
rs1000433337 Chr.10: 28677274-28677274 on Build GRCh38
rs949200545 Chr.10: 28677573-28677573 on Build GRCh38
rs971629151 Chr.10: 28677303-28677303 on Build GRCh38
rs900556184 Chr.10: 28677625-28677625 on Build GRCh38
rs1024378967 Chr.10: 28677472-28677472 on Build GRCh38
rs971411411 Chr.10: 28677368-28677368 on Build GRCh38
rs980316792 Chr.10: 28677325-28677326 on Build GRCh38
rs930863153 Chr.10: 28677318-28677318 on Build GRCh38
rs1030481505 Chr.10: 28677549-28677549 on Build GRCh38
rs959029371 Chr.10: 28677281-28677281 on Build GRCh38
rs955089385 Chr.10: 28677509-28677509 on Build GRCh38
rs894260745 Chr.10: 28677457-28677457 on Build GRCh38
rs972250827 Chr.10: 28677308-28677308 on Build GRCh38
rs868171290 Chr.10: 28677616-28677616 on Build GRCh38
rs1045557244 Chr.10: 28677577-28677577 on Build GRCh38
rs949147107 Chr.10: 28677730-28677730 on Build GRCh38
rs182455644 Chr.10: 28677558-28677558 on Build GRCh38
rs1030534652 Chr.10: 28677646-28677646 on Build GRCh38
rs1041390460 Chr.10: 28677440-28677440 on Build GRCh38
rs574033493 Chr.10: 28677483-28677483 on Build GRCh38
rs71492575 Chr.10: 28677304-28677304 on Build GRCh38
rs894577479 Chr.10: 28677357-28677357 on Build GRCh38
rs369460072 Chr.10: 28677480-28677480 on Build GRCh38
rs113559127 Chr.10: 28677396-28677396 on Build GRCh38
rs1054095950 Chr.10: 28677598-28677598 on Build GRCh38
rs983747788 Chr.10: 28677651-28677651 on Build GRCh38
rs786873 Chr.10: 28677702-28677702 on Build GRCh38
rs560721213 Chr.10: 28677590-28677590 on Build GRCh38
rs186200792 Chr.10: 28677667-28677667 on Build GRCh38
rs963094668 Chr.10: 28677664-28677664 on Build GRCh38
rs887733686 Chr.10: 28677539-28677539 on Build GRCh38
rs7099888 Chr.10: 28677370-28677370 on Build GRCh38
rs993485058 Chr.10: 28677487-28677487 on Build GRCh38
rs1046227683 Chr.10: 28677576-28677576 on Build GRCh38
rs1011505088 Chr.10: 28677608-28677608 on Build GRCh38
rs549279775 Chr.10: 28677605-28677605 on Build GRCh38
rs928439842 Chr.10: 28677578-28677578 on Build GRCh38
rs538997133 Chr.10: 28677724-28677724 on Build GRCh38
rs924382872 Chr.10: 28677555-28677555 on Build GRCh38
rs942470656 Chr.10: 28677397-28677397 on Build GRCh38
rs1054303900 Chr.10: 28677453-28677453 on Build GRCh38
rs550291941 Chr.10: 28677263-28677263 on Build GRCh38
rs1018305863 Chr.10: 28677541-28677541 on Build GRCh38
rs900772658 Chr.10: 28677613-28677613 on Build GRCh38
rs813095 Chr.10: 28677687-28677687 on Build GRCh38
rs933676065 Chr.10: 28677423-28677423 on Build GRCh38
rs528012770 Chr.10: 28677594-28677594 on Build GRCh38
rs919508823 Chr.10: 28677314-28677314 on Build GRCh38
rs1033131953 Chr.10: 28677277-28677277 on Build GRCh38
rs976238229 Chr.10: 28677504-28677504 on Build GRCh38
rs1045525611 Chr.10: 28677524-28677524 on Build GRCh38
rs562868226 Chr.10: 28677552-28677552 on Build GRCh38
rs1000060792 Chr.10: 28677355-28677355 on Build GRCh38
rs964943629 Chr.10: 28677543-28677543 on Build GRCh38
rs1033241469 Chr.10: 28677356-28677356 on Build GRCh38
rs545429345 Chr.10: 28677581-28677581 on Build GRCh38
rs933896941 Chr.10: 28677327-28677327 on Build GRCh38
rs915839762 Chr.10: 28677576-28677577 on Build GRCh38
rs1017664331 Chr.10: 28677617-28677617 on Build GRCh38
rs1085557 Chr.10: 28677656-28677656 on Build GRCh38
rs528960991 Chr.10: 28677647-28677647 on Build GRCh38
rs924242741 Chr.10: 28677512-28677512 on Build GRCh38
rs544719740 Chr.10: 28677530-28677530 on Build GRCh38
rs1888085 Chr.10: 28677565-28677565 on Build GRCh38
rs893167221 Chr.10: 28677606-28677606 on Build GRCh38
rs1024231778 Chr.10: 28677302-28677302 on Build GRCh38
rs557587889 Chr.10: 28677317-28677317 on Build GRCh38
rs935301041 Chr.10: 28677595-28677595 on Build GRCh38
rs989819436 Chr.10: 28677571-28677571 on Build GRCh38
rs977103540 Chr.10: 28677510-28677510 on Build GRCh38
rs1026554229 Chr.10: 28677379-28677379 on Build GRCh38
rs1026712378 Chr.10: 28677499-28677499 on Build GRCh38
rs564241355 Chr.10: 28677305-28677317 on Build GRCh38
rs907317378 Chr.10: 28677525-28677525 on Build GRCh38
rs903090397 Chr.10: 28677352-28677352 on Build GRCh38
rs1039398779 Chr.10: 28677326-28677326 on Build GRCh38
rs916339156 Chr.10: 28677727-28677727 on Build GRCh38
rs957127367 Chr.10: 28677696-28677696 on Build GRCh38

View Full Product Details