Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45440125-45440646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3867228 c.1131G>A p.E377E Chr.10: 45440579-45440579 on Build GRCh38
918278 c.1038C>T p.Y346Y Chr.10: 45440486-45440486 on Build GRCh38
6890775 c.1044G>A p.W348* Chr.10: 45440492-45440492 on Build GRCh38
6598703 c.1111C>T p.R371* Chr.10: 45440559-45440559 on Build GRCh38
4661211 c.1154G>A p.R385H Chr.10: 45440602-45440602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749106042 Chr.10: 45440503-45440503 on Build GRCh38
rs964692372 Chr.10: 45440332-45440332 on Build GRCh38
rs1003091123 Chr.10: 45440290-45440290 on Build GRCh38
rs1009787385 Chr.10: 45440476-45440476 on Build GRCh38
rs553216686 Chr.10: 45440373-45440373 on Build GRCh38
rs948067239 Chr.10: 45440237-45440237 on Build GRCh38
rs754005003 Chr.10: 45440568-45440569 on Build GRCh38
rs763945960 Chr.10: 45440591-45440591 on Build GRCh38
rs755648894 Chr.10: 45440587-45440587 on Build GRCh38
rs111859727 Chr.10: 45440422-45440422 on Build GRCh38
rs780812745 Chr.10: 45440470-45440470 on Build GRCh38
rs776007287 Chr.10: 45440406-45440406 on Build GRCh38
rs765649864 Chr.10: 45440566-45440566 on Build GRCh38
rs886295898 Chr.10: 45440292-45440292 on Build GRCh38
rs748963509 Chr.10: 45440529-45440529 on Build GRCh38
rs573162552 Chr.10: 45440301-45440301 on Build GRCh38
rs1004801305 Chr.10: 45440293-45440293 on Build GRCh38
rs747097580 Chr.10: 45440541-45440541 on Build GRCh38
rs746046438 Chr.10: 45440515-45440515 on Build GRCh38
rs774180136 Chr.10: 45440573-45440573 on Build GRCh38
rs766524341 Chr.10: 45440412-45440412 on Build GRCh38
rs577789262 Chr.10: 45440379-45440379 on Build GRCh38
rs762229821 Chr.10: 45440449-45440449 on Build GRCh38
rs373773649 Chr.10: 45440487-45440487 on Build GRCh38
rs923147311 Chr.10: 45440371-45440371 on Build GRCh38
rs759303080 Chr.10: 45440574-45440574 on Build GRCh38
rs1045059978 Chr.10: 45440513-45440513 on Build GRCh38
rs774553469 Chr.10: 45440388-45440388 on Build GRCh38
rs962999571 Chr.10: 45440361-45440361 on Build GRCh38
rs191951798 Chr.10: 45440561-45440561 on Build GRCh38
rs772088915 Chr.10: 45440403-45440403 on Build GRCh38
rs779501045 Chr.10: 45440460-45440460 on Build GRCh38
rs371711392 Chr.10: 45440486-45440486 on Build GRCh38
rs954471371 Chr.10: 45440443-45440443 on Build GRCh38
rs773001478 Chr.10: 45440282-45440282 on Build GRCh38
rs905509183 Chr.10: 45440198-45440198 on Build GRCh38
rs976113188 Chr.10: 45440362-45440362 on Build GRCh38
rs751201354 Chr.10: 45440456-45440456 on Build GRCh38
rs749088572 Chr.10: 45440384-45440384 on Build GRCh38
rs968129739 Chr.10: 45440280-45440280 on Build GRCh38
rs964795685 Chr.10: 45440613-45440613 on Build GRCh38
rs757072938 Chr.10: 45440598-45440598 on Build GRCh38
rs1006398476 Chr.10: 45440166-45440166 on Build GRCh38
rs762095751 Chr.10: 45440563-45440563 on Build GRCh38
rs200185014 Chr.10: 45440532-45440532 on Build GRCh38
rs367766299 Chr.10: 45440474-45440474 on Build GRCh38
rs28395870 Chr.10: 45440500-45440500 on Build GRCh38
rs1017710645 Chr.10: 45440305-45440305 on Build GRCh38
rs370360074 Chr.10: 45440418-45440418 on Build GRCh38
rs781545895 Chr.10: 45440229-45440229 on Build GRCh38
rs1047630934 Chr.10: 45440560-45440560 on Build GRCh38
rs779128700 Chr.10: 45440514-45440514 on Build GRCh38
rs138855994 Chr.10: 45440434-45440434 on Build GRCh38
rs368161745 Chr.10: 45440451-45440451 on Build GRCh38
rs547998614 Chr.10: 45440421-45440421 on Build GRCh38
rs778716193 Chr.10: 45440604-45440604 on Build GRCh38
rs767260151 Chr.10: 45440577-45440577 on Build GRCh38
rs1002522981 Chr.10: 45440199-45440199 on Build GRCh38
rs989659749 Chr.10: 45440473-45440473 on Build GRCh38
rs887502405 Chr.10: 45440145-45440145 on Build GRCh38
rs755310822 Chr.10: 45440618-45440618 on Build GRCh38
rs569008598 Chr.10: 45440200-45440200 on Build GRCh38
rs534383217 Chr.10: 45440167-45440167 on Build GRCh38
rs542698432 Chr.10: 45440559-45440559 on Build GRCh38
rs1045090025 Chr.10: 45440241-45440241 on Build GRCh38
rs3780914 Chr.10: 45440296-45440296 on Build GRCh38
rs142255723 Chr.10: 45440218-45440218 on Build GRCh38
rs1056953890 Chr.10: 45440291-45440291 on Build GRCh38
rs906638463 Chr.10: 45440289-45440289 on Build GRCh38
rs770647919 Chr.10: 45440511-45440511 on Build GRCh38
rs987668520 Chr.10: 45440179-45440179 on Build GRCh38
rs28395860 Chr.10: 45440457-45440457 on Build GRCh38
rs1021548136 Chr.10: 45440240-45440240 on Build GRCh38
rs935576287 Chr.10: 45440213-45440213 on Build GRCh38
rs528054699 Chr.10: 45440615-45440615 on Build GRCh38
rs143675525 Chr.10: 45440593-45440593 on Build GRCh38
rs745636462 Chr.10: 45440618-45440618 on Build GRCh38
rs759623514 Chr.10: 45440399-45440399 on Build GRCh38
rs534625984 Chr.10: 45440317-45440317 on Build GRCh38
rs146362560 Chr.10: 45440444-45440444 on Build GRCh38
rs777753439 Chr.10: 45440497-45440497 on Build GRCh38

View Full Product Details