Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87057361-87057876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUD1
Entrez Gene ID
2746
Gene Name
glutamate dehydrogenase 1
Gene Aliases
GDH, GDH1, GLUD
UniGene
Hs.500409
Gene Chromosome Location
Chr.10: 87050202-87095047 on Build GRCh38
Transcripts
NM_001318900.1 NM_001318901.1 NM_001318902.1 NM_001318904.1 NM_001318905.1 NM_001318906.1 NM_005271.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402913 c.1418G>A p.S473N Chr.10: 87057767-87057767 on Build GRCh38
921013 c.1452T>C p.T484T Chr.10: 87057733-87057733 on Build GRCh38
6386753 c.1486A>G p.R496G Chr.10: 87057699-87057699 on Build GRCh38
6390355 c.1451C>A p.T484N Chr.10: 87057734-87057734 on Build GRCh38
1349433 c.1447G>T p.G483* Chr.10: 87057738-87057738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749840458 Chr.10: 87057817-87057817 on Build GRCh38
rs753000338 Chr.10: 87057800-87057800 on Build GRCh38
rs771182791 Chr.10: 87057677-87057677 on Build GRCh38
rs547803673 Chr.10: 87057570-87057570 on Build GRCh38
rs1018370498 Chr.10: 87057463-87057463 on Build GRCh38
rs36079084 Chr.10: 87057567-87057567 on Build GRCh38
rs200311046 Chr.10: 87057657-87057657 on Build GRCh38
rs778427422 Chr.10: 87057472-87057472 on Build GRCh38
rs141844887 Chr.10: 87057715-87057715 on Build GRCh38
rs937164395 Chr.10: 87057459-87057459 on Build GRCh38
rs113338732 Chr.10: 87057457-87057457 on Build GRCh38
rs564117047 Chr.10: 87057575-87057576 on Build GRCh38
rs35186250 Chr.10: 87057822-87057822 on Build GRCh38
rs763061216 Chr.10: 87057822-87057824 on Build GRCh38
rs121909731 Chr.10: 87057692-87057692 on Build GRCh38
rs761630039 Chr.10: 87057821-87057822 on Build GRCh38
rs747444397 Chr.10: 87057651-87057652 on Build GRCh38
rs746410100 Chr.10: 87057808-87057813 on Build GRCh38
rs775680889 Chr.10: 87057821-87057821 on Build GRCh38
rs114713986 Chr.10: 87057458-87057458 on Build GRCh38
rs918025178 Chr.10: 87057577-87057577 on Build GRCh38
rs1013898320 Chr.10: 87057528-87057528 on Build GRCh38
rs546653532 Chr.10: 87057476-87057476 on Build GRCh38
rs182162017 Chr.10: 87057600-87057600 on Build GRCh38
rs759865842 Chr.10: 87057691-87057691 on Build GRCh38
rs7069764 Chr.10: 87057507-87057507 on Build GRCh38
rs550609501 Chr.10: 87057786-87057787 on Build GRCh38
rs566646935 Chr.10: 87057733-87057733 on Build GRCh38
rs774681697 Chr.10: 87057680-87057680 on Build GRCh38
rs753288576 Chr.10: 87057645-87057645 on Build GRCh38
rs777065865 Chr.10: 87057794-87057821 on Build GRCh38
rs936611720 Chr.10: 87057630-87057630 on Build GRCh38
rs758062208 Chr.10: 87057750-87057750 on Build GRCh38
rs566809359 Chr.10: 87057822-87057825 on Build GRCh38
rs773836257 Chr.10: 87057673-87057673 on Build GRCh38
rs781469869 Chr.10: 87057790-87057790 on Build GRCh38
rs537863410 Chr.10: 87057823-87057825 on Build GRCh38
rs764466967 Chr.10: 87057822-87057823 on Build GRCh38
rs748631904 Chr.10: 87057666-87057666 on Build GRCh38
rs1007689728 Chr.10: 87057787-87057787 on Build GRCh38
rs564330077 Chr.10: 87057445-87057445 on Build GRCh38
rs781559964 Chr.10: 87057664-87057664 on Build GRCh38
rs771092987 Chr.10: 87057819-87057819 on Build GRCh38
rs753866508 Chr.10: 87057732-87057732 on Build GRCh38
rs749313694 Chr.10: 87057675-87057675 on Build GRCh38
rs769723479 Chr.10: 87057819-87057820 on Build GRCh38
rs532528971 Chr.10: 87057510-87057510 on Build GRCh38
rs944472420 Chr.10: 87057839-87057839 on Build GRCh38
rs71477638 Chr.10: 87057837-87057837 on Build GRCh38
rs555550604 Chr.10: 87057837-87057837 on Build GRCh38
rs531061878 Chr.10: 87057399-87057399 on Build GRCh38
rs756854673 Chr.10: 87057651-87057651 on Build GRCh38
rs949468337 Chr.10: 87057583-87057583 on Build GRCh38
rs373872795 Chr.10: 87057724-87057724 on Build GRCh38
rs1000857570 Chr.10: 87057637-87057637 on Build GRCh38
rs886047374 Chr.10: 87057793-87057793 on Build GRCh38
rs1039852622 Chr.10: 87057793-87057793 on Build GRCh38
rs558023062 Chr.10: 87057825-87057825 on Build GRCh38
rs548114658 Chr.10: 87057431-87057431 on Build GRCh38
rs778517853 Chr.10: 87057656-87057656 on Build GRCh38
rs1026985026 Chr.10: 87057530-87057530 on Build GRCh38
rs869215937 Chr.10: 87057836-87057837 on Build GRCh38
rs200210202 Chr.10: 87057812-87057812 on Build GRCh38
rs774963322 Chr.10: 87057805-87057805 on Build GRCh38
rs926403572 Chr.10: 87057605-87057605 on Build GRCh38
rs756559520 Chr.10: 87057804-87057804 on Build GRCh38
rs879233103 Chr.10: 87057821-87057821 on Build GRCh38
rs755297500 Chr.10: 87057660-87057660 on Build GRCh38
rs763929519 Chr.10: 87057726-87057726 on Build GRCh38
rs764760316 Chr.10: 87057744-87057744 on Build GRCh38
rs4370842 Chr.10: 87057424-87057424 on Build GRCh38
rs750030628 Chr.10: 87057745-87057745 on Build GRCh38
rs779154751 Chr.10: 87057834-87057834 on Build GRCh38
rs370428834 Chr.10: 87057670-87057670 on Build GRCh38
rs112563956 Chr.10: 87057823-87057823 on Build GRCh38

View Full Product Details